Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26203983-26204481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADHA
Entrez Gene ID
3030
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
UniGene
Hs.516032
Gene Chromosome Location
Chr.2: 26190635-26244726 on Build GRCh38
Transcripts
NM_000182.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4414193 c.1098T>G p.I366M Chr.2: 26204184-26204184 on Build GRCh38
4420700 c.1208A>T p.Q403L Chr.2: 26204074-26204074 on Build GRCh38
5150510 c.1165C>G p.L389V Chr.2: 26204117-26204117 on Build GRCh38
5354100 c.1195C>T p.R399* Chr.2: 26204087-26204087 on Build GRCh38
6157720 c.1168A>G p.K390E Chr.2: 26204114-26204114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748661804 Chr.2: 26204095-26204095 on Build GRCh38
rs200715496 Chr.2: 26204086-26204086 on Build GRCh38
rs760588476 Chr.2: 26204140-26204140 on Build GRCh38
rs377267994 Chr.2: 26204084-26204084 on Build GRCh38
rs139485335 Chr.2: 26204129-26204129 on Build GRCh38
rs1020395662 Chr.2: 26204352-26204352 on Build GRCh38
rs755296795 Chr.2: 26204193-26204193 on Build GRCh38
rs751880451 Chr.2: 26204105-26204105 on Build GRCh38
rs370054250 Chr.2: 26204120-26204120 on Build GRCh38
rs773829713 Chr.2: 26204119-26204119 on Build GRCh38
rs150014280 Chr.2: 26204223-26204223 on Build GRCh38
rs760608614 Chr.2: 26204050-26204050 on Build GRCh38
rs116396996 Chr.2: 26204070-26204070 on Build GRCh38
rs767692992 Chr.2: 26204208-26204208 on Build GRCh38
rs998658096 Chr.2: 26204265-26204265 on Build GRCh38
rs974017529 Chr.2: 26204104-26204104 on Build GRCh38
rs137852770 Chr.2: 26204150-26204150 on Build GRCh38
rs756580741 Chr.2: 26204093-26204093 on Build GRCh38
rs552191498 Chr.2: 26204065-26204065 on Build GRCh38
rs534553558 Chr.2: 26204108-26204108 on Build GRCh38
rs936290489 Chr.2: 26204246-26204246 on Build GRCh38
rs145182116 Chr.2: 26204224-26204224 on Build GRCh38
rs539814267 Chr.2: 26204053-26204053 on Build GRCh38
rs1050693944 Chr.2: 26204430-26204430 on Build GRCh38
rs958911692 Chr.2: 26204405-26204405 on Build GRCh38
rs752875053 Chr.2: 26204222-26204222 on Build GRCh38
rs184302027 Chr.2: 26204354-26204354 on Build GRCh38
rs751667312 Chr.2: 26204161-26204161 on Build GRCh38
rs767080380 Chr.2: 26204031-26204031 on Build GRCh38
rs764067083 Chr.2: 26204141-26204141 on Build GRCh38
rs1006820230 Chr.2: 26204195-26204195 on Build GRCh38
rs978761036 Chr.2: 26204357-26204357 on Build GRCh38
rs991742855 Chr.2: 26204408-26204408 on Build GRCh38
rs770481891 Chr.2: 26204096-26204096 on Build GRCh38
rs199727889 Chr.2: 26204142-26204142 on Build GRCh38
rs1032974573 Chr.2: 26204366-26204366 on Build GRCh38
rs1057517087 Chr.2: 26204080-26204080 on Build GRCh38
rs997033672 Chr.2: 26204043-26204046 on Build GRCh38
rs781338540 Chr.2: 26204094-26204094 on Build GRCh38
rs761474139 Chr.2: 26204153-26204153 on Build GRCh38
rs752570147 Chr.2: 26204032-26204032 on Build GRCh38
rs900047023 Chr.2: 26204317-26204317 on Build GRCh38
rs778404466 Chr.2: 26204199-26204199 on Build GRCh38
rs778263925 Chr.2: 26204098-26204098 on Build GRCh38
rs899495340 Chr.2: 26204006-26204006 on Build GRCh38
rs1057516460 Chr.2: 26204190-26204199 on Build GRCh38
rs780178287 Chr.2: 26204073-26204073 on Build GRCh38
rs765101895 Chr.2: 26204154-26204154 on Build GRCh38
rs768757660 Chr.2: 26204245-26204248 on Build GRCh38
rs749569391 Chr.2: 26204367-26204371 on Build GRCh38

View Full Product Details