Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27094398-27094902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHK
Entrez Gene ID
3795
Gene Name
ketohexokinase
Gene Aliases
-
UniGene
Hs.567297
Gene Chromosome Location
Chr.2: 27086738-27100751 on Build GRCh38
Transcripts
NM_000221.2 NM_006488.2 XM_005264294.3 XM_005264296.3 XM_005264298.3 XM_006712008.3 XM_006712009.3 XM_006712010.3 XM_006712011.3 XM_006712012.3 XM_006712013.3 XM_006712014.3 XM_017004060.1 XM_017004061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5684756 c.236G>T p.R79L Chr.2: 27094826-27094826 on Build GRCh38
6692864 c.312_313insA p.N105fs*10 Chr.2: 27094902-27094903 on Build GRCh38
6091022 c.214G>C p.V72L Chr.2: 27094481-27094481 on Build GRCh38
6192288 c.303C>G p.I101M Chr.2: 27094570-27094570 on Build GRCh38
1494925 c.230G>A p.R77H Chr.2: 27094497-27094497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922525069 Chr.2: 27094808-27094808 on Build GRCh38
rs776109581 Chr.2: 27094511-27094517 on Build GRCh38
rs764363512 Chr.2: 27094631-27094631 on Build GRCh38
rs773599887 Chr.2: 27094644-27094645 on Build GRCh38
rs558348945 Chr.2: 27094695-27094695 on Build GRCh38
rs933725089 Chr.2: 27094430-27094430 on Build GRCh38
rs374146345 Chr.2: 27094456-27094456 on Build GRCh38
rs771957787 Chr.2: 27094573-27094573 on Build GRCh38
rs747821283 Chr.2: 27094457-27094457 on Build GRCh38
rs536698201 Chr.2: 27094688-27094688 on Build GRCh38
rs148370039 Chr.2: 27094560-27094560 on Build GRCh38
rs187413849 Chr.2: 27094526-27094526 on Build GRCh38
rs964100786 Chr.2: 27094783-27094783 on Build GRCh38
rs923190823 Chr.2: 27094719-27094719 on Build GRCh38
rs763131018 Chr.2: 27094518-27094518 on Build GRCh38
rs201650300 Chr.2: 27094601-27094601 on Build GRCh38
rs768121919 Chr.2: 27094449-27094449 on Build GRCh38
rs936569928 Chr.2: 27094624-27094624 on Build GRCh38
rs376933692 Chr.2: 27094650-27094650 on Build GRCh38
rs764917935 Chr.2: 27094786-27094786 on Build GRCh38
rs773548056 Chr.2: 27094588-27094588 on Build GRCh38
rs772905836 Chr.2: 27094846-27094846 on Build GRCh38
rs993845886 Chr.2: 27094505-27094505 on Build GRCh38
rs774508030 Chr.2: 27094592-27094592 on Build GRCh38
rs149502628 Chr.2: 27094850-27094850 on Build GRCh38
rs201070693 Chr.2: 27094658-27094658 on Build GRCh38
rs768550766 Chr.2: 27094752-27094752 on Build GRCh38
rs775285617 Chr.2: 27094583-27094583 on Build GRCh38
rs1046612039 Chr.2: 27094691-27094691 on Build GRCh38
rs922409647 Chr.2: 27094653-27094653 on Build GRCh38
rs376378105 Chr.2: 27094651-27094651 on Build GRCh38
rs761142964 Chr.2: 27094612-27094612 on Build GRCh38
rs764807382 Chr.2: 27094633-27094634 on Build GRCh38
rs753540628 Chr.2: 27094636-27094636 on Build GRCh38
rs760155933 Chr.2: 27094594-27094594 on Build GRCh38
rs1063551 Chr.2: 27094877-27094877 on Build GRCh38
rs771022630 Chr.2: 27094759-27094759 on Build GRCh38
rs978774323 Chr.2: 27094599-27094599 on Build GRCh38
rs138367050 Chr.2: 27094590-27094590 on Build GRCh38
rs762530854 Chr.2: 27094851-27094851 on Build GRCh38
rs775197292 Chr.2: 27094497-27094497 on Build GRCh38
rs183829513 Chr.2: 27094750-27094750 on Build GRCh38
rs200630753 Chr.2: 27094826-27094826 on Build GRCh38
rs766362038 Chr.2: 27094523-27094523 on Build GRCh38
rs895977779 Chr.2: 27094749-27094749 on Build GRCh38
rs748785153 Chr.2: 27094468-27094468 on Build GRCh38
rs756792330 Chr.2: 27094646-27094646 on Build GRCh38
rs756859250 Chr.2: 27094558-27094558 on Build GRCh38
rs1063550 Chr.2: 27094859-27094859 on Build GRCh38
rs764522438 Chr.2: 27094782-27094782 on Build GRCh38
rs965961137 Chr.2: 27094790-27094790 on Build GRCh38
rs765447076 Chr.2: 27094502-27094502 on Build GRCh38
rs1056076303 Chr.2: 27094740-27094740 on Build GRCh38
rs201334254 Chr.2: 27094822-27094822 on Build GRCh38
rs368871098 Chr.2: 27094466-27094466 on Build GRCh38
rs780961189 Chr.2: 27094547-27094547 on Build GRCh38
rs1063552 Chr.2: 27094879-27094879 on Build GRCh38
rs201669686 Chr.2: 27094825-27094825 on Build GRCh38
rs745808103 Chr.2: 27094561-27094561 on Build GRCh38
rs1002194243 Chr.2: 27094716-27094716 on Build GRCh38
rs202153228 Chr.2: 27094555-27094555 on Build GRCh38
rs955166007 Chr.2: 27094810-27094810 on Build GRCh38
rs768299940 Chr.2: 27094586-27094586 on Build GRCh38
rs913305349 Chr.2: 27094593-27094593 on Build GRCh38
rs778425761 Chr.2: 27094648-27094648 on Build GRCh38
rs769613433 Chr.2: 27094824-27094824 on Build GRCh38
rs757966336 Chr.2: 27094801-27094801 on Build GRCh38
rs972489629 Chr.2: 27094696-27094696 on Build GRCh38
rs750042875 Chr.2: 27094797-27094797 on Build GRCh38
rs775231097 Chr.2: 27094831-27094831 on Build GRCh38
rs200737810 Chr.2: 27094748-27094748 on Build GRCh38
rs191575888 Chr.2: 27094708-27094708 on Build GRCh38
rs17850057 Chr.2: 27094501-27094501 on Build GRCh38
rs200568467 Chr.2: 27094819-27094819 on Build GRCh38
rs751976199 Chr.2: 27094445-27094445 on Build GRCh38
rs957958227 Chr.2: 27094829-27094829 on Build GRCh38
rs899413777 Chr.2: 27094474-27094474 on Build GRCh38
rs146252182 Chr.2: 27094837-27094837 on Build GRCh38
rs62128660 Chr.2: 27094767-27094767 on Build GRCh38
rs755488258 Chr.2: 27094531-27094531 on Build GRCh38
rs774386956 Chr.2: 27094492-27094492 on Build GRCh38
rs779523527 Chr.2: 27094811-27094811 on Build GRCh38
rs777173573 Chr.2: 27094836-27094836 on Build GRCh38
rs201995559 Chr.2: 27094496-27094496 on Build GRCh38
rs200318560 Chr.2: 27094508-27094508 on Build GRCh38
rs886055886 Chr.2: 27094517-27094517 on Build GRCh38
rs761073979 Chr.2: 27094779-27094779 on Build GRCh38
rs201170990 Chr.2: 27094812-27094812 on Build GRCh38
rs776207525 Chr.2: 27094774-27094774 on Build GRCh38
rs562937111 Chr.2: 27094840-27094840 on Build GRCh38
rs746852007 Chr.2: 27094584-27094584 on Build GRCh38
rs762051940 Chr.2: 27094499-27094499 on Build GRCh38
rs768002486 Chr.2: 27094541-27094541 on Build GRCh38
rs749532793 Chr.2: 27094755-27094755 on Build GRCh38
rs780795012 Chr.2: 27094753-27094753 on Build GRCh38
rs370270679 Chr.2: 27094784-27094784 on Build GRCh38
rs141417422 Chr.2: 27094589-27094589 on Build GRCh38
rs550720212 Chr.2: 27094830-27094830 on Build GRCh38
rs1046870019 Chr.2: 27094437-27094437 on Build GRCh38
rs761470895 Chr.2: 27094598-27094602 on Build GRCh38
rs774599729 Chr.2: 27094765-27094765 on Build GRCh38
rs1033424024 Chr.2: 27094778-27094778 on Build GRCh38
rs766956071 Chr.2: 27094426-27094426 on Build GRCh38
rs200402287 Chr.2: 27094766-27094766 on Build GRCh38
rs755457721 Chr.2: 27094452-27094452 on Build GRCh38
rs754808415 Chr.2: 27094813-27094813 on Build GRCh38
rs753234761 Chr.2: 27094611-27094611 on Build GRCh38
rs765880895 Chr.2: 27094857-27094857 on Build GRCh38
rs74537742 Chr.2: 27094472-27094472 on Build GRCh38
rs1046816887 Chr.2: 27094676-27094678 on Build GRCh38
rs777180821 Chr.2: 27094467-27094467 on Build GRCh38
rs753075444 Chr.2: 27094546-27094546 on Build GRCh38
rs372511538 Chr.2: 27094785-27094785 on Build GRCh38
rs565695602 Chr.2: 27094556-27094556 on Build GRCh38
rs747581100 Chr.2: 27094832-27094832 on Build GRCh38
rs148679626 Chr.2: 27094863-27094863 on Build GRCh38
rs368277164 Chr.2: 27094871-27094871 on Build GRCh38

View Full Product Details