Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36988599-36989085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR5B
Entrez Gene ID
54497
Gene Name
HEAT repeat containing 5B
Gene Aliases
-
UniGene
Hs.744084
Gene Chromosome Location
Chr.2: 36980999-37084384 on Build GRCh38
Transcripts
NM_019024.2 XM_006712034.2 XM_006712035.3 XM_011532934.2 XM_011532935.2 XM_017004378.1 XM_017004379.1 XM_017004380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1683320 c.5826_5827insC p.A1944fs*3 Chr.2: 36988730-36988731 on Build GRCh38
1167776 c.5762delC p.T1921fs*7 Chr.2: 36988795-36988795 on Build GRCh38
74879 c.5765C>G p.P1922R Chr.2: 36988792-36988792 on Build GRCh38
3910293 c.5765C>T p.P1922L Chr.2: 36988792-36988792 on Build GRCh38
4689824 c.5791G>A p.V1931M Chr.2: 36988766-36988766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886176894 Chr.2: 36988776-36988776 on Build GRCh38
rs992148664 Chr.2: 36988966-36988966 on Build GRCh38
rs374403014 Chr.2: 36988911-36988911 on Build GRCh38
rs1047218378 Chr.2: 36989055-36989055 on Build GRCh38
rs779331420 Chr.2: 36988732-36988732 on Build GRCh38
rs755314889 Chr.2: 36988865-36988865 on Build GRCh38
rs775795139 Chr.2: 36988654-36988654 on Build GRCh38
rs557834104 Chr.2: 36988876-36988878 on Build GRCh38
rs965064700 Chr.2: 36988742-36988742 on Build GRCh38
rs771234566 Chr.2: 36988625-36988625 on Build GRCh38
rs373768266 Chr.2: 36988769-36988769 on Build GRCh38
rs756283655 Chr.2: 36988866-36988868 on Build GRCh38
rs546673365 Chr.2: 36988761-36988761 on Build GRCh38
rs866467549 Chr.2: 36988689-36988689 on Build GRCh38
rs748218112 Chr.2: 36988680-36988682 on Build GRCh38
rs925114294 Chr.2: 36989030-36989030 on Build GRCh38
rs765893925 Chr.2: 36988712-36988712 on Build GRCh38
rs571654596 Chr.2: 36988875-36988875 on Build GRCh38
rs761249432 Chr.2: 36989021-36989021 on Build GRCh38
rs746070399 Chr.2: 36988644-36988644 on Build GRCh38
rs772249009 Chr.2: 36988650-36988650 on Build GRCh38
rs776571673 Chr.2: 36988706-36988706 on Build GRCh38
rs200769161 Chr.2: 36988810-36988810 on Build GRCh38
rs148256206 Chr.2: 36988717-36988717 on Build GRCh38
rs914025396 Chr.2: 36988974-36988974 on Build GRCh38
rs780515071 Chr.2: 36988847-36988847 on Build GRCh38
rs567847253 Chr.2: 36988718-36988718 on Build GRCh38
rs761893882 Chr.2: 36988693-36988693 on Build GRCh38
rs762648818 Chr.2: 36988705-36988705 on Build GRCh38
rs747200626 Chr.2: 36988863-36988863 on Build GRCh38
rs553321359 Chr.2: 36989040-36989040 on Build GRCh38
rs767867410 Chr.2: 36988958-36988960 on Build GRCh38
rs781405171 Chr.2: 36988871-36988871 on Build GRCh38
rs1014319608 Chr.2: 36988802-36988802 on Build GRCh38
rs769925475 Chr.2: 36988886-36988886 on Build GRCh38
rs775485172 Chr.2: 36988772-36988772 on Build GRCh38
rs772656351 Chr.2: 36988890-36988890 on Build GRCh38
rs879331965 Chr.2: 36988747-36988747 on Build GRCh38
rs966779549 Chr.2: 36988994-36988994 on Build GRCh38
rs539089248 Chr.2: 36988923-36988923 on Build GRCh38
rs765096453 Chr.2: 36988699-36988699 on Build GRCh38
rs761834580 Chr.2: 36988763-36988763 on Build GRCh38
rs1039380399 Chr.2: 36989036-36989036 on Build GRCh38
rs926202243 Chr.2: 36989017-36989017 on Build GRCh38
rs768831786 Chr.2: 36988745-36988745 on Build GRCh38
rs933016441 Chr.2: 36989044-36989044 on Build GRCh38
rs751234684 Chr.2: 36989032-36989032 on Build GRCh38
rs1026043894 Chr.2: 36988734-36988734 on Build GRCh38
rs772489807 Chr.2: 36988733-36988733 on Build GRCh38
rs77915442 Chr.2: 36988738-36988738 on Build GRCh38
rs1014246137 Chr.2: 36989049-36989049 on Build GRCh38
rs766983676 Chr.2: 36988807-36988807 on Build GRCh38
rs754322899 Chr.2: 36988729-36988729 on Build GRCh38
rs762569667 Chr.2: 36988958-36988958 on Build GRCh38
rs764797044 Chr.2: 36988884-36988884 on Build GRCh38
rs535356684 Chr.2: 36988740-36988740 on Build GRCh38
rs761660142 Chr.2: 36988819-36988819 on Build GRCh38
rs762591962 Chr.2: 36988775-36988775 on Build GRCh38
rs189376180 Chr.2: 36988950-36988950 on Build GRCh38
rs765595300 Chr.2: 36988838-36988838 on Build GRCh38
rs1038914628 Chr.2: 36988698-36988698 on Build GRCh38
rs759237314 Chr.2: 36988803-36988803 on Build GRCh38
rs767779877 Chr.2: 36989060-36989060 on Build GRCh38
rs765976882 Chr.2: 36989021-36989021 on Build GRCh38
rs779193313 Chr.2: 36988637-36988637 on Build GRCh38
rs751232798 Chr.2: 36988714-36988714 on Build GRCh38
rs199514151 Chr.2: 36988731-36988731 on Build GRCh38
rs143175652 Chr.2: 36988671-36988671 on Build GRCh38
rs369803308 Chr.2: 36989051-36989051 on Build GRCh38
rs751126880 Chr.2: 36988792-36988792 on Build GRCh38
rs777224445 Chr.2: 36988890-36988891 on Build GRCh38
rs750970688 Chr.2: 36988844-36988844 on Build GRCh38
rs760606119 Chr.2: 36988661-36988661 on Build GRCh38
rs750248993 Chr.2: 36988704-36988704 on Build GRCh38
rs749272370 Chr.2: 36988891-36988891 on Build GRCh38
rs879194049 Chr.2: 36988952-36988952 on Build GRCh38
rs191963276 Chr.2: 36988990-36988990 on Build GRCh38
rs766230413 Chr.2: 36988777-36988777 on Build GRCh38
rs921578709 Chr.2: 36988912-36988912 on Build GRCh38
rs912963664 Chr.2: 36988963-36988963 on Build GRCh38
rs987208580 Chr.2: 36988945-36988945 on Build GRCh38
rs961016042 Chr.2: 36988953-36988953 on Build GRCh38
rs776664150 Chr.2: 36988675-36988675 on Build GRCh38
rs759898262 Chr.2: 36988842-36988842 on Build GRCh38
rs752271740 Chr.2: 36988808-36988808 on Build GRCh38

View Full Product Details