Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102026005-102026535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1R2
Entrez Gene ID
7850
Gene Name
interleukin 1 receptor type 2
Gene Aliases
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB
UniGene
Hs.25333
Gene Chromosome Location
Chr.2: 101991805-102028544 on Build GRCh38
Transcripts
NM_004633.3 NR_048564.1 XM_006712734.3 XM_006712736.3 XM_011511801.2 XM_011511803.2 XM_011511804.2 XM_011511805.2 XM_011511808.2 XM_017004889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
399066 c.913T>A p.Y305N Chr.2: 102026136-102026136 on Build GRCh38
3708784 c.896C>T p.S299L Chr.2: 102026119-102026119 on Build GRCh38
4851666 c.919G>A p.E307K Chr.2: 102026142-102026142 on Build GRCh38
1210546 c.946A>G p.T316A Chr.2: 102026169-102026169 on Build GRCh38
2980453 c.1013G>T p.R338L Chr.2: 102026236-102026236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376015781 Chr.2: 102026079-102026079 on Build GRCh38
rs775442025 Chr.2: 102026096-102026096 on Build GRCh38
rs910662097 Chr.2: 102026197-102026197 on Build GRCh38
rs757713985 Chr.2: 102026298-102026298 on Build GRCh38
rs151307899 Chr.2: 102026208-102026208 on Build GRCh38
rs35369567 Chr.2: 102026094-102026094 on Build GRCh38
rs754263352 Chr.2: 102026138-102026148 on Build GRCh38
rs1013745067 Chr.2: 102026306-102026306 on Build GRCh38
rs1031893178 Chr.2: 102026095-102026095 on Build GRCh38
rs976573513 Chr.2: 102026424-102026424 on Build GRCh38
rs775603167 Chr.2: 102026230-102026230 on Build GRCh38
rs957590733 Chr.2: 102026383-102026383 on Build GRCh38
rs1009385508 Chr.2: 102026406-102026406 on Build GRCh38
rs545439323 Chr.2: 102026408-102026408 on Build GRCh38
rs761112510 Chr.2: 102026102-102026103 on Build GRCh38
rs139885028 Chr.2: 102026149-102026149 on Build GRCh38
rs941719784 Chr.2: 102026271-102026271 on Build GRCh38
rs773616121 Chr.2: 102026073-102026073 on Build GRCh38
rs999004196 Chr.2: 102026359-102026359 on Build GRCh38
rs753535615 Chr.2: 102026263-102026263 on Build GRCh38
rs764483579 Chr.2: 102026135-102026135 on Build GRCh38
rs144482163 Chr.2: 102026155-102026155 on Build GRCh38
rs1033651210 Chr.2: 102026497-102026497 on Build GRCh38
rs566880360 Chr.2: 102026148-102026148 on Build GRCh38
rs941023806 Chr.2: 102026329-102026329 on Build GRCh38
rs200467327 Chr.2: 102026138-102026138 on Build GRCh38
rs778071941 Chr.2: 102026067-102026067 on Build GRCh38
rs201541415 Chr.2: 102026300-102026300 on Build GRCh38
rs779567286 Chr.2: 102026163-102026163 on Build GRCh38
rs975973343 Chr.2: 102026209-102026209 on Build GRCh38
rs760333497 Chr.2: 102026260-102026260 on Build GRCh38
rs781410987 Chr.2: 102026203-102026203 on Build GRCh38
rs747569608 Chr.2: 102026199-102026199 on Build GRCh38
rs559960579 Chr.2: 102026388-102026388 on Build GRCh38
rs28362315 Chr.2: 102026264-102026264 on Build GRCh38
rs372920040 Chr.2: 102026272-102026272 on Build GRCh38
rs753377905 Chr.2: 102026170-102026170 on Build GRCh38
rs762540353 Chr.2: 102026404-102026404 on Build GRCh38
rs1002606831 Chr.2: 102026489-102026489 on Build GRCh38
rs3212326 Chr.2: 102026096-102026097 on Build GRCh38
rs1049311722 Chr.2: 102026310-102026310 on Build GRCh38
rs142489461 Chr.2: 102026173-102026173 on Build GRCh38
rs929117282 Chr.2: 102026217-102026217 on Build GRCh38
rs28362314 Chr.2: 102026097-102026097 on Build GRCh38
rs1022148952 Chr.2: 102026441-102026441 on Build GRCh38
rs71378180 Chr.2: 102026103-102026104 on Build GRCh38
rs960991783 Chr.2: 102026508-102026508 on Build GRCh38
rs3218981 Chr.2: 102026405-102026405 on Build GRCh38
rs899622582 Chr.2: 102026112-102026112 on Build GRCh38
rs762938627 Chr.2: 102026102-102026102 on Build GRCh38
rs190822853 Chr.2: 102026049-102026049 on Build GRCh38
rs565578862 Chr.2: 102026456-102026456 on Build GRCh38
rs139357503 Chr.2: 102026236-102026236 on Build GRCh38
rs554382459 Chr.2: 102026253-102026253 on Build GRCh38
rs199726471 Chr.2: 102026301-102026301 on Build GRCh38
rs375568355 Chr.2: 102026076-102026076 on Build GRCh38
rs142449580 Chr.2: 102026063-102026063 on Build GRCh38
rs779498318 Chr.2: 102026060-102026060 on Build GRCh38
rs759741038 Chr.2: 102026077-102026077 on Build GRCh38
rs751993491 Chr.2: 102026285-102026285 on Build GRCh38
rs556845442 Chr.2: 102026351-102026351 on Build GRCh38
rs767276614 Chr.2: 102026246-102026246 on Build GRCh38
rs998422530 Chr.2: 102026115-102026115 on Build GRCh38
rs909106024 Chr.2: 102026270-102026270 on Build GRCh38
rs771277638 Chr.2: 102026201-102026201 on Build GRCh38
rs980819488 Chr.2: 102026216-102026216 on Build GRCh38
rs764160421 Chr.2: 102026113-102026113 on Build GRCh38
rs765916036 Chr.2: 102026261-102026261 on Build GRCh38
rs757535710 Chr.2: 102026294-102026295 on Build GRCh38
rs899507278 Chr.2: 102026357-102026357 on Build GRCh38
rs34916865 Chr.2: 102026399-102026400 on Build GRCh38
rs778229564 Chr.2: 102026176-102026176 on Build GRCh38
rs377633681 Chr.2: 102026282-102026282 on Build GRCh38
rs2072473 Chr.2: 102026033-102026033 on Build GRCh38
rs747340354 Chr.2: 102026072-102026072 on Build GRCh38
rs767882698 Chr.2: 102026096-102026096 on Build GRCh38
rs201396625 Chr.2: 102026234-102026234 on Build GRCh38
rs761679495 Chr.2: 102026139-102026139 on Build GRCh38
rs748621887 Chr.2: 102026062-102026062 on Build GRCh38
rs964860936 Chr.2: 102026407-102026407 on Build GRCh38
rs1039407635 Chr.2: 102026330-102026330 on Build GRCh38
rs888060574 Chr.2: 102026318-102026318 on Build GRCh38
rs201032704 Chr.2: 102026140-102026140 on Build GRCh38
rs553313886 Chr.2: 102026207-102026207 on Build GRCh38
rs147924293 Chr.2: 102026235-102026235 on Build GRCh38
rs997906416 Chr.2: 102026363-102026363 on Build GRCh38
rs114973158 Chr.2: 102026303-102026303 on Build GRCh38

View Full Product Details