Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102337172-102337677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RL1
Entrez Gene ID
9173
Gene Name
interleukin 1 receptor like 1
Gene Aliases
DER4, FIT-1, IL33R, ST2, ST2L, ST2V, T1
UniGene
Hs.66
Gene Chromosome Location
Chr.2: 102311502-102352367 on Build GRCh38
Transcripts
NM_001282408.1 NM_003856.3 NM_016232.4 NR_104167.1 XM_006712839.3 XM_011512151.1

SNP Detail

SNP ID Chromosome Location
rs985318484 Chr.2: 102337275-102337275 on Build GRCh38
rs1010208172 Chr.2: 102337628-102337628 on Build GRCh38
rs371990950 Chr.2: 102337364-102337364 on Build GRCh38
rs995184574 Chr.2: 102337594-102337594 on Build GRCh38
rs939870842 Chr.2: 102337422-102337422 on Build GRCh38
rs1057142127 Chr.2: 102337623-102337623 on Build GRCh38
rs938359056 Chr.2: 102337564-102337564 on Build GRCh38
rs1048753103 Chr.2: 102337613-102337613 on Build GRCh38
rs368103424 Chr.2: 102337213-102337213 on Build GRCh38
rs185771771 Chr.2: 102337317-102337317 on Build GRCh38
rs896851441 Chr.2: 102337212-102337212 on Build GRCh38
rs561815616 Chr.2: 102337411-102337411 on Build GRCh38
rs896820467 Chr.2: 102337559-102337559 on Build GRCh38
rs905502980 Chr.2: 102337222-102337222 on Build GRCh38
rs948192614 Chr.2: 102337506-102337506 on Build GRCh38
rs1021026098 Chr.2: 102337296-102337296 on Build GRCh38
rs1007112559 Chr.2: 102337644-102337644 on Build GRCh38
rs931068422 Chr.2: 102337473-102337473 on Build GRCh38
rs114092839 Chr.2: 102337634-102337634 on Build GRCh38
rs147926215 Chr.2: 102337240-102337240 on Build GRCh38
rs759391248 Chr.2: 102337259-102337259 on Build GRCh38
rs764105310 Chr.2: 102337627-102337627 on Build GRCh38
rs903993760 Chr.2: 102337639-102337639 on Build GRCh38
rs375168088 Chr.2: 102337556-102337556 on Build GRCh38
rs905561589 Chr.2: 102337534-102337534 on Build GRCh38
rs1021099626 Chr.2: 102337633-102337633 on Build GRCh38
rs954035866 Chr.2: 102337388-102337388 on Build GRCh38
rs978176618 Chr.2: 102337321-102337321 on Build GRCh38
rs938396812 Chr.2: 102337540-102337540 on Build GRCh38
rs763141079 Chr.2: 102337610-102337610 on Build GRCh38
rs1020087020 Chr.2: 102337286-102337286 on Build GRCh38
rs928143055 Chr.2: 102337510-102337510 on Build GRCh38
rs7571371 Chr.2: 102337267-102337267 on Build GRCh38
rs919728717 Chr.2: 102337445-102337445 on Build GRCh38
rs1046580220 Chr.2: 102337509-102337509 on Build GRCh38
rs990101734 Chr.2: 102337416-102337416 on Build GRCh38
rs987155027 Chr.2: 102337405-102337405 on Build GRCh38
rs936715418 Chr.2: 102337378-102337378 on Build GRCh38
rs190139999 Chr.2: 102337418-102337418 on Build GRCh38
rs1028285572 Chr.2: 102337341-102337341 on Build GRCh38
rs556539852 Chr.2: 102337208-102337208 on Build GRCh38
rs995413185 Chr.2: 102337237-102337237 on Build GRCh38
rs923935725 Chr.2: 102337376-102337376 on Build GRCh38
rs3771179 Chr.2: 102337432-102337432 on Build GRCh38

View Full Product Details