Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102342902-102343401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RL1
Entrez Gene ID
9173
Gene Name
interleukin 1 receptor like 1
Gene Aliases
DER4, FIT-1, IL33R, ST2, ST2L, ST2V, T1
UniGene
Hs.66
Gene Chromosome Location
Chr.2: 102311502-102352367 on Build GRCh38
Transcripts
NM_001282408.1 NM_003856.3 NM_016232.4 NR_104167.1 XM_006712839.3 XM_011512151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4567923 c.928_929GG>AA p.G310N Chr.2: 102343373-102343374 on Build GRCh38
5681272 c.696C>T p.N232N Chr.2: 102343049-102343049 on Build GRCh38
3564480 c.918G>A p.L306L Chr.2: 102343363-102343363 on Build GRCh38
5665896 c.775A>G p.T259A Chr.2: 102343128-102343128 on Build GRCh38
3894410 c.805G>A p.E269K Chr.2: 102343158-102343158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928387996 Chr.2: 102343338-102343338 on Build GRCh38
rs749350355 Chr.2: 102343017-102343017 on Build GRCh38
rs377734212 Chr.2: 102343192-102343192 on Build GRCh38
rs768009306 Chr.2: 102343287-102343287 on Build GRCh38
rs749714484 Chr.2: 102343013-102343013 on Build GRCh38
rs370287545 Chr.2: 102343181-102343181 on Build GRCh38
rs532769660 Chr.2: 102342999-102342999 on Build GRCh38
rs201949519 Chr.2: 102343209-102343209 on Build GRCh38
rs772006885 Chr.2: 102343053-102343053 on Build GRCh38
rs978330352 Chr.2: 102343037-102343037 on Build GRCh38
rs534062161 Chr.2: 102343241-102343241 on Build GRCh38
rs768766790 Chr.2: 102343027-102343027 on Build GRCh38
rs756486507 Chr.2: 102343102-102343102 on Build GRCh38
rs35298562 Chr.2: 102343318-102343318 on Build GRCh38
rs374285935 Chr.2: 102343191-102343191 on Build GRCh38
rs370167757 Chr.2: 102343219-102343219 on Build GRCh38
rs778910308 Chr.2: 102343109-102343109 on Build GRCh38
rs764994383 Chr.2: 102343160-102343160 on Build GRCh38
rs1027004945 Chr.2: 102342976-102342976 on Build GRCh38
rs370716558 Chr.2: 102343242-102343242 on Build GRCh38
rs769555931 Chr.2: 102343261-102343261 on Build GRCh38
rs150894779 Chr.2: 102343097-102343097 on Build GRCh38
rs756672386 Chr.2: 102343319-102343319 on Build GRCh38
rs773085067 Chr.2: 102343129-102343129 on Build GRCh38
rs774449571 Chr.2: 102343038-102343038 on Build GRCh38
rs755219544 Chr.2: 102343106-102343106 on Build GRCh38
rs754210668 Chr.2: 102343213-102343213 on Build GRCh38
rs752764247 Chr.2: 102343232-102343232 on Build GRCh38
rs896954344 Chr.2: 102342937-102342937 on Build GRCh38
rs199635856 Chr.2: 102343303-102343303 on Build GRCh38
rs780405232 Chr.2: 102343054-102343054 on Build GRCh38
rs368714053 Chr.2: 102343374-102343374 on Build GRCh38
rs200327718 Chr.2: 102342990-102342990 on Build GRCh38
rs140073514 Chr.2: 102343105-102343105 on Build GRCh38
rs781336039 Chr.2: 102343342-102343342 on Build GRCh38
rs75779188 Chr.2: 102343044-102343044 on Build GRCh38
rs760450823 Chr.2: 102343075-102343075 on Build GRCh38
rs756355328 Chr.2: 102343011-102343011 on Build GRCh38
rs939652072 Chr.2: 102342971-102342971 on Build GRCh38
rs768364736 Chr.2: 102343356-102343356 on Build GRCh38
rs772989633 Chr.2: 102343065-102343065 on Build GRCh38
rs770673070 Chr.2: 102343134-102343134 on Build GRCh38
rs979723160 Chr.2: 102343119-102343119 on Build GRCh38
rs931255538 Chr.2: 102343188-102343188 on Build GRCh38
rs539437762 Chr.2: 102343290-102343290 on Build GRCh38
rs141407066 Chr.2: 102343256-102343256 on Build GRCh38
rs777666935 Chr.2: 102343333-102343333 on Build GRCh38
rs781076436 Chr.2: 102343253-102343253 on Build GRCh38
rs750788599 Chr.2: 102343101-102343101 on Build GRCh38
rs760313036 Chr.2: 102342992-102342992 on Build GRCh38
rs773664206 Chr.2: 102343360-102343360 on Build GRCh38
rs562021540 Chr.2: 102343220-102343220 on Build GRCh38
rs958208369 Chr.2: 102343089-102343089 on Build GRCh38
rs775330170 Chr.2: 102343279-102343279 on Build GRCh38
rs1049770509 Chr.2: 102343257-102343257 on Build GRCh38
rs746223192 Chr.2: 102343009-102343009 on Build GRCh38
rs1031024685 Chr.2: 102343283-102343283 on Build GRCh38
rs73004217 Chr.2: 102342998-102342998 on Build GRCh38
rs770481705 Chr.2: 102342988-102342988 on Build GRCh38
rs879599400 Chr.2: 102342957-102342957 on Build GRCh38
rs1050821999 Chr.2: 102342933-102342933 on Build GRCh38
rs759217103 Chr.2: 102343155-102343155 on Build GRCh38
rs953854983 Chr.2: 102343161-102343161 on Build GRCh38
rs768045569 Chr.2: 102343189-102343189 on Build GRCh38
rs62152661 Chr.2: 102343186-102343186 on Build GRCh38
rs905400426 Chr.2: 102343311-102343311 on Build GRCh38
rs1037357069 Chr.2: 102342927-102342927 on Build GRCh38
rs776194504 Chr.2: 102343086-102343086 on Build GRCh38
rs146105446 Chr.2: 102343098-102343098 on Build GRCh38
rs781571426 Chr.2: 102343004-102343004 on Build GRCh38
rs748839614 Chr.2: 102343352-102343352 on Build GRCh38
rs745635768 Chr.2: 102343348-102343348 on Build GRCh38
rs748235808 Chr.2: 102343113-102343113 on Build GRCh38
rs140301260 Chr.2: 102343335-102343335 on Build GRCh38
rs372957266 Chr.2: 102343100-102343100 on Build GRCh38
rs146406137 Chr.2: 102343331-102343331 on Build GRCh38
rs759085241 Chr.2: 102343095-102343095 on Build GRCh38
rs989629022 Chr.2: 102343096-102343096 on Build GRCh38
rs267598807 Chr.2: 102343325-102343325 on Build GRCh38
rs569442080 Chr.2: 102343084-102343084 on Build GRCh38
rs921140270 Chr.2: 102343125-102343125 on Build GRCh38
rs752958696 Chr.2: 102343330-102343330 on Build GRCh38
rs928279716 Chr.2: 102343117-102343117 on Build GRCh38
rs570868780 Chr.2: 102343286-102343286 on Build GRCh38
rs761457425 Chr.2: 102343371-102343371 on Build GRCh38
rs150347157 Chr.2: 102343349-102343349 on Build GRCh38
rs967994109 Chr.2: 102343103-102343103 on Build GRCh38
rs766871179 Chr.2: 102343301-102343301 on Build GRCh38
rs374411011 Chr.2: 102343223-102343223 on Build GRCh38
rs999952587 Chr.2: 102343258-102343258 on Build GRCh38
rs772045141 Chr.2: 102343120-102343120 on Build GRCh38
rs373664545 Chr.2: 102343336-102343336 on Build GRCh38
rs995761400 Chr.2: 102342956-102342956 on Build GRCh38
rs376290660 Chr.2: 102343237-102343237 on Build GRCh38
rs776600848 Chr.2: 102343154-102343154 on Build GRCh38
rs541096350 Chr.2: 102342977-102342977 on Build GRCh38
rs761026559 Chr.2: 102343298-102343298 on Build GRCh38
rs1031682271 Chr.2: 102343077-102343077 on Build GRCh38
rs748033475 Chr.2: 102343040-102343040 on Build GRCh38
rs374124803 Chr.2: 102343250-102343250 on Build GRCh38
rs201461361 Chr.2: 102342993-102342993 on Build GRCh38

View Full Product Details