Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102371875-102372376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18R1
Entrez Gene ID
8809
Gene Name
interleukin 18 receptor 1
Gene Aliases
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
UniGene
Hs.469521
Gene Chromosome Location
Chr.2: 102355817-102398777 on Build GRCh38
Transcripts
NM_001282399.1 NM_003855.3 XM_005264039.3 XM_005264040.3 XM_011512099.1 XM_017005181.1 XM_017005182.1 XM_017005183.1 XM_017005184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960898 c.321G>A p.W107* Chr.2: 102371971-102371971 on Build GRCh38
4914259 c.400A>C p.K134Q Chr.2: 102372050-102372050 on Build GRCh38
5859787 c.416C>T p.T139I Chr.2: 102372066-102372066 on Build GRCh38
1004663 c.401A>C p.K134T Chr.2: 102372051-102372051 on Build GRCh38
1398522 c.406T>G p.F136V Chr.2: 102372056-102372056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011729520 Chr.2: 102372259-102372259 on Build GRCh38
rs754650932 Chr.2: 102372208-102372208 on Build GRCh38
rs1019788925 Chr.2: 102372232-102372232 on Build GRCh38
rs750592389 Chr.2: 102372010-102372010 on Build GRCh38
rs770520562 Chr.2: 102371957-102371957 on Build GRCh38
rs545609877 Chr.2: 102372178-102372178 on Build GRCh38
rs774568545 Chr.2: 102371983-102371983 on Build GRCh38
rs1008847835 Chr.2: 102372220-102372220 on Build GRCh38
rs994224401 Chr.2: 102372294-102372294 on Build GRCh38
rs756145601 Chr.2: 102371931-102371931 on Build GRCh38
rs764729992 Chr.2: 102371981-102371981 on Build GRCh38
rs745445571 Chr.2: 102372136-102372136 on Build GRCh38
rs980067134 Chr.2: 102372351-102372351 on Build GRCh38
rs774878937 Chr.2: 102372153-102372153 on Build GRCh38
rs1031921067 Chr.2: 102372236-102372236 on Build GRCh38
rs904204970 Chr.2: 102372084-102372084 on Build GRCh38
rs956305800 Chr.2: 102372249-102372249 on Build GRCh38
rs758938686 Chr.2: 102372136-102372136 on Build GRCh38
rs776375145 Chr.2: 102372276-102372287 on Build GRCh38
rs202088343 Chr.2: 102371909-102371909 on Build GRCh38
rs776004141 Chr.2: 102372114-102372114 on Build GRCh38
rs748030895 Chr.2: 102371937-102371937 on Build GRCh38
rs34216045 Chr.2: 102372066-102372066 on Build GRCh38
rs762276510 Chr.2: 102371998-102371998 on Build GRCh38
rs750865901 Chr.2: 102371998-102371999 on Build GRCh38
rs890041500 Chr.2: 102372201-102372201 on Build GRCh38
rs201204631 Chr.2: 102372000-102372000 on Build GRCh38
rs778519965 Chr.2: 102372288-102372288 on Build GRCh38
rs758228602 Chr.2: 102372095-102372095 on Build GRCh38
rs1027013074 Chr.2: 102372335-102372335 on Build GRCh38
rs929140233 Chr.2: 102372043-102372043 on Build GRCh38
rs375658526 Chr.2: 102372145-102372145 on Build GRCh38
rs41342249 Chr.2: 102371931-102371931 on Build GRCh38
rs970267722 Chr.2: 102372318-102372318 on Build GRCh38
rs766379820 Chr.2: 102372040-102372040 on Build GRCh38
rs41379550 Chr.2: 102371997-102371997 on Build GRCh38
rs776299372 Chr.2: 102371959-102371959 on Build GRCh38
rs778565125 Chr.2: 102371935-102371935 on Build GRCh38
rs763302680 Chr.2: 102371979-102371979 on Build GRCh38
rs778956300 Chr.2: 102372065-102372065 on Build GRCh38
rs980887341 Chr.2: 102372337-102372337 on Build GRCh38
rs771826503 Chr.2: 102371940-102371940 on Build GRCh38
rs756338040 Chr.2: 102372034-102372034 on Build GRCh38
rs73006117 Chr.2: 102371918-102371918 on Build GRCh38
rs773041896 Chr.2: 102371948-102371948 on Build GRCh38
rs781461957 Chr.2: 102372060-102372060 on Build GRCh38
rs1033427547 Chr.2: 102372342-102372342 on Build GRCh38
rs562692326 Chr.2: 102372347-102372347 on Build GRCh38
rs770716680 Chr.2: 102372112-102372112 on Build GRCh38
rs201106563 Chr.2: 102371932-102371932 on Build GRCh38
rs754016574 Chr.2: 102372052-102372052 on Build GRCh38
rs1008028202 Chr.2: 102372085-102372085 on Build GRCh38
rs540194919 Chr.2: 102371949-102371949 on Build GRCh38
rs901806108 Chr.2: 102372169-102372169 on Build GRCh38
rs776296491 Chr.2: 102372316-102372318 on Build GRCh38
rs746601953 Chr.2: 102372098-102372098 on Build GRCh38
rs200049313 Chr.2: 102371929-102371929 on Build GRCh38

View Full Product Details