Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102384746-102385251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18R1
Entrez Gene ID
8809
Gene Name
interleukin 18 receptor 1
Gene Aliases
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
UniGene
Hs.469521
Gene Chromosome Location
Chr.2: 102355817-102398777 on Build GRCh38
Transcripts
NM_001282399.1 NM_003855.3 XM_005264039.3 XM_005264040.3 XM_011512099.1 XM_017005181.1 XM_017005182.1 XM_017005183.1 XM_017005184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5514870 c.809+6T>C p.? Chr.2: 102385004-102385004 on Build GRCh38
3564508 c.796G>A p.E266K Chr.2: 102384985-102384985 on Build GRCh38
3564507 c.770C>T p.S257L Chr.2: 102384959-102384959 on Build GRCh38
3564506 c.757G>A p.E253K Chr.2: 102384946-102384946 on Build GRCh38
440915 c.696C>T p.N232N Chr.2: 102384885-102384885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771995155 Chr.2: 102384891-102384891 on Build GRCh38
rs773707267 Chr.2: 102385043-102385043 on Build GRCh38
rs550798512 Chr.2: 102384809-102384809 on Build GRCh38
rs1033505123 Chr.2: 102385122-102385122 on Build GRCh38
rs992860517 Chr.2: 102384812-102384812 on Build GRCh38
rs377457925 Chr.2: 102384959-102384959 on Build GRCh38
rs749498728 Chr.2: 102384911-102384911 on Build GRCh38
rs752232575 Chr.2: 102384838-102384838 on Build GRCh38
rs374788069 Chr.2: 102384943-102384943 on Build GRCh38
rs918178947 Chr.2: 102384821-102384821 on Build GRCh38
rs150063521 Chr.2: 102384960-102384960 on Build GRCh38
rs768409809 Chr.2: 102385042-102385042 on Build GRCh38
rs929906738 Chr.2: 102385172-102385172 on Build GRCh38
rs1019877524 Chr.2: 102385063-102385065 on Build GRCh38
rs745840990 Chr.2: 102385019-102385019 on Build GRCh38
rs569732144 Chr.2: 102385022-102385022 on Build GRCh38
rs933732430 Chr.2: 102385007-102385007 on Build GRCh38
rs997892018 Chr.2: 102384792-102384792 on Build GRCh38
rs1035130 Chr.2: 102384942-102384942 on Build GRCh38
rs148457935 Chr.2: 102384884-102384884 on Build GRCh38
rs768005869 Chr.2: 102385183-102385183 on Build GRCh38
rs888323386 Chr.2: 102385225-102385225 on Build GRCh38
rs1009675414 Chr.2: 102385077-102385077 on Build GRCh38
rs958192447 Chr.2: 102385129-102385129 on Build GRCh38
rs61731174 Chr.2: 102384967-102384967 on Build GRCh38
rs892163704 Chr.2: 102384835-102384835 on Build GRCh38
rs181578441 Chr.2: 102385168-102385168 on Build GRCh38
rs780559206 Chr.2: 102384883-102384883 on Build GRCh38
rs985175043 Chr.2: 102385182-102385182 on Build GRCh38
rs76660141 Chr.2: 102384979-102384979 on Build GRCh38
rs11465645 Chr.2: 102385018-102385018 on Build GRCh38
rs967893307 Chr.2: 102384865-102384865 on Build GRCh38
rs892111523 Chr.2: 102385052-102385052 on Build GRCh38
rs184063962 Chr.2: 102385110-102385110 on Build GRCh38
rs746925277 Chr.2: 102384851-102384858 on Build GRCh38
rs770833967 Chr.2: 102385014-102385014 on Build GRCh38
rs752802414 Chr.2: 102384981-102384981 on Build GRCh38
rs11465647 Chr.2: 102385096-102385096 on Build GRCh38
rs1043411283 Chr.2: 102385082-102385082 on Build GRCh38
rs764852813 Chr.2: 102385023-102385023 on Build GRCh38
rs758653418 Chr.2: 102384985-102384985 on Build GRCh38
rs188483839 Chr.2: 102384912-102384912 on Build GRCh38
rs11465643 Chr.2: 102384834-102384834 on Build GRCh38
rs11465644 Chr.2: 102384885-102384885 on Build GRCh38
rs944085615 Chr.2: 102385145-102385145 on Build GRCh38
rs763458027 Chr.2: 102384958-102384958 on Build GRCh38
rs756690082 Chr.2: 102384872-102384872 on Build GRCh38
rs776417898 Chr.2: 102384827-102384827 on Build GRCh38
rs746856515 Chr.2: 102384910-102384910 on Build GRCh38
rs959726256 Chr.2: 102384798-102384798 on Build GRCh38
rs1033531106 Chr.2: 102384868-102384868 on Build GRCh38
rs762504755 Chr.2: 102385031-102385031 on Build GRCh38
rs188808538 Chr.2: 102385167-102385167 on Build GRCh38
rs750850460 Chr.2: 102384863-102384863 on Build GRCh38
rs899571383 Chr.2: 102384780-102384780 on Build GRCh38
rs3752659 Chr.2: 102385138-102385138 on Build GRCh38
rs56151680 Chr.2: 102385130-102385130 on Build GRCh38
rs776397632 Chr.2: 102384915-102384915 on Build GRCh38
rs980983838 Chr.2: 102384867-102384867 on Build GRCh38
rs769853872 Chr.2: 102384906-102384932 on Build GRCh38
rs925430867 Chr.2: 102384866-102384866 on Build GRCh38
rs775372066 Chr.2: 102385029-102385029 on Build GRCh38
rs200122759 Chr.2: 102385001-102385001 on Build GRCh38
rs770010642 Chr.2: 102384801-102384801 on Build GRCh38
rs150066880 Chr.2: 102384851-102384851 on Build GRCh38
rs374004770 Chr.2: 102384965-102384965 on Build GRCh38
rs769408772 Chr.2: 102384934-102384934 on Build GRCh38
rs546758907 Chr.2: 102385186-102385186 on Build GRCh38
rs377639309 Chr.2: 102384988-102384988 on Build GRCh38
rs779355793 Chr.2: 102384975-102384975 on Build GRCh38
rs909220113 Chr.2: 102385184-102385184 on Build GRCh38
rs17651485 Chr.2: 102385190-102385190 on Build GRCh38
rs200811959 Chr.2: 102384886-102384886 on Build GRCh38
rs777921680 Chr.2: 102384892-102384892 on Build GRCh38
rs1013931455 Chr.2: 102385057-102385057 on Build GRCh38
rs759376271 Chr.2: 102384921-102384921 on Build GRCh38
rs757928874 Chr.2: 102385123-102385123 on Build GRCh38
rs866580151 Chr.2: 102384955-102384955 on Build GRCh38
rs11465646 Chr.2: 102385063-102385063 on Build GRCh38
rs769531549 Chr.2: 102385024-102385024 on Build GRCh38
rs879652844 Chr.2: 102385152-102385153 on Build GRCh38
rs200874450 Chr.2: 102384848-102384848 on Build GRCh38
rs567351994 Chr.2: 102384810-102384810 on Build GRCh38

View Full Product Details