Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102389938-102390476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18R1
Entrez Gene ID
8809
Gene Name
interleukin 18 receptor 1
Gene Aliases
CD218a, CDw218a, IL-1Rrp, IL18RA, IL1RRP
UniGene
Hs.469521
Gene Chromosome Location
Chr.2: 102355817-102398777 on Build GRCh38
Transcripts
NM_001282399.1 NM_003855.3 XM_005264039.3 XM_005264040.3 XM_011512099.1 XM_017005181.1 XM_017005182.1 XM_017005183.1 XM_017005184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1004666 c.961G>T p.D321Y Chr.2: 102390067-102390067 on Build GRCh38
4963889 c.983C>T p.T328I Chr.2: 102390089-102390089 on Build GRCh38
5454867 c.1019C>T p.A340V Chr.2: 102390125-102390125 on Build GRCh38
7083192 c.1069C>T p.L357L Chr.2: 102390175-102390175 on Build GRCh38
4563720 c.986G>C p.R329T Chr.2: 102390092-102390092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760947382 Chr.2: 102390269-102390269 on Build GRCh38
rs758677366 Chr.2: 102390073-102390073 on Build GRCh38
rs773431854 Chr.2: 102390104-102390104 on Build GRCh38
rs1041043265 Chr.2: 102389995-102389995 on Build GRCh38
rs746672963 Chr.2: 102390064-102390064 on Build GRCh38
rs762201530 Chr.2: 102390095-102390095 on Build GRCh38
rs1000721131 Chr.2: 102390313-102390313 on Build GRCh38
rs758101806 Chr.2: 102390061-102390061 on Build GRCh38
rs952649690 Chr.2: 102390323-102390323 on Build GRCh38
rs556979740 Chr.2: 102390074-102390074 on Build GRCh38
rs1025487494 Chr.2: 102390307-102390307 on Build GRCh38
rs912211172 Chr.2: 102390012-102390012 on Build GRCh38
rs772362542 Chr.2: 102390232-102390232 on Build GRCh38
rs370858300 Chr.2: 102390040-102390040 on Build GRCh38
rs758228666 Chr.2: 102390167-102390167 on Build GRCh38
rs764314997 Chr.2: 102390081-102390081 on Build GRCh38
rs752509428 Chr.2: 102390145-102390145 on Build GRCh38
rs772155555 Chr.2: 102390218-102390219 on Build GRCh38
rs868564603 Chr.2: 102390190-102390190 on Build GRCh38
rs564527674 Chr.2: 102390452-102390452 on Build GRCh38
rs562395917 Chr.2: 102390346-102390346 on Build GRCh38
rs11465652 Chr.2: 102390231-102390231 on Build GRCh38
rs765315093 Chr.2: 102390138-102390138 on Build GRCh38
rs1028553222 Chr.2: 102390320-102390320 on Build GRCh38
rs760819708 Chr.2: 102390115-102390115 on Build GRCh38
rs778945671 Chr.2: 102390348-102390348 on Build GRCh38
rs187081342 Chr.2: 102390194-102390194 on Build GRCh38
rs769222530 Chr.2: 102390082-102390082 on Build GRCh38
rs371019881 Chr.2: 102390195-102390195 on Build GRCh38
rs752457819 Chr.2: 102390013-102390013 on Build GRCh38
rs374245841 Chr.2: 102390126-102390126 on Build GRCh38
rs760765603 Chr.2: 102390030-102390030 on Build GRCh38
rs541692501 Chr.2: 102390391-102390391 on Build GRCh38
rs745431033 Chr.2: 102390203-102390203 on Build GRCh38
rs1009939975 Chr.2: 102390299-102390299 on Build GRCh38
rs777733568 Chr.2: 102390168-102390168 on Build GRCh38
rs922614534 Chr.2: 102390441-102390441 on Build GRCh38
rs149204147 Chr.2: 102390063-102390063 on Build GRCh38
rs753861064 Chr.2: 102390199-102390199 on Build GRCh38
rs143336529 Chr.2: 102390103-102390103 on Build GRCh38
rs983881137 Chr.2: 102390329-102390329 on Build GRCh38
rs960000932 Chr.2: 102390270-102390270 on Build GRCh38
rs377144148 Chr.2: 102389989-102389989 on Build GRCh38
rs374553958 Chr.2: 102390202-102390202 on Build GRCh38
rs6749014 Chr.2: 102389988-102389988 on Build GRCh38
rs752599188 Chr.2: 102390054-102390054 on Build GRCh38
rs769116542 Chr.2: 102390214-102390214 on Build GRCh38
rs773342417 Chr.2: 102390357-102390357 on Build GRCh38
rs557522969 Chr.2: 102390014-102390014 on Build GRCh38
rs972665671 Chr.2: 102390291-102390291 on Build GRCh38
rs977264580 Chr.2: 102390383-102390383 on Build GRCh38
rs779348174 Chr.2: 102390226-102390226 on Build GRCh38
rs371466872 Chr.2: 102390267-102390267 on Build GRCh38
rs774672103 Chr.2: 102390090-102390090 on Build GRCh38
rs745758703 Chr.2: 102390094-102390094 on Build GRCh38
rs778846346 Chr.2: 102390044-102390044 on Build GRCh38
rs953749641 Chr.2: 102389970-102389970 on Build GRCh38
rs748407275 Chr.2: 102390421-102390421 on Build GRCh38
rs139664529 Chr.2: 102390036-102390036 on Build GRCh38
rs778003138 Chr.2: 102390021-102390023 on Build GRCh38
rs766413944 Chr.2: 102390123-102390123 on Build GRCh38
rs759503831 Chr.2: 102390127-102390127 on Build GRCh38
rs149700362 Chr.2: 102390380-102390380 on Build GRCh38
rs903736556 Chr.2: 102390308-102390308 on Build GRCh38
rs750466609 Chr.2: 102390022-102390022 on Build GRCh38

View Full Product Details