Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102423636-102424145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18RAP
Entrez Gene ID
8807
Gene Name
interleukin 18 receptor accessory protein
Gene Aliases
ACPL, CD218b, CDw218b, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1R-7, IL-1R7, IL-1RAcPL, IL18RB
UniGene
Hs.158315
Gene Chromosome Location
Chr.2: 102418551-102452568 on Build GRCh38
Transcripts
NM_003853.3 XM_011512087.2 XM_011512088.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4544257 c.352G>A p.V118M Chr.2: 102424092-102424092 on Build GRCh38
221952 c.181C>T p.R61* Chr.2: 102423921-102423921 on Build GRCh38
714932 c.376T>A p.C126S Chr.2: 102424116-102424116 on Build GRCh38
7276701 c.80_81delCA p.T27fs*12 Chr.2: 102423820-102423821 on Build GRCh38
5532653 c.331C>T p.L111F Chr.2: 102424071-102424071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753851163 Chr.2: 102423857-102423857 on Build GRCh38
rs1031452014 Chr.2: 102424070-102424070 on Build GRCh38
rs776909502 Chr.2: 102424061-102424061 on Build GRCh38
rs756319084 Chr.2: 102423821-102423821 on Build GRCh38
rs143234495 Chr.2: 102424018-102424018 on Build GRCh38
rs6759588 Chr.2: 102423699-102423699 on Build GRCh38
rs377337034 Chr.2: 102424065-102424065 on Build GRCh38
rs145032053 Chr.2: 102424094-102424094 on Build GRCh38
rs767014153 Chr.2: 102424103-102424103 on Build GRCh38
rs767626133 Chr.2: 102423827-102423827 on Build GRCh38
rs547962493 Chr.2: 102424082-102424082 on Build GRCh38
rs759560607 Chr.2: 102423975-102423975 on Build GRCh38
rs369898608 Chr.2: 102423867-102423867 on Build GRCh38
rs775331372 Chr.2: 102424085-102424085 on Build GRCh38
rs778856475 Chr.2: 102423882-102423882 on Build GRCh38
rs774512653 Chr.2: 102423815-102423815 on Build GRCh38
rs765304854 Chr.2: 102424081-102424081 on Build GRCh38
rs199952525 Chr.2: 102424088-102424088 on Build GRCh38
rs867163754 Chr.2: 102423921-102423921 on Build GRCh38
rs761536970 Chr.2: 102424092-102424092 on Build GRCh38
rs772226393 Chr.2: 102423960-102423960 on Build GRCh38
rs9807934 Chr.2: 102423722-102423722 on Build GRCh38
rs869042654 Chr.2: 102423793-102423794 on Build GRCh38
rs965434986 Chr.2: 102423942-102423942 on Build GRCh38
rs970437397 Chr.2: 102423885-102423885 on Build GRCh38
rs770263971 Chr.2: 102423785-102423785 on Build GRCh38
rs748401163 Chr.2: 102423957-102423957 on Build GRCh38
rs777152543 Chr.2: 102423774-102423774 on Build GRCh38
rs955890703 Chr.2: 102423878-102423878 on Build GRCh38
rs1028469626 Chr.2: 102423778-102423778 on Build GRCh38
rs1049415338 Chr.2: 102424039-102424039 on Build GRCh38
rs752605497 Chr.2: 102423992-102423992 on Build GRCh38
rs773267891 Chr.2: 102424050-102424050 on Build GRCh38
rs765110724 Chr.2: 102423979-102423979 on Build GRCh38
rs746383375 Chr.2: 102423776-102423776 on Build GRCh38
rs773321602 Chr.2: 102423963-102423963 on Build GRCh38
rs764677669 Chr.2: 102423971-102423971 on Build GRCh38
rs1014104606 Chr.2: 102424106-102424106 on Build GRCh38
rs771076732 Chr.2: 102424060-102424060 on Build GRCh38
rs398104566 Chr.2: 102423788-102423789 on Build GRCh38
rs766278345 Chr.2: 102423851-102423851 on Build GRCh38
rs143062976 Chr.2: 102423721-102423721 on Build GRCh38
rs762023114 Chr.2: 102423823-102423823 on Build GRCh38
rs920641913 Chr.2: 102423953-102423953 on Build GRCh38
rs774759814 Chr.2: 102423947-102423947 on Build GRCh38
rs371728977 Chr.2: 102423801-102423801 on Build GRCh38
rs936895996 Chr.2: 102423677-102423677 on Build GRCh38
rs751866033 Chr.2: 102423785-102423786 on Build GRCh38
rs3215320 Chr.2: 102423792-102423793 on Build GRCh38
rs756620639 Chr.2: 102423922-102423922 on Build GRCh38
rs780487193 Chr.2: 102423926-102423926 on Build GRCh38
rs924088939 Chr.2: 102423664-102423664 on Build GRCh38
rs750538918 Chr.2: 102423837-102423837 on Build GRCh38
rs758037243 Chr.2: 102423995-102423995 on Build GRCh38
rs777331337 Chr.2: 102423911-102423911 on Build GRCh38
rs565679467 Chr.2: 102424028-102424028 on Build GRCh38
rs989581110 Chr.2: 102424067-102424067 on Build GRCh38
rs1026827083 Chr.2: 102423814-102423814 on Build GRCh38
rs749425651 Chr.2: 102424069-102424069 on Build GRCh38
rs758043204 Chr.2: 102423887-102423887 on Build GRCh38
rs113634441 Chr.2: 102424013-102424013 on Build GRCh38
rs752339778 Chr.2: 102423884-102423884 on Build GRCh38
rs11377261 Chr.2: 102423787-102423788 on Build GRCh38
rs113314735 Chr.2: 102423955-102423955 on Build GRCh38
rs751244914 Chr.2: 102424006-102424006 on Build GRCh38
rs201832137 Chr.2: 102424091-102424091 on Build GRCh38
rs755458821 Chr.2: 102424117-102424117 on Build GRCh38
rs747260933 Chr.2: 102424053-102424053 on Build GRCh38
rs746539787 Chr.2: 102423913-102423913 on Build GRCh38
rs201039117 Chr.2: 102423789-102423790 on Build GRCh38
rs760633536 Chr.2: 102423850-102423850 on Build GRCh38
rs145458658 Chr.2: 102423939-102423939 on Build GRCh38
rs376556225 Chr.2: 102423789-102423789 on Build GRCh38
rs772510649 Chr.2: 102424043-102424043 on Build GRCh38
rs11465694 Chr.2: 102424007-102424007 on Build GRCh38
rs749979341 Chr.2: 102424109-102424109 on Build GRCh38
rs777947995 Chr.2: 102423890-102423909 on Build GRCh38
rs924180042 Chr.2: 102424029-102424029 on Build GRCh38
rs149062947 Chr.2: 102423695-102423695 on Build GRCh38
rs374476221 Chr.2: 102424025-102424025 on Build GRCh38
rs1047891368 Chr.2: 102423705-102423705 on Build GRCh38
rs2272129 Chr.2: 102423789-102423789 on Build GRCh38
rs1003190722 Chr.2: 102423719-102423719 on Build GRCh38
rs752348672 Chr.2: 102423972-102423972 on Build GRCh38
rs763700480 Chr.2: 102423996-102423996 on Build GRCh38
rs6543133 Chr.2: 102423717-102423717 on Build GRCh38
rs138870268 Chr.2: 102423689-102423689 on Build GRCh38
rs760624664 Chr.2: 102423968-102423968 on Build GRCh38
rs752896941 Chr.2: 102423791-102423793 on Build GRCh38
rs62154968 Chr.2: 102424037-102424037 on Build GRCh38
rs780681353 Chr.2: 102424017-102424017 on Build GRCh38
rs11465689 Chr.2: 102423707-102423707 on Build GRCh38
rs1044017953 Chr.2: 102423710-102423710 on Build GRCh38
rs768938502 Chr.2: 102423943-102423943 on Build GRCh38
rs866477583 Chr.2: 102424083-102424083 on Build GRCh38

View Full Product Details