Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162371601-162372097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCA
Entrez Gene ID
25801
Gene Name
grancalcin
Gene Aliases
GCL
UniGene
Hs.377894
Gene Chromosome Location
Chr.2: 162318768-162371605 on Build GRCh38
Transcripts
XM_005246446.2 XM_006712398.3 XM_006712400.3 XM_011510926.2 XM_011510928.2 XM_017003767.1 NM_033272.3 XM_011512109.2 XM_017005218.1 XM_017005219.1 XM_017005220.1

Gene Symbol
KCNH7
Entrez Gene ID
90134
Gene Name
potassium voltage-gated channel subfamily H member 7
Gene Aliases
ERG3, HERG3, Kv11.3
UniGene
Hs.657413
Gene Chromosome Location
Chr.2: 162371407-162838747 on Build GRCh38
Transcripts
XM_005246446.2 XM_006712398.3 XM_006712400.3 XM_011510926.2 XM_011510928.2 XM_017003767.1 NM_033272.3 XM_011512109.2 XM_017005218.1 XM_017005219.1 XM_017005220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3568538 c.3539G>A p.G1180E Chr.2: 162371881-162371881 on Build GRCh38
5437127 c.3487G>A p.V1163I Chr.2: 162371933-162371933 on Build GRCh38
5922167 c.3584G>A p.G1195E Chr.2: 162371836-162371836 on Build GRCh38
1564733 c.3486C>T p.Y1162Y Chr.2: 162371934-162371934 on Build GRCh38
4340464 c.3470G>A p.R1157Q Chr.2: 162371950-162371950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758195799 Chr.2: 162371916-162371916 on Build GRCh38
rs534785053 Chr.2: 162371659-162371659 on Build GRCh38
rs372831112 Chr.2: 162371951-162371951 on Build GRCh38
rs942531071 Chr.2: 162371884-162371884 on Build GRCh38
rs200298014 Chr.2: 162371861-162371861 on Build GRCh38
rs900545594 Chr.2: 162371624-162371624 on Build GRCh38
rs766264346 Chr.2: 162371923-162371923 on Build GRCh38
rs748046524 Chr.2: 162371934-162371934 on Build GRCh38
rs775263383 Chr.2: 162371938-162371938 on Build GRCh38
rs909768655 Chr.2: 162371877-162371877 on Build GRCh38
rs769428413 Chr.2: 162372004-162372004 on Build GRCh38
rs1019306743 Chr.2: 162371936-162371936 on Build GRCh38
rs573562870 Chr.2: 162371891-162371891 on Build GRCh38
rs750125664 Chr.2: 162371804-162371804 on Build GRCh38
rs557313177 Chr.2: 162371641-162371641 on Build GRCh38
rs751204689 Chr.2: 162372028-162372028 on Build GRCh38
rs142210415 Chr.2: 162371933-162371933 on Build GRCh38
rs149607273 Chr.2: 162372040-162372040 on Build GRCh38
rs373993413 Chr.2: 162371814-162371814 on Build GRCh38
rs755000626 Chr.2: 162371838-162371838 on Build GRCh38
rs183509596 Chr.2: 162371728-162371728 on Build GRCh38
rs774496305 Chr.2: 162371852-162371852 on Build GRCh38
rs775977072 Chr.2: 162371889-162371889 on Build GRCh38
rs756827076 Chr.2: 162371793-162371793 on Build GRCh38
rs370636080 Chr.2: 162371894-162371894 on Build GRCh38
rs78734560 Chr.2: 162371955-162371955 on Build GRCh38
rs776649582 Chr.2: 162371810-162371810 on Build GRCh38
rs746917781 Chr.2: 162371834-162371834 on Build GRCh38
rs764856967 Chr.2: 162371905-162371905 on Build GRCh38
rs756102525 Chr.2: 162372044-162372044 on Build GRCh38
rs770713731 Chr.2: 162371729-162371729 on Build GRCh38
rs149916560 Chr.2: 162371932-162371932 on Build GRCh38
rs762674054 Chr.2: 162372011-162372011 on Build GRCh38
rs573163082 Chr.2: 162371893-162371893 on Build GRCh38
rs1057508923 Chr.2: 162371734-162371734 on Build GRCh38
rs1005044259 Chr.2: 162371817-162371817 on Build GRCh38
rs779617348 Chr.2: 162371791-162371791 on Build GRCh38
rs969305335 Chr.2: 162371765-162371765 on Build GRCh38
rs764940868 Chr.2: 162371803-162371803 on Build GRCh38
rs768921396 Chr.2: 162371971-162371971 on Build GRCh38
rs377226541 Chr.2: 162372030-162372030 on Build GRCh38
rs927375670 Chr.2: 162371972-162371972 on Build GRCh38
rs374095828 Chr.2: 162371950-162371950 on Build GRCh38
rs1010515194 Chr.2: 162371722-162371722 on Build GRCh38
rs940951599 Chr.2: 162372046-162372046 on Build GRCh38
rs777140982 Chr.2: 162371983-162371983 on Build GRCh38
rs956010553 Chr.2: 162371720-162371720 on Build GRCh38
rs777033667 Chr.2: 162371898-162371898 on Build GRCh38
rs771839703 Chr.2: 162371642-162371642 on Build GRCh38
rs187984820 Chr.2: 162371839-162371839 on Build GRCh38
rs1030372118 Chr.2: 162371694-162371694 on Build GRCh38
rs753432481 Chr.2: 162371783-162371783 on Build GRCh38
rs778936991 Chr.2: 162372069-162372069 on Build GRCh38
rs929622373 Chr.2: 162371860-162371860 on Build GRCh38
rs761360521 Chr.2: 162371901-162371901 on Build GRCh38
rs144025747 Chr.2: 162371876-162371876 on Build GRCh38
rs771019746 Chr.2: 162371850-162371850 on Build GRCh38
rs777608961 Chr.2: 162371670-162371670 on Build GRCh38
rs772919279 Chr.2: 162372010-162372010 on Build GRCh38
rs886918307 Chr.2: 162371975-162371975 on Build GRCh38
rs947449815 Chr.2: 162371716-162371716 on Build GRCh38
rs750033552 Chr.2: 162371906-162371906 on Build GRCh38
rs201273498 Chr.2: 162371767-162371767 on Build GRCh38
rs980257719 Chr.2: 162371781-162371781 on Build GRCh38
rs749426625 Chr.2: 162371847-162371847 on Build GRCh38
rs913353313 Chr.2: 162371683-162371683 on Build GRCh38
rs766096984 Chr.2: 162372027-162372027 on Build GRCh38
rs762417294 Chr.2: 162371986-162371986 on Build GRCh38
rs147843561 Chr.2: 162371952-162371952 on Build GRCh38
rs150975356 Chr.2: 162371778-162371779 on Build GRCh38
rs918278123 Chr.2: 162371853-162371853 on Build GRCh38

View Full Product Details