Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162400053-162400556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH7
Entrez Gene ID
90134
Gene Name
potassium voltage-gated channel subfamily H member 7
Gene Aliases
ERG3, HERG3, Kv11.3
UniGene
Hs.657413
Gene Chromosome Location
Chr.2: 162371407-162838747 on Build GRCh38
Transcripts
NM_033272.3 XM_011512109.2 XM_017005218.1 XM_017005219.1 XM_017005220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5686063 c.2213C>A p.T738K Chr.2: 162400383-162400383 on Build GRCh38
4189067 c.2234C>T p.A745V Chr.2: 162400362-162400362 on Build GRCh38
6154556 c.2360G>T p.R787I Chr.2: 162400236-162400236 on Build GRCh38
1613581 c.2227T>A p.C743S Chr.2: 162400369-162400369 on Build GRCh38
5930916 c.2303C>T p.P768L Chr.2: 162400293-162400293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145691395 Chr.2: 162400456-162400456 on Build GRCh38
rs762884891 Chr.2: 162400238-162400238 on Build GRCh38
rs770485337 Chr.2: 162400143-162400143 on Build GRCh38
rs760878823 Chr.2: 162400436-162400436 on Build GRCh38
rs199733546 Chr.2: 162400357-162400357 on Build GRCh38
rs757276514 Chr.2: 162400350-162400351 on Build GRCh38
rs767811837 Chr.2: 162400430-162400430 on Build GRCh38
rs34590296 Chr.2: 162400270-162400271 on Build GRCh38
rs868752914 Chr.2: 162400213-162400213 on Build GRCh38
rs778712528 Chr.2: 162400447-162400448 on Build GRCh38
rs760572122 Chr.2: 162400207-162400207 on Build GRCh38
rs577155444 Chr.2: 162400470-162400470 on Build GRCh38
rs768511699 Chr.2: 162400375-162400375 on Build GRCh38
rs746679682 Chr.2: 162400486-162400486 on Build GRCh38
rs112706514 Chr.2: 162400222-162400222 on Build GRCh38
rs779625568 Chr.2: 162400482-162400482 on Build GRCh38
rs759898629 Chr.2: 162400274-162400274 on Build GRCh38
rs773807112 Chr.2: 162400147-162400147 on Build GRCh38
rs139632449 Chr.2: 162400215-162400215 on Build GRCh38
rs771839647 Chr.2: 162400166-162400166 on Build GRCh38
rs756713019 Chr.2: 162400302-162400302 on Build GRCh38
rs775312379 Chr.2: 162400193-162400193 on Build GRCh38
rs953413757 Chr.2: 162400100-162400100 on Build GRCh38
rs745497316 Chr.2: 162400342-162400342 on Build GRCh38
rs934107690 Chr.2: 162400171-162400171 on Build GRCh38
rs76459697 Chr.2: 162400534-162400534 on Build GRCh38
rs866203323 Chr.2: 162400308-162400308 on Build GRCh38
rs376541522 Chr.2: 162400432-162400432 on Build GRCh38
rs559866535 Chr.2: 162400376-162400376 on Build GRCh38
rs371886389 Chr.2: 162400394-162400394 on Build GRCh38
rs757925463 Chr.2: 162400477-162400477 on Build GRCh38
rs748710544 Chr.2: 162400138-162400138 on Build GRCh38
rs990308361 Chr.2: 162400355-162400355 on Build GRCh38
rs995330302 Chr.2: 162400084-162400084 on Build GRCh38
rs898359706 Chr.2: 162400077-162400077 on Build GRCh38
rs1045380146 Chr.2: 162400384-162400384 on Build GRCh38
rs751827337 Chr.2: 162400269-162400269 on Build GRCh38
rs761876733 Chr.2: 162400393-162400393 on Build GRCh38
rs750223366 Chr.2: 162400487-162400488 on Build GRCh38
rs774341855 Chr.2: 162400156-162400156 on Build GRCh38
rs16846665 Chr.2: 162400439-162400439 on Build GRCh38
rs370109886 Chr.2: 162400385-162400385 on Build GRCh38
rs200321479 Chr.2: 162400164-162400164 on Build GRCh38
rs767975805 Chr.2: 162400279-162400279 on Build GRCh38
rs776408897 Chr.2: 162400194-162400194 on Build GRCh38
rs79976943 Chr.2: 162400190-162400190 on Build GRCh38
rs149972724 Chr.2: 162400283-162400283 on Build GRCh38
rs778362235 Chr.2: 162400469-162400469 on Build GRCh38
rs766485529 Chr.2: 162400256-162400256 on Build GRCh38
rs756536838 Chr.2: 162400280-162400280 on Build GRCh38
rs563639308 Chr.2: 162400468-162400468 on Build GRCh38
rs901600220 Chr.2: 162400137-162400137 on Build GRCh38
rs745584575 Chr.2: 162400403-162400403 on Build GRCh38
rs577435003 Chr.2: 162400331-162400331 on Build GRCh38
rs771751716 Chr.2: 162400356-162400356 on Build GRCh38
rs143736950 Chr.2: 162400361-162400361 on Build GRCh38
rs1013343141 Chr.2: 162400467-162400467 on Build GRCh38
rs774626303 Chr.2: 162400405-162400405 on Build GRCh38
rs754372322 Chr.2: 162400298-162400298 on Build GRCh38
rs768334436 Chr.2: 162400489-162400489 on Build GRCh38
rs749895516 Chr.2: 162400471-162400471 on Build GRCh38
rs759803008 Chr.2: 162400408-162400408 on Build GRCh38
rs1028183460 Chr.2: 162400088-162400088 on Build GRCh38

View Full Product Details