Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176151307-176151804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD3
Entrez Gene ID
3232
Gene Name
homeobox D3
Gene Aliases
HOX1D, HOX4, HOX4A, Hox-4.1
UniGene
Hs.93574
Gene Chromosome Location
Chr.2: 176136921-176173102 on Build GRCh38
Transcripts
XM_005246510.4 XM_006712477.2 NM_014621.2 XM_005246514.4

Gene Symbol
HOXD4
Entrez Gene ID
3233
Gene Name
homeobox D4
Gene Aliases
HHO.C13, HOX-5.1, HOX4, HOX4B, Hox-4.2
UniGene
Hs.591609
Gene Chromosome Location
Chr.2: 176150394-176156078 on Build GRCh38
Transcripts
XM_005246510.4 XM_006712477.2 NM_014621.2 XM_005246514.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
141006 c.52C>T p.P18S Chr.2: 176151685-176151685 on Build GRCh38
5665916 c.103G>C p.D35H Chr.2: 176151736-176151736 on Build GRCh38
5643765 c.155A>T p.Y52F Chr.2: 176151788-176151788 on Build GRCh38
4086958 c.51C>A p.F17L Chr.2: 176151684-176151684 on Build GRCh38
5634177 c.72G>T p.L24F Chr.2: 176151705-176151705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780066078 Chr.2: 176151778-176151778 on Build GRCh38
rs943521878 Chr.2: 176151435-176151435 on Build GRCh38
rs774592120 Chr.2: 176151726-176151755 on Build GRCh38
rs1041044179 Chr.2: 176151486-176151486 on Build GRCh38
rs762100298 Chr.2: 176151618-176151618 on Build GRCh38
rs997611203 Chr.2: 176151447-176151447 on Build GRCh38
rs766920993 Chr.2: 176151648-176151648 on Build GRCh38
rs1052876017 Chr.2: 176151361-176151361 on Build GRCh38
rs1026267238 Chr.2: 176151568-176151568 on Build GRCh38
rs936436450 Chr.2: 176151729-176151729 on Build GRCh38
rs779976253 Chr.2: 176151732-176151732 on Build GRCh38
rs1017814987 Chr.2: 176151438-176151438 on Build GRCh38
rs568413462 Chr.2: 176151439-176151439 on Build GRCh38
rs542100496 Chr.2: 176151751-176151751 on Build GRCh38
rs758400794 Chr.2: 176151776-176151776 on Build GRCh38
rs746810203 Chr.2: 176151779-176151779 on Build GRCh38
rs924937832 Chr.2: 176151723-176151723 on Build GRCh38
rs16863572 Chr.2: 176151566-176151566 on Build GRCh38
rs138948127 Chr.2: 176151763-176151763 on Build GRCh38
rs777162481 Chr.2: 176151410-176151410 on Build GRCh38
rs780918058 Chr.2: 176151583-176151583 on Build GRCh38
rs930015171 Chr.2: 176151736-176151736 on Build GRCh38
rs925031375 Chr.2: 176151376-176151376 on Build GRCh38
rs779403544 Chr.2: 176151565-176151565 on Build GRCh38
rs777027994 Chr.2: 176151612-176151612 on Build GRCh38
rs187203814 Chr.2: 176151724-176151724 on Build GRCh38
rs928513975 Chr.2: 176151593-176151595 on Build GRCh38
rs766668683 Chr.2: 176151592-176151594 on Build GRCh38
rs755660748 Chr.2: 176151595-176151595 on Build GRCh38
rs569812979 Chr.2: 176151560-176151560 on Build GRCh38
rs770897971 Chr.2: 176151607-176151607 on Build GRCh38
rs372977271 Chr.2: 176151622-176151622 on Build GRCh38
rs767926468 Chr.2: 176151758-176151758 on Build GRCh38
rs376860683 Chr.2: 176151690-176151690 on Build GRCh38
rs928466833 Chr.2: 176151346-176151346 on Build GRCh38
rs549994105 Chr.2: 176151406-176151406 on Build GRCh38
rs150414482 Chr.2: 176151697-176151697 on Build GRCh38
rs181916447 Chr.2: 176151623-176151623 on Build GRCh38
rs763287731 Chr.2: 176151584-176151585 on Build GRCh38
rs773418329 Chr.2: 176151635-176151635 on Build GRCh38
rs564014954 Chr.2: 176151771-176151771 on Build GRCh38
rs979995758 Chr.2: 176151344-176151344 on Build GRCh38
rs752794283 Chr.2: 176151763-176151764 on Build GRCh38
rs78015126 Chr.2: 176151768-176151768 on Build GRCh38
rs556334519 Chr.2: 176151701-176151701 on Build GRCh38
rs751958240 Chr.2: 176151664-176151664 on Build GRCh38
rs887903097 Chr.2: 176151570-176151570 on Build GRCh38
rs549838512 Chr.2: 176151372-176151372 on Build GRCh38
rs755750496 Chr.2: 176151686-176151686 on Build GRCh38
rs141531995 Chr.2: 176151647-176151647 on Build GRCh38
rs774960831 Chr.2: 176151755-176151755 on Build GRCh38
rs553839993 Chr.2: 176151556-176151556 on Build GRCh38
rs749281568 Chr.2: 176151600-176151600 on Build GRCh38
rs977746363 Chr.2: 176151368-176151368 on Build GRCh38
rs374205432 Chr.2: 176151750-176151750 on Build GRCh38
rs144430144 Chr.2: 176151487-176151487 on Build GRCh38
rs772454886 Chr.2: 176151562-176151562 on Build GRCh38
rs760964216 Chr.2: 176151762-176151762 on Build GRCh38
rs767834489 Chr.2: 176151681-176151681 on Build GRCh38
rs947067323 Chr.2: 176151403-176151403 on Build GRCh38
rs749437372 Chr.2: 176151785-176151785 on Build GRCh38
rs768519064 Chr.2: 176151780-176151780 on Build GRCh38
rs774091416 Chr.2: 176151608-176151608 on Build GRCh38
rs936397264 Chr.2: 176151393-176151393 on Build GRCh38
rs747757026 Chr.2: 176151594-176151594 on Build GRCh38
rs759975586 Chr.2: 176151669-176151669 on Build GRCh38
rs771719217 Chr.2: 176151611-176151611 on Build GRCh38
rs778935489 Chr.2: 176151719-176151719 on Build GRCh38
rs200815560 Chr.2: 176151746-176151746 on Build GRCh38
rs753056547 Chr.2: 176151760-176151760 on Build GRCh38
rs745690579 Chr.2: 176151609-176151609 on Build GRCh38
rs977785447 Chr.2: 176151712-176151712 on Build GRCh38
rs377617802 Chr.2: 176151775-176151775 on Build GRCh38
rs950785163 Chr.2: 176151522-176151522 on Build GRCh38
rs759937750 Chr.2: 176151756-176151756 on Build GRCh38
rs759709428 Chr.2: 176151737-176151739 on Build GRCh38
rs891525120 Chr.2: 176151397-176151397 on Build GRCh38
rs752396313 Chr.2: 176151684-176151684 on Build GRCh38
rs149156170 Chr.2: 176151731-176151731 on Build GRCh38
rs773508238 Chr.2: 176151749-176151749 on Build GRCh38
rs776033572 Chr.2: 176151715-176151715 on Build GRCh38
rs984848971 Chr.2: 176151527-176151527 on Build GRCh38
rs938458565 Chr.2: 176151352-176151352 on Build GRCh38
rs770140508 Chr.2: 176151747-176151747 on Build GRCh38
rs767546885 Chr.2: 176151745-176151758 on Build GRCh38

View Full Product Details