Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 188477471-188477998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GULP1
Entrez Gene ID
51454
Gene Name
GULP, engulfment adaptor PTB domain containing 1
Gene Aliases
CED-6, CED6, GULP
UniGene
Hs.470887
Gene Chromosome Location
Chr.2: 188291669-188595926 on Build GRCh38
Transcripts
NM_001252668.1 NM_001252669.1 NM_016315.3 NR_045562.1 NR_045563.1 XM_006712580.3 XM_006712581.1 XM_006712582.1 XM_006712583.3 XM_006712584.3 XM_006712585.1 XM_006712589.3 XM_011511329.1 XM_011511332.1 XM_011511333.1 XM_011511335.2 XM_017004303.1 XM_017004304.1 XM_017004305.1 XM_017004306.1 XM_017004307.1 XM_017004308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1253922 c.5A>G p.N2S Chr.2: 188477707-188477707 on Build GRCh38
4474169 c.18C>G p.S6R Chr.2: 188477720-188477720 on Build GRCh38
5804050 c.22A>T p.K8* Chr.2: 188477724-188477724 on Build GRCh38
5024167 c.8G>A p.R3H Chr.2: 188477710-188477710 on Build GRCh38
4474168 c.18C>G p.S6R Chr.2: 188477720-188477720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751120789 Chr.2: 188477759-188477759 on Build GRCh38
rs764790887 Chr.2: 188477680-188477680 on Build GRCh38
rs1018744653 Chr.2: 188477594-188477594 on Build GRCh38
rs529216963 Chr.2: 188477807-188477807 on Build GRCh38
rs1049426658 Chr.2: 188477791-188477791 on Build GRCh38
rs958995441 Chr.2: 188477934-188477934 on Build GRCh38
rs577375609 Chr.2: 188477575-188477575 on Build GRCh38
rs1024426261 Chr.2: 188477940-188477940 on Build GRCh38
rs1033652895 Chr.2: 188477920-188477920 on Build GRCh38
rs567493068 Chr.2: 188477949-188477949 on Build GRCh38
rs868536466 Chr.2: 188477863-188477863 on Build GRCh38
rs528148498 Chr.2: 188477770-188477770 on Build GRCh38
rs899579773 Chr.2: 188477544-188477544 on Build GRCh38
rs889051426 Chr.2: 188477792-188477792 on Build GRCh38
rs867010993 Chr.2: 188477614-188477614 on Build GRCh38
rs755514186 Chr.2: 188477700-188477700 on Build GRCh38
rs768384047 Chr.2: 188477763-188477763 on Build GRCh38
rs115341282 Chr.2: 188477907-188477907 on Build GRCh38
rs890739294 Chr.2: 188477583-188477583 on Build GRCh38
rs968685275 Chr.2: 188477631-188477631 on Build GRCh38
rs141512059 Chr.2: 188477917-188477917 on Build GRCh38
rs1040208126 Chr.2: 188477867-188477867 on Build GRCh38
rs971571864 Chr.2: 188477954-188477954 on Build GRCh38
rs992028107 Chr.2: 188477939-188477939 on Build GRCh38
rs756509880 Chr.2: 188477710-188477710 on Build GRCh38
rs781725363 Chr.2: 188477707-188477707 on Build GRCh38
rs78363380 Chr.2: 188477876-188477876 on Build GRCh38
rs1034346398 Chr.2: 188477952-188477952 on Build GRCh38
rs753137025 Chr.2: 188477709-188477709 on Build GRCh38
rs959077057 Chr.2: 188477966-188477966 on Build GRCh38
rs967403543 Chr.2: 188477649-188477649 on Build GRCh38
rs13403508 Chr.2: 188477875-188477875 on Build GRCh38
rs374687427 Chr.2: 188477757-188477757 on Build GRCh38
rs764209008 Chr.2: 188477953-188477953 on Build GRCh38
rs746848199 Chr.2: 188477951-188477952 on Build GRCh38
rs148360745 Chr.2: 188477788-188477788 on Build GRCh38
rs371783059 Chr.2: 188477692-188477692 on Build GRCh38
rs185698058 Chr.2: 188477599-188477599 on Build GRCh38
rs979413843 Chr.2: 188477656-188477656 on Build GRCh38
rs749930031 Chr.2: 188477771-188477771 on Build GRCh38
rs541664911 Chr.2: 188477576-188477576 on Build GRCh38
rs13414327 Chr.2: 188477667-188477667 on Build GRCh38
rs535077489 Chr.2: 188477956-188477956 on Build GRCh38
rs951771822 Chr.2: 188477973-188477973 on Build GRCh38
rs367777352 Chr.2: 188477767-188477767 on Build GRCh38
rs188907784 Chr.2: 188477762-188477762 on Build GRCh38
rs996772389 Chr.2: 188477548-188477548 on Build GRCh38
rs200557671 Chr.2: 188477679-188477679 on Build GRCh38
rs371721822 Chr.2: 188477773-188477773 on Build GRCh38
rs369610819 Chr.2: 188477810-188477810 on Build GRCh38
rs909566763 Chr.2: 188477855-188477855 on Build GRCh38
rs570959186 Chr.2: 188477925-188477925 on Build GRCh38
rs745833001 Chr.2: 188477745-188477745 on Build GRCh38
rs879917645 Chr.2: 188477505-188477505 on Build GRCh38
rs775355361 Chr.2: 188477758-188477758 on Build GRCh38
rs553555245 Chr.2: 188477596-188477596 on Build GRCh38
rs73040496 Chr.2: 188477967-188477967 on Build GRCh38
rs1020406567 Chr.2: 188477894-188477894 on Build GRCh38
rs991687483 Chr.2: 188477672-188477672 on Build GRCh38
rs1010925019 Chr.2: 188477919-188477919 on Build GRCh38
rs749465498 Chr.2: 188477726-188477726 on Build GRCh38
rs779046130 Chr.2: 188477743-188477743 on Build GRCh38
rs778143176 Chr.2: 188477713-188477713 on Build GRCh38
rs763125808 Chr.2: 188477937-188477937 on Build GRCh38
rs546394419 Chr.2: 188477778-188477778 on Build GRCh38
rs201523693 Chr.2: 188477739-188477739 on Build GRCh38
rs949807339 Chr.2: 188477754-188477754 on Build GRCh38
rs760365739 Chr.2: 188477662-188477662 on Build GRCh38
rs542498733 Chr.2: 188477609-188477609 on Build GRCh38
rs1020291991 Chr.2: 188477632-188477632 on Build GRCh38
rs761243576 Chr.2: 188477669-188477669 on Build GRCh38
rs750218502 Chr.2: 188477730-188477731 on Build GRCh38
rs200628153 Chr.2: 188477668-188477668 on Build GRCh38
rs534805521 Chr.2: 188477950-188477950 on Build GRCh38

View Full Product Details