Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 188569897-188570421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GULP1
Entrez Gene ID
51454
Gene Name
GULP, engulfment adaptor PTB domain containing 1
Gene Aliases
CED-6, CED6, GULP
UniGene
Hs.470887
Gene Chromosome Location
Chr.2: 188291669-188595926 on Build GRCh38
Transcripts
NM_001252668.1 NM_001252669.1 NM_016315.3 NR_045562.1 NR_045563.1 XM_006712580.3 XM_006712581.1 XM_006712582.1 XM_006712583.3 XM_006712584.3 XM_006712585.1 XM_006712589.3 XM_011511329.1 XM_011511331.1 XM_011511332.1 XM_011511333.1 XM_011511335.2 XM_017004303.1 XM_017004304.1 XM_017004305.1 XM_017004306.1 XM_017004307.1 XM_017004308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6296080 c.520C>A p.Q174K Chr.2: 188570031-188570031 on Build GRCh38
5775375 c.522A>T p.Q174H Chr.2: 188570033-188570033 on Build GRCh38
1614044 c.593A>C p.Q198P Chr.2: 188570104-188570104 on Build GRCh38
1013927 c.561A>G p.V187V Chr.2: 188570072-188570072 on Build GRCh38
1403984 c.580C>T p.L194L Chr.2: 188570091-188570091 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775067927 Chr.2: 188570152-188570152 on Build GRCh38
rs761188427 Chr.2: 188570055-188570055 on Build GRCh38
rs778606359 Chr.2: 188569947-188569947 on Build GRCh38
rs904193397 Chr.2: 188570165-188570165 on Build GRCh38
rs377527204 Chr.2: 188570013-188570013 on Build GRCh38
rs183912239 Chr.2: 188569974-188569974 on Build GRCh38
rs1042853058 Chr.2: 188570309-188570309 on Build GRCh38
rs748020874 Chr.2: 188569973-188569973 on Build GRCh38
rs749229744 Chr.2: 188570001-188570001 on Build GRCh38
rs758131087 Chr.2: 188569984-188569985 on Build GRCh38
rs996369599 Chr.2: 188570213-188570213 on Build GRCh38
rs771796407 Chr.2: 188570031-188570031 on Build GRCh38
rs561226614 Chr.2: 188569928-188569928 on Build GRCh38
rs895711831 Chr.2: 188570365-188570365 on Build GRCh38
rs778832408 Chr.2: 188570116-188570116 on Build GRCh38
rs187302982 Chr.2: 188570156-188570156 on Build GRCh38
rs368740275 Chr.2: 188570285-188570285 on Build GRCh38
rs775180915 Chr.2: 188570037-188570037 on Build GRCh38
rs373024772 Chr.2: 188570149-188570149 on Build GRCh38
rs977297425 Chr.2: 188570266-188570266 on Build GRCh38
rs753411759 Chr.2: 188570053-188570053 on Build GRCh38
rs771919175 Chr.2: 188570119-188570119 on Build GRCh38
rs191741660 Chr.2: 188570259-188570259 on Build GRCh38
rs961383152 Chr.2: 188569941-188569941 on Build GRCh38
rs752906524 Chr.2: 188570086-188570086 on Build GRCh38
rs925602224 Chr.2: 188570342-188570342 on Build GRCh38
rs767992135 Chr.2: 188570083-188570083 on Build GRCh38
rs751526439 Chr.2: 188569996-188569997 on Build GRCh38
rs985666525 Chr.2: 188570260-188570260 on Build GRCh38
rs994268984 Chr.2: 188570254-188570254 on Build GRCh38
rs951031088 Chr.2: 188570360-188570360 on Build GRCh38
rs374345703 Chr.2: 188570170-188570170 on Build GRCh38
rs776083655 Chr.2: 188570147-188570147 on Build GRCh38
rs770920774 Chr.2: 188570007-188570007 on Build GRCh38
rs1014527621 Chr.2: 188570212-188570212 on Build GRCh38
rs978584504 Chr.2: 188569987-188569987 on Build GRCh38
rs775234108 Chr.2: 188570122-188570122 on Build GRCh38
rs753123839 Chr.2: 188570166-188570166 on Build GRCh38
rs767000174 Chr.2: 188570057-188570057 on Build GRCh38
rs151074734 Chr.2: 188570072-188570072 on Build GRCh38
rs911500013 Chr.2: 188570263-188570263 on Build GRCh38
rs768387068 Chr.2: 188570133-188570133 on Build GRCh38
rs1025508319 Chr.2: 188569940-188569940 on Build GRCh38
rs576073671 Chr.2: 188570289-188570289 on Build GRCh38
rs777750297 Chr.2: 188570174-188570174 on Build GRCh38
rs200711089 Chr.2: 188570157-188570157 on Build GRCh38
rs755455550 Chr.2: 188570074-188570074 on Build GRCh38
rs919939382 Chr.2: 188569953-188569953 on Build GRCh38
rs757385151 Chr.2: 188570172-188570172 on Build GRCh38
rs558818193 Chr.2: 188570386-188570386 on Build GRCh38
rs777805792 Chr.2: 188570091-188570091 on Build GRCh38
rs777966236 Chr.2: 188570060-188570065 on Build GRCh38
rs780061753 Chr.2: 188569985-188569985 on Build GRCh38
rs755012491 Chr.2: 188570039-188570044 on Build GRCh38
rs746688944 Chr.2: 188570128-188570128 on Build GRCh38
rs769833915 Chr.2: 188569999-188569999 on Build GRCh38
rs954819286 Chr.2: 188569971-188569971 on Build GRCh38
rs746931879 Chr.2: 188570184-188570184 on Build GRCh38
rs115951524 Chr.2: 188570276-188570276 on Build GRCh38
rs756340887 Chr.2: 188570087-188570087 on Build GRCh38
rs183573379 Chr.2: 188570264-188570264 on Build GRCh38
rs968823559 Chr.2: 188570271-188570271 on Build GRCh38
rs937019916 Chr.2: 188570353-188570353 on Build GRCh38
rs1037869280 Chr.2: 188570160-188570160 on Build GRCh38
rs1046868113 Chr.2: 188570138-188570138 on Build GRCh38
rs770441140 Chr.2: 188570236-188570236 on Build GRCh38
rs547757588 Chr.2: 188570225-188570225 on Build GRCh38
rs748053743 Chr.2: 188570231-188570231 on Build GRCh38
rs756390336 Chr.2: 188570169-188570169 on Build GRCh38
rs958641942 Chr.2: 188570322-188570322 on Build GRCh38
rs771729850 Chr.2: 188570150-188570150 on Build GRCh38
rs1055701792 Chr.2: 188570357-188570357 on Build GRCh38
rs912951508 Chr.2: 188570111-188570111 on Build GRCh38
rs759327196 Chr.2: 188570027-188570027 on Build GRCh38
rs763552013 Chr.2: 188570044-188570044 on Build GRCh38
rs1026901841 Chr.2: 188570258-188570258 on Build GRCh38
rs992758243 Chr.2: 188570348-188570348 on Build GRCh38
rs376441217 Chr.2: 188570039-188570039 on Build GRCh38
rs537116303 Chr.2: 188570382-188570382 on Build GRCh38
rs748371849 Chr.2: 188569994-188569994 on Build GRCh38
rs143831338 Chr.2: 188570106-188570106 on Build GRCh38

View Full Product Details