Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197499675-197500168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPD1
Entrez Gene ID
3329
Gene Name
heat shock protein family D (Hsp60) member 1
Gene Aliases
CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13
UniGene
Hs.595053
Gene Chromosome Location
Chr.2: 197486584-197500274 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NM_002157.2 NM_001202485.1

Gene Symbol
HSPE1
Entrez Gene ID
3336
Gene Name
heat shock protein family E (Hsp10) member 1
Gene Aliases
CPN10, EPF, GROES, HSP10
UniGene
Hs.1197
Gene Chromosome Location
Chr.2: 197499997-197503463 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NM_002157.2 NM_001202485.1

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197499997-197553699 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NM_002157.2 NM_001202485.1

SNP Detail

SNP ID Chromosome Location
rs1047023513 Chr.2: 197499870-197499870 on Build GRCh38
rs1040568685 Chr.2: 197499969-197499969 on Build GRCh38
rs764257215 Chr.2: 197499884-197499884 on Build GRCh38
rs539674750 Chr.2: 197500089-197500089 on Build GRCh38
rs972502657 Chr.2: 197499986-197499986 on Build GRCh38
rs553287323 Chr.2: 197499990-197499990 on Build GRCh38
rs763053276 Chr.2: 197500045-197500045 on Build GRCh38
rs1005311001 Chr.2: 197500093-197500093 on Build GRCh38
rs1026766351 Chr.2: 197500141-197500141 on Build GRCh38
rs34097997 Chr.2: 197500079-197500080 on Build GRCh38
rs998350491 Chr.2: 197500054-197500054 on Build GRCh38
rs889206644 Chr.2: 197500145-197500145 on Build GRCh38
rs34519984 Chr.2: 197499845-197499845 on Build GRCh38
rs568292027 Chr.2: 197499970-197499970 on Build GRCh38
rs113661821 Chr.2: 197500024-197500024 on Build GRCh38
rs769972758 Chr.2: 197499977-197499977 on Build GRCh38
rs1029073804 Chr.2: 197499737-197499737 on Build GRCh38
rs952403147 Chr.2: 197500018-197500018 on Build GRCh38
rs893844729 Chr.2: 197499910-197499910 on Build GRCh38
rs535199888 Chr.2: 197500117-197500117 on Build GRCh38
rs949538232 Chr.2: 197499988-197499988 on Build GRCh38
rs963358022 Chr.2: 197499982-197499982 on Build GRCh38
rs1025515216 Chr.2: 197500017-197500017 on Build GRCh38
rs572474963 Chr.2: 197500110-197500110 on Build GRCh38
rs985272441 Chr.2: 197499787-197499787 on Build GRCh38
rs961299999 Chr.2: 197500135-197500135 on Build GRCh38
rs751750227 Chr.2: 197499917-197499917 on Build GRCh38
rs1036423930 Chr.2: 197499855-197499855 on Build GRCh38
rs953845012 Chr.2: 197499964-197499964 on Build GRCh38
rs762763033 Chr.2: 197499854-197499854 on Build GRCh38
rs926787615 Chr.2: 197500006-197500006 on Build GRCh38
rs1016885746 Chr.2: 197499781-197499782 on Build GRCh38
rs982307511 Chr.2: 197500044-197500044 on Build GRCh38
rs377275803 Chr.2: 197499766-197499766 on Build GRCh38
rs972672400 Chr.2: 197500136-197500136 on Build GRCh38
rs1002436934 Chr.2: 197499708-197499708 on Build GRCh38
rs1052453050 Chr.2: 197499907-197499907 on Build GRCh38
rs371832805 Chr.2: 197499993-197499993 on Build GRCh38
rs59701865 Chr.2: 197499767-197499767 on Build GRCh38
rs67255036 Chr.2: 197499987-197499987 on Build GRCh38
rs766379968 Chr.2: 197500079-197500079 on Build GRCh38
rs1016781641 Chr.2: 197499980-197499980 on Build GRCh38
rs983876273 Chr.2: 197499861-197499861 on Build GRCh38
rs570654765 Chr.2: 197500065-197500065 on Build GRCh38
rs994527532 Chr.2: 197499998-197499998 on Build GRCh38
rs528611166 Chr.2: 197499894-197499894 on Build GRCh38
rs945594583 Chr.2: 197500023-197500023 on Build GRCh38
rs13165 Chr.2: 197499794-197499794 on Build GRCh38
rs1032049594 Chr.2: 197499721-197499721 on Build GRCh38
rs552704943 Chr.2: 197500092-197500092 on Build GRCh38
rs1020605337 Chr.2: 197499913-197499913 on Build GRCh38
rs886055390 Chr.2: 197499907-197499907 on Build GRCh38
rs557264264 Chr.2: 197500008-197500008 on Build GRCh38
rs938137406 Chr.2: 197500007-197500007 on Build GRCh38
rs532584313 Chr.2: 197499838-197499838 on Build GRCh38
rs935540915 Chr.2: 197499902-197499902 on Build GRCh38
rs906801199 Chr.2: 197499895-197499895 on Build GRCh38
rs16866252 Chr.2: 197499919-197499919 on Build GRCh38
rs935507239 Chr.2: 197500049-197500049 on Build GRCh38
rs1019540905 Chr.2: 197499789-197499789 on Build GRCh38
rs1050394889 Chr.2: 197500143-197500143 on Build GRCh38
rs1021102167 Chr.2: 197499757-197499757 on Build GRCh38
rs977021087 Chr.2: 197499775-197499775 on Build GRCh38
rs555025799 Chr.2: 197500132-197500132 on Build GRCh38
rs972430072 Chr.2: 197499795-197499795 on Build GRCh38
rs983736965 Chr.2: 197500019-197500019 on Build GRCh38
rs563816458 Chr.2: 197499799-197499799 on Build GRCh38
rs1028610503 Chr.2: 197499939-197499939 on Build GRCh38
rs919303896 Chr.2: 197499859-197499860 on Build GRCh38
rs892061773 Chr.2: 197499755-197499755 on Build GRCh38
rs918214173 Chr.2: 197499831-197499831 on Build GRCh38
rs905469872 Chr.2: 197499704-197499704 on Build GRCh38
rs969089471 Chr.2: 197500033-197500033 on Build GRCh38
rs777039079 Chr.2: 197499866-197499866 on Build GRCh38
rs909223870 Chr.2: 197499801-197499801 on Build GRCh38
rs566023346 Chr.2: 197499888-197499888 on Build GRCh38
rs983993949 Chr.2: 197499800-197499800 on Build GRCh38
rs1054003374 Chr.2: 197499890-197499890 on Build GRCh38
rs1028032399 Chr.2: 197500058-197500058 on Build GRCh38
rs1009324527 Chr.2: 197499752-197499752 on Build GRCh38
rs984003770 Chr.2: 197499970-197499970 on Build GRCh38
rs916314956 Chr.2: 197499865-197499865 on Build GRCh38

View Full Product Details