Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197499859-197500357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPD1
Entrez Gene ID
3329
Gene Name
heat shock protein family D (Hsp60) member 1
Gene Aliases
CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13
UniGene
Hs.595053
Gene Chromosome Location
Chr.2: 197486584-197500274 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NM_002157.2 NM_001202485.1

Gene Symbol
HSPE1
Entrez Gene ID
3336
Gene Name
heat shock protein family E (Hsp10) member 1
Gene Aliases
CPN10, EPF, GROES, HSP10
UniGene
Hs.1197
Gene Chromosome Location
Chr.2: 197499997-197503463 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NM_002157.2 NM_001202485.1

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197499997-197553699 on Build GRCh38
Transcripts
NM_002156.4 NM_199440.1 NM_002157.2 NM_001202485.1

SNP Detail

SNP ID Chromosome Location
rs764257215 Chr.2: 197499884-197499884 on Build GRCh38
rs539674750 Chr.2: 197500089-197500089 on Build GRCh38
rs577475520 Chr.2: 197500273-197500273 on Build GRCh38
rs553287323 Chr.2: 197499990-197499990 on Build GRCh38
rs763053276 Chr.2: 197500045-197500045 on Build GRCh38
rs1026766351 Chr.2: 197500141-197500141 on Build GRCh38
rs889206644 Chr.2: 197500145-197500145 on Build GRCh38
rs568292027 Chr.2: 197499970-197499970 on Build GRCh38
rs113661821 Chr.2: 197500024-197500024 on Build GRCh38
rs1046854341 Chr.2: 197500304-197500304 on Build GRCh38
rs952599576 Chr.2: 197500213-197500213 on Build GRCh38
rs528474896 Chr.2: 197500316-197500316 on Build GRCh38
rs543605862 Chr.2: 197500206-197500206 on Build GRCh38
rs982623817 Chr.2: 197500180-197500180 on Build GRCh38
rs535199888 Chr.2: 197500117-197500117 on Build GRCh38
rs996257279 Chr.2: 197500291-197500291 on Build GRCh38
rs949538232 Chr.2: 197499988-197499988 on Build GRCh38
rs1025515216 Chr.2: 197500017-197500017 on Build GRCh38
rs3749094 Chr.2: 197500148-197500148 on Build GRCh38
rs751750227 Chr.2: 197499917-197499917 on Build GRCh38
rs926665699 Chr.2: 197500193-197500193 on Build GRCh38
rs926787615 Chr.2: 197500006-197500006 on Build GRCh38
rs1005196440 Chr.2: 197500214-197500214 on Build GRCh38
rs559511143 Chr.2: 197500299-197500299 on Build GRCh38
rs1052453050 Chr.2: 197499907-197499907 on Build GRCh38
rs371832805 Chr.2: 197499993-197499993 on Build GRCh38
rs67255036 Chr.2: 197499987-197499987 on Build GRCh38
rs927734022 Chr.2: 197500231-197500231 on Build GRCh38
rs1042611498 Chr.2: 197500240-197500240 on Build GRCh38
rs1016781641 Chr.2: 197499980-197499980 on Build GRCh38
rs567109406 Chr.2: 197500249-197500249 on Build GRCh38
rs944720454 Chr.2: 197500279-197500279 on Build GRCh38
rs763864525 Chr.2: 197500257-197500257 on Build GRCh38
rs570654765 Chr.2: 197500065-197500065 on Build GRCh38
rs528611166 Chr.2: 197499894-197499894 on Build GRCh38
rs945594583 Chr.2: 197500023-197500023 on Build GRCh38
rs66504718 Chr.2: 197500296-197500296 on Build GRCh38
rs552704943 Chr.2: 197500092-197500092 on Build GRCh38
rs1020605337 Chr.2: 197499913-197499913 on Build GRCh38
rs16866252 Chr.2: 197499919-197499919 on Build GRCh38
rs906801199 Chr.2: 197499895-197499895 on Build GRCh38
rs935540915 Chr.2: 197499902-197499902 on Build GRCh38
rs935507239 Chr.2: 197500049-197500049 on Build GRCh38
rs1050394889 Chr.2: 197500143-197500143 on Build GRCh38
rs778733975 Chr.2: 197500333-197500333 on Build GRCh38
rs555025799 Chr.2: 197500132-197500132 on Build GRCh38
rs1005240591 Chr.2: 197500280-197500280 on Build GRCh38
rs113142336 Chr.2: 197500212-197500212 on Build GRCh38
rs566023346 Chr.2: 197499888-197499888 on Build GRCh38
rs1054003374 Chr.2: 197499890-197499890 on Build GRCh38
rs1040568685 Chr.2: 197499969-197499969 on Build GRCh38
rs933646010 Chr.2: 197500251-197500251 on Build GRCh38
rs972502657 Chr.2: 197499986-197499986 on Build GRCh38
rs560829300 Chr.2: 197500211-197500211 on Build GRCh38
rs1005311001 Chr.2: 197500093-197500093 on Build GRCh38
rs945654766 Chr.2: 197500234-197500234 on Build GRCh38
rs34097997 Chr.2: 197500079-197500080 on Build GRCh38
rs998350491 Chr.2: 197500054-197500054 on Build GRCh38
rs994498318 Chr.2: 197500328-197500328 on Build GRCh38
rs769972758 Chr.2: 197499977-197499977 on Build GRCh38
rs952403147 Chr.2: 197500018-197500018 on Build GRCh38
rs904897811 Chr.2: 197500321-197500321 on Build GRCh38
rs893844729 Chr.2: 197499910-197499910 on Build GRCh38
rs67198751 Chr.2: 197500322-197500322 on Build GRCh38
rs901039688 Chr.2: 197500210-197500210 on Build GRCh38
rs1023219923 Chr.2: 197500215-197500215 on Build GRCh38
rs963358022 Chr.2: 197499982-197499982 on Build GRCh38
rs572474963 Chr.2: 197500110-197500110 on Build GRCh38
rs961299999 Chr.2: 197500135-197500135 on Build GRCh38
rs957116940 Chr.2: 197500241-197500241 on Build GRCh38
rs953845012 Chr.2: 197499964-197499964 on Build GRCh38
rs915509814 Chr.2: 197500269-197500269 on Build GRCh38
rs982307511 Chr.2: 197500044-197500044 on Build GRCh38
rs1116734 Chr.2: 197500319-197500319 on Build GRCh38
rs994418033 Chr.2: 197500303-197500303 on Build GRCh38
rs972672400 Chr.2: 197500136-197500136 on Build GRCh38
rs373346246 Chr.2: 197500176-197500176 on Build GRCh38
rs766379968 Chr.2: 197500079-197500079 on Build GRCh38
rs994527532 Chr.2: 197499998-197499998 on Build GRCh38
rs969175107 Chr.2: 197500218-197500218 on Build GRCh38
rs910588793 Chr.2: 197500278-197500278 on Build GRCh38
rs946912020 Chr.2: 197500272-197500272 on Build GRCh38
rs886055390 Chr.2: 197499907-197499907 on Build GRCh38
rs557264264 Chr.2: 197500008-197500008 on Build GRCh38
rs1040451158 Chr.2: 197500195-197500195 on Build GRCh38
rs938137406 Chr.2: 197500007-197500007 on Build GRCh38
rs938063822 Chr.2: 197500194-197500194 on Build GRCh38
rs922262307 Chr.2: 197500242-197500242 on Build GRCh38
rs1005392910 Chr.2: 197500318-197500318 on Build GRCh38
rs983736965 Chr.2: 197500019-197500019 on Build GRCh38
rs1028610503 Chr.2: 197499939-197499939 on Build GRCh38
rs67387011 Chr.2: 197500302-197500303 on Build GRCh38
rs760649940 Chr.2: 197500229-197500229 on Build GRCh38
rs753639844 Chr.2: 197500312-197500312 on Build GRCh38
rs969089471 Chr.2: 197500033-197500033 on Build GRCh38
rs982305100 Chr.2: 197500225-197500225 on Build GRCh38
rs1028032399 Chr.2: 197500058-197500058 on Build GRCh38
rs984003770 Chr.2: 197499970-197499970 on Build GRCh38

View Full Product Details