Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226731549-226732068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

SNP Detail

SNP ID Chromosome Location
rs1001719197 Chr.2: 226731849-226731849 on Build GRCh38
rs867507867 Chr.2: 226732018-226732018 on Build GRCh38
rs569140464 Chr.2: 226731574-226731574 on Build GRCh38
rs1035809970 Chr.2: 226731862-226731862 on Build GRCh38
rs886679302 Chr.2: 226731797-226731797 on Build GRCh38
rs989814158 Chr.2: 226731918-226731918 on Build GRCh38
rs750360622 Chr.2: 226731781-226731781 on Build GRCh38
rs2288588 Chr.2: 226732023-226732023 on Build GRCh38
rs922396266 Chr.2: 226731935-226731935 on Build GRCh38
rs554836269 Chr.2: 226731686-226731686 on Build GRCh38
rs1047250115 Chr.2: 226731990-226731990 on Build GRCh38
rs894692010 Chr.2: 226732007-226732007 on Build GRCh38
rs558750343 Chr.2: 226731847-226731847 on Build GRCh38
rs933725573 Chr.2: 226731966-226731966 on Build GRCh38
rs943456370 Chr.2: 226731695-226731695 on Build GRCh38
rs750829143 Chr.2: 226731737-226731737 on Build GRCh38
rs577397572 Chr.2: 226731924-226731924 on Build GRCh38
rs1039433321 Chr.2: 226731617-226731617 on Build GRCh38
rs956676333 Chr.2: 226731888-226731888 on Build GRCh38
rs2288589 Chr.2: 226732024-226732024 on Build GRCh38
rs1037871142 Chr.2: 226731843-226731843 on Build GRCh38
rs981979926 Chr.2: 226731848-226731848 on Build GRCh38
rs1039029090 Chr.2: 226732004-226732004 on Build GRCh38
rs146578458 Chr.2: 226732003-226732003 on Build GRCh38
rs547483692 Chr.2: 226731925-226731925 on Build GRCh38
rs2288587 Chr.2: 226731796-226731796 on Build GRCh38
rs1031725278 Chr.2: 226732033-226732033 on Build GRCh38
rs1047219045 Chr.2: 226731664-226731664 on Build GRCh38
rs563439240 Chr.2: 226732011-226732011 on Build GRCh38
rs536689276 Chr.2: 226731657-226731657 on Build GRCh38
rs887287866 Chr.2: 226731998-226731998 on Build GRCh38
rs887392423 Chr.2: 226731665-226731665 on Build GRCh38
rs1024307536 Chr.2: 226732022-226732022 on Build GRCh38
rs923749942 Chr.2: 226731876-226731876 on Build GRCh38
rs386655835 Chr.2: 226732023-226732024 on Build GRCh38
rs370588757 Chr.2: 226731795-226731795 on Build GRCh38
rs573200584 Chr.2: 226731772-226731772 on Build GRCh38
rs530690231 Chr.2: 226731598-226731598 on Build GRCh38
rs143983427 Chr.2: 226731768-226731769 on Build GRCh38
rs1024403737 Chr.2: 226731841-226731841 on Build GRCh38
rs1039339206 Chr.2: 226731933-226731933 on Build GRCh38
rs868342652 Chr.2: 226731647-226731647 on Build GRCh38
rs1012954897 Chr.2: 226732015-226732015 on Build GRCh38
rs895588861 Chr.2: 226731863-226731863 on Build GRCh38
rs900898396 Chr.2: 226731629-226731629 on Build GRCh38
rs1006205403 Chr.2: 226731763-226731763 on Build GRCh38
rs765159798 Chr.2: 226731668-226731668 on Build GRCh38

View Full Product Details