Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226795284-226795780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752387 c.3312C>T p.F1104F Chr.2: 226795427-226795427 on Build GRCh38
6687390 c.3185G>A p.G1062D Chr.2: 226795554-226795554 on Build GRCh38
6499616 c.3130C>A p.P1044T Chr.2: 226795609-226795609 on Build GRCh38
4952221 c.3354C>A p.P1118P Chr.2: 226795385-226795385 on Build GRCh38
4583266 c.3051A>G p.T1017T Chr.2: 226795688-226795688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200422050 Chr.2: 226795644-226795644 on Build GRCh38
rs768153308 Chr.2: 226795674-226795674 on Build GRCh38
rs767546323 Chr.2: 226795521-226795521 on Build GRCh38
rs202243502 Chr.2: 226795442-226795442 on Build GRCh38
rs1051989771 Chr.2: 226795390-226795390 on Build GRCh38
rs772844464 Chr.2: 226795696-226795696 on Build GRCh38
rs769318852 Chr.2: 226795582-226795582 on Build GRCh38
rs544798746 Chr.2: 226795404-226795404 on Build GRCh38
rs752864956 Chr.2: 226795639-226795639 on Build GRCh38
rs767474165 Chr.2: 226795333-226795334 on Build GRCh38
rs751434622 Chr.2: 226795413-226795413 on Build GRCh38
rs143760685 Chr.2: 226795746-226795746 on Build GRCh38
rs775770356 Chr.2: 226795320-226795320 on Build GRCh38
rs373248004 Chr.2: 226795725-226795725 on Build GRCh38
rs369672495 Chr.2: 226795714-226795714 on Build GRCh38
rs899984257 Chr.2: 226795327-226795327 on Build GRCh38
rs974159919 Chr.2: 226795389-226795389 on Build GRCh38
rs773839567 Chr.2: 226795392-226795392 on Build GRCh38
rs141372270 Chr.2: 226795529-226795529 on Build GRCh38
rs899312279 Chr.2: 226795649-226795649 on Build GRCh38
rs750831376 Chr.2: 226795728-226795728 on Build GRCh38
rs771049966 Chr.2: 226795707-226795707 on Build GRCh38
rs41272723 Chr.2: 226795652-226795652 on Build GRCh38
rs771924555 Chr.2: 226795693-226795693 on Build GRCh38
rs780147733 Chr.2: 226795376-226795376 on Build GRCh38
rs775121566 Chr.2: 226795450-226795450 on Build GRCh38
rs112888661 Chr.2: 226795374-226795374 on Build GRCh38
rs748578334 Chr.2: 226795435-226795435 on Build GRCh38
rs780609564 Chr.2: 226795735-226795735 on Build GRCh38
rs765349547 Chr.2: 226795533-226795533 on Build GRCh38
rs988225202 Chr.2: 226795460-226795460 on Build GRCh38
rs146268112 Chr.2: 226795518-226795518 on Build GRCh38
rs763295813 Chr.2: 226795321-226795321 on Build GRCh38
rs779252822 Chr.2: 226795427-226795427 on Build GRCh38
rs144820119 Chr.2: 226795393-226795393 on Build GRCh38
rs748385460 Chr.2: 226795608-226795608 on Build GRCh38
rs758373069 Chr.2: 226795367-226795367 on Build GRCh38
rs749508598 Chr.2: 226795424-226795424 on Build GRCh38
rs201918946 Chr.2: 226795405-226795405 on Build GRCh38
rs112432659 Chr.2: 226795426-226795426 on Build GRCh38
rs748859206 Chr.2: 226795665-226795665 on Build GRCh38
rs750648397 Chr.2: 226795391-226795392 on Build GRCh38
rs756165343 Chr.2: 226795415-226795415 on Build GRCh38
rs573529594 Chr.2: 226795331-226795331 on Build GRCh38
rs771700786 Chr.2: 226795610-226795610 on Build GRCh38
rs772751270 Chr.2: 226795391-226795391 on Build GRCh38
rs751524439 Chr.2: 226795457-226795457 on Build GRCh38
rs200670773 Chr.2: 226795498-226795498 on Build GRCh38
rs201525785 Chr.2: 226795473-226795473 on Build GRCh38
rs756650445 Chr.2: 226795660-226795660 on Build GRCh38
rs199755602 Chr.2: 226795645-226795645 on Build GRCh38
rs777666532 Chr.2: 226795372-226795372 on Build GRCh38
rs913587902 Chr.2: 226795753-226795753 on Build GRCh38
rs141391800 Chr.2: 226795602-226795602 on Build GRCh38
rs748312477 Chr.2: 226795493-226795493 on Build GRCh38
rs146121894 Chr.2: 226795417-226795417 on Build GRCh38
rs747464908 Chr.2: 226795557-226795557 on Build GRCh38
rs763911795 Chr.2: 226795519-226795519 on Build GRCh38
rs770949631 Chr.2: 226795740-226795740 on Build GRCh38
rs540624592 Chr.2: 226795736-226795736 on Build GRCh38
rs769228929 Chr.2: 226795506-226795506 on Build GRCh38
rs770051216 Chr.2: 226795317-226795317 on Build GRCh38
rs746305011 Chr.2: 226795569-226795569 on Build GRCh38
rs758653179 Chr.2: 226795544-226795544 on Build GRCh38
rs148962208 Chr.2: 226795504-226795504 on Build GRCh38
rs372681714 Chr.2: 226795497-226795497 on Build GRCh38
rs751664998 Chr.2: 226795648-226795648 on Build GRCh38
rs776704743 Chr.2: 226795712-226795712 on Build GRCh38
rs138710153 Chr.2: 226795381-226795381 on Build GRCh38
rs770370233 Chr.2: 226795439-226795439 on Build GRCh38
rs777756375 Chr.2: 226795408-226795408 on Build GRCh38
rs767163776 Chr.2: 226795349-226795349 on Build GRCh38
rs751826393 Chr.2: 226795720-226795720 on Build GRCh38
rs765846492 Chr.2: 226795402-226795402 on Build GRCh38
rs781725718 Chr.2: 226795568-226795568 on Build GRCh38
rs368961254 Chr.2: 226795490-226795490 on Build GRCh38
rs145977916 Chr.2: 226795351-226795351 on Build GRCh38
rs3731594 Chr.2: 226795330-226795330 on Build GRCh38
rs766003841 Chr.2: 226795352-226795352 on Build GRCh38
rs752662327 Chr.2: 226795359-226795359 on Build GRCh38
rs373714010 Chr.2: 226795587-226795587 on Build GRCh38
rs533260174 Chr.2: 226795358-226795358 on Build GRCh38
rs536862124 Chr.2: 226795650-226795650 on Build GRCh38
rs762233329 Chr.2: 226795386-226795386 on Build GRCh38
rs774812399 Chr.2: 226795383-226795383 on Build GRCh38
rs747491337 Chr.2: 226795438-226795438 on Build GRCh38
rs761486707 Chr.2: 226795520-226795520 on Build GRCh38
rs757182552 Chr.2: 226795459-226795459 on Build GRCh38
rs761897696 Chr.2: 226795724-226795724 on Build GRCh38
rs748769427 Chr.2: 226795593-226795593 on Build GRCh38
rs773858412 Chr.2: 226795607-226795607 on Build GRCh38
rs894163217 Chr.2: 226795706-226795706 on Build GRCh38
rs539361581 Chr.2: 226795630-226795630 on Build GRCh38
rs183414071 Chr.2: 226795447-226795447 on Build GRCh38
rs762744077 Chr.2: 226795452-226795452 on Build GRCh38
rs752989640 Chr.2: 226795534-226795534 on Build GRCh38
rs753853576 Chr.2: 226795334-226795336 on Build GRCh38
rs199554914 Chr.2: 226795453-226795453 on Build GRCh38
rs146703195 Chr.2: 226795486-226795486 on Build GRCh38
rs567766900 Chr.2: 226795491-226795491 on Build GRCh38
rs764021548 Chr.2: 226795717-226795717 on Build GRCh38
rs140117962 Chr.2: 226795683-226795683 on Build GRCh38
rs745918297 Chr.2: 226795316-226795316 on Build GRCh38
rs143913301 Chr.2: 226795377-226795377 on Build GRCh38
rs200821911 Chr.2: 226795309-226795309 on Build GRCh38
rs758947835 Chr.2: 226795446-226795446 on Build GRCh38
rs865939502 Chr.2: 226795655-226795656 on Build GRCh38
rs372034600 Chr.2: 226795739-226795739 on Build GRCh38
rs756462316 Chr.2: 226795730-226795730 on Build GRCh38
rs757558816 Chr.2: 226795559-226795559 on Build GRCh38
rs1043936986 Chr.2: 226795323-226795323 on Build GRCh38
rs758177031 Chr.2: 226795407-226795407 on Build GRCh38
rs200142054 Chr.2: 226795333-226795333 on Build GRCh38
rs140150448 Chr.2: 226795436-226795436 on Build GRCh38
rs752537670 Chr.2: 226795406-226795406 on Build GRCh38
rs765435936 Chr.2: 226795374-226795379 on Build GRCh38
rs758845449 Chr.2: 226795496-226795496 on Build GRCh38
rs750968532 Chr.2: 226795657-226795657 on Build GRCh38
rs891382490 Chr.2: 226795542-226795542 on Build GRCh38
rs755959354 Chr.2: 226795613-226795613 on Build GRCh38
rs750320720 Chr.2: 226795528-226795528 on Build GRCh38
rs747700323 Chr.2: 226795692-226795692 on Build GRCh38
rs760515770 Chr.2: 226795703-226795703 on Build GRCh38
rs148328229 Chr.2: 226795550-226795550 on Build GRCh38
rs988057825 Chr.2: 226795750-226795750 on Build GRCh38
rs781408207 Chr.2: 226795474-226795474 on Build GRCh38
rs745452023 Chr.2: 226795505-226795505 on Build GRCh38
rs780511813 Chr.2: 226795664-226795664 on Build GRCh38
rs377465384 Chr.2: 226795758-226795758 on Build GRCh38
rs747866903 Chr.2: 226795742-226795742 on Build GRCh38
rs12999226 Chr.2: 226795635-226795635 on Build GRCh38
rs367762669 Chr.2: 226795747-226795747 on Build GRCh38
rs898476192 Chr.2: 226795643-226795643 on Build GRCh38
rs371306259 Chr.2: 226795357-226795357 on Build GRCh38
rs145050860 Chr.2: 226795499-226795499 on Build GRCh38

View Full Product Details