Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 239048420-239048915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC4
Entrez Gene ID
9759
Gene Name
histone deacetylase 4
Gene Aliases
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
UniGene
Hs.20516
Gene Chromosome Location
Chr.2: 239048168-239401647 on Build GRCh38
Transcripts
NM_006037.3 XM_006712877.3 XM_006712878.3 XM_006712879.3 XM_006712880.3 XM_011512217.2 XM_011512218.2 XM_011512219.2 XM_011512220.2 XM_011512221.1 XM_011512222.2 XM_011512223.2 XM_011512224.2 XM_011512225.2 XM_011512226.2 XM_011512227.2 XM_011512230.1 XM_017005394.1 XM_017005395.1

SNP Detail

SNP ID Chromosome Location
rs561960524 Chr.2: 239048642-239048642 on Build GRCh38
rs534560920 Chr.2: 239048527-239048527 on Build GRCh38
rs895808 Chr.2: 239048664-239048664 on Build GRCh38
rs1039735199 Chr.2: 239048798-239048798 on Build GRCh38
rs75312371 Chr.2: 239048594-239048595 on Build GRCh38
rs774044012 Chr.2: 239048751-239048751 on Build GRCh38
rs188132185 Chr.2: 239048842-239048842 on Build GRCh38
rs905958316 Chr.2: 239048554-239048554 on Build GRCh38
rs997693504 Chr.2: 239048560-239048560 on Build GRCh38
rs989069151 Chr.2: 239048690-239048690 on Build GRCh38
rs190084219 Chr.2: 239048439-239048439 on Build GRCh38
rs962541616 Chr.2: 239048647-239048647 on Build GRCh38
rs1002804505 Chr.2: 239048605-239048605 on Build GRCh38
rs914814175 Chr.2: 239048699-239048699 on Build GRCh38
rs918381402 Chr.2: 239048672-239048672 on Build GRCh38
rs938546796 Chr.2: 239048662-239048665 on Build GRCh38
rs558813131 Chr.2: 239048561-239048564 on Build GRCh38
rs1031460169 Chr.2: 239048566-239048566 on Build GRCh38
rs867499072 Chr.2: 239048774-239048774 on Build GRCh38
rs553209188 Chr.2: 239048561-239048568 on Build GRCh38
rs570459200 Chr.2: 239048498-239048498 on Build GRCh38
rs552249717 Chr.2: 239048473-239048473 on Build GRCh38
rs533402447 Chr.2: 239048459-239048459 on Build GRCh38
rs3683 Chr.2: 239048749-239048749 on Build GRCh38
rs560535182 Chr.2: 239048564-239048565 on Build GRCh38
rs927497352 Chr.2: 239048737-239048737 on Build GRCh38
rs897905368 Chr.2: 239048520-239048520 on Build GRCh38
rs78601632 Chr.2: 239048448-239048448 on Build GRCh38
rs995323570 Chr.2: 239048830-239048830 on Build GRCh38
rs559635949 Chr.2: 239048582-239048583 on Build GRCh38
rs912789458 Chr.2: 239048494-239048494 on Build GRCh38
rs373503880 Chr.2: 239048750-239048750 on Build GRCh38
rs778140876 Chr.2: 239048653-239048653 on Build GRCh38
rs989873949 Chr.2: 239048597-239048597 on Build GRCh38
rs976268587 Chr.2: 239048447-239048447 on Build GRCh38
rs558190217 Chr.2: 239048865-239048865 on Build GRCh38
rs921023407 Chr.2: 239048463-239048463 on Build GRCh38
rs1036912207 Chr.2: 239048819-239048819 on Build GRCh38
rs953790446 Chr.2: 239048654-239048654 on Build GRCh38
rs557008947 Chr.2: 239048738-239048738 on Build GRCh38
rs1044422200 Chr.2: 239048538-239048538 on Build GRCh38
rs866478688 Chr.2: 239048466-239048466 on Build GRCh38
rs995749902 Chr.2: 239048652-239048652 on Build GRCh38
rs1057049438 Chr.2: 239048620-239048620 on Build GRCh38
rs998144456 Chr.2: 239048829-239048829 on Build GRCh38
rs928566034 Chr.2: 239048656-239048657 on Build GRCh38
rs79040455 Chr.2: 239048834-239048834 on Build GRCh38
rs914199705 Chr.2: 239048460-239048460 on Build GRCh38
rs748776694 Chr.2: 239048521-239048521 on Build GRCh38
rs886435936 Chr.2: 239048761-239048761 on Build GRCh38
rs1019409121 Chr.2: 239048889-239048889 on Build GRCh38
rs546635836 Chr.2: 239048572-239048572 on Build GRCh38
rs868751754 Chr.2: 239048859-239048859 on Build GRCh38
rs886466738 Chr.2: 239048628-239048628 on Build GRCh38
rs955732487 Chr.2: 239048449-239048449 on Build GRCh38
rs570044162 Chr.2: 239048590-239048591 on Build GRCh38
rs186474651 Chr.2: 239048787-239048787 on Build GRCh38
rs1015963449 Chr.2: 239048637-239048637 on Build GRCh38
rs181161360 Chr.2: 239048680-239048680 on Build GRCh38
rs564848332 Chr.2: 239048568-239048569 on Build GRCh38
rs914743209 Chr.2: 239048522-239048522 on Build GRCh38
rs898426524 Chr.2: 239048823-239048823 on Build GRCh38
rs953655547 Chr.2: 239048464-239048464 on Build GRCh38
rs201034860 Chr.2: 239048559-239048559 on Build GRCh38
rs3841081 Chr.2: 239048560-239048561 on Build GRCh38
rs1004133124 Chr.2: 239048631-239048631 on Build GRCh38
rs60534011 Chr.2: 239048591-239048594 on Build GRCh38
rs1044559455 Chr.2: 239048722-239048722 on Build GRCh38
rs947537803 Chr.2: 239048719-239048719 on Build GRCh38
rs778988288 Chr.2: 239048873-239048873 on Build GRCh38
rs912212742 Chr.2: 239048671-239048671 on Build GRCh38
rs1008238898 Chr.2: 239048531-239048531 on Build GRCh38

View Full Product Details