Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 239049131-239049664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC4
Entrez Gene ID
9759
Gene Name
histone deacetylase 4
Gene Aliases
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
UniGene
Hs.20516
Gene Chromosome Location
Chr.2: 239048168-239401647 on Build GRCh38
Transcripts
NM_006037.3 XM_006712877.3 XM_006712878.3 XM_006712879.3 XM_006712880.3 XM_011512217.2 XM_011512218.2 XM_011512219.2 XM_011512220.2 XM_011512221.1 XM_011512222.2 XM_011512223.2 XM_011512224.2 XM_011512225.2 XM_011512226.2 XM_011512227.2 XM_011512230.1 XM_017005394.1 XM_017005395.1

SNP Detail

SNP ID Chromosome Location
rs1041287567 Chr.2: 239049515-239049515 on Build GRCh38
rs574731549 Chr.2: 239049458-239049458 on Build GRCh38
rs1018119068 Chr.2: 239049586-239049586 on Build GRCh38
rs764669762 Chr.2: 239049195-239049195 on Build GRCh38
rs777026958 Chr.2: 239049604-239049604 on Build GRCh38
rs1021410140 Chr.2: 239049555-239049555 on Build GRCh38
rs904164991 Chr.2: 239049559-239049559 on Build GRCh38
rs906003079 Chr.2: 239049457-239049464 on Build GRCh38
rs778222061 Chr.2: 239049378-239049378 on Build GRCh38
rs1030827237 Chr.2: 239049610-239049610 on Build GRCh38
rs973536435 Chr.2: 239049411-239049411 on Build GRCh38
rs191721960 Chr.2: 239049551-239049551 on Build GRCh38
rs1001652704 Chr.2: 239049156-239049157 on Build GRCh38
rs1021692450 Chr.2: 239049158-239049158 on Build GRCh38
rs771572425 Chr.2: 239049423-239049423 on Build GRCh38
rs1043249085 Chr.2: 239049435-239049435 on Build GRCh38
rs567627295 Chr.2: 239049543-239049543 on Build GRCh38
rs370700441 Chr.2: 239049617-239049617 on Build GRCh38
rs1049639559 Chr.2: 239049462-239049462 on Build GRCh38
rs1034540182 Chr.2: 239049615-239049615 on Build GRCh38
rs1010399901 Chr.2: 239049541-239049541 on Build GRCh38
rs1034576046 Chr.2: 239049248-239049248 on Build GRCh38
rs762046984 Chr.2: 239049609-239049609 on Build GRCh38
rs992975677 Chr.2: 239049348-239049348 on Build GRCh38
rs113877638 Chr.2: 239049247-239049247 on Build GRCh38
rs183188895 Chr.2: 239049625-239049625 on Build GRCh38
rs1001306397 Chr.2: 239049504-239049504 on Build GRCh38
rs1001161930 Chr.2: 239049614-239049614 on Build GRCh38
rs1025552954 Chr.2: 239049552-239049552 on Build GRCh38
rs947583715 Chr.2: 239049415-239049415 on Build GRCh38
rs546746017 Chr.2: 239049312-239049312 on Build GRCh38
rs758997394 Chr.2: 239049182-239049185 on Build GRCh38
rs932003842 Chr.2: 239049440-239049440 on Build GRCh38
rs943826955 Chr.2: 239049502-239049502 on Build GRCh38
rs923323296 Chr.2: 239049379-239049379 on Build GRCh38
rs911013603 Chr.2: 239049357-239049357 on Build GRCh38
rs933288355 Chr.2: 239049389-239049389 on Build GRCh38
rs891653675 Chr.2: 239049516-239049516 on Build GRCh38
rs557688284 Chr.2: 239049640-239049640 on Build GRCh38
rs539584245 Chr.2: 239049633-239049633 on Build GRCh38
rs959947011 Chr.2: 239049171-239049171 on Build GRCh38
rs575963342 Chr.2: 239049580-239049581 on Build GRCh38
rs964829096 Chr.2: 239049370-239049370 on Build GRCh38
rs960159090 Chr.2: 239049260-239049260 on Build GRCh38
rs889707605 Chr.2: 239049482-239049482 on Build GRCh38
rs1053404517 Chr.2: 239049395-239049395 on Build GRCh38
rs183967576 Chr.2: 239049398-239049398 on Build GRCh38
rs993794314 Chr.2: 239049528-239049528 on Build GRCh38
rs968454434 Chr.2: 239049219-239049219 on Build GRCh38
rs1057270827 Chr.2: 239049513-239049513 on Build GRCh38
rs557829649 Chr.2: 239049606-239049606 on Build GRCh38
rs186834320 Chr.2: 239049544-239049544 on Build GRCh38
rs954727058 Chr.2: 239049613-239049613 on Build GRCh38
rs983913796 Chr.2: 239049345-239049345 on Build GRCh38

View Full Product Details