Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 239051479-239052013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC4
Entrez Gene ID
9759
Gene Name
histone deacetylase 4
Gene Aliases
AHO3, BDMR, HA6116, HD4, HDAC-4, HDAC-A, HDACA
UniGene
Hs.20516
Gene Chromosome Location
Chr.2: 239048168-239401647 on Build GRCh38
Transcripts
NM_006037.3 XM_006712877.3 XM_006712878.3 XM_006712879.3 XM_006712880.3 XM_011512217.2 XM_011512218.2 XM_011512219.2 XM_011512220.2 XM_011512221.1 XM_011512222.2 XM_011512223.2 XM_011512224.2 XM_011512225.2 XM_011512226.2 XM_011512227.2 XM_011512230.1 XM_017005394.1 XM_017005395.1

SNP Detail

SNP ID Chromosome Location
rs1003733995 Chr.2: 239051814-239051814 on Build GRCh38
rs964072033 Chr.2: 239051856-239051856 on Build GRCh38
rs148739450 Chr.2: 239051943-239051943 on Build GRCh38
rs146303968 Chr.2: 239051548-239051548 on Build GRCh38
rs144267489 Chr.2: 239051831-239051831 on Build GRCh38
rs1019654454 Chr.2: 239051921-239051924 on Build GRCh38
rs547945640 Chr.2: 239051844-239051844 on Build GRCh38
rs941905837 Chr.2: 239051636-239051636 on Build GRCh38
rs772286505 Chr.2: 239051714-239051714 on Build GRCh38
rs139462269 Chr.2: 239051796-239051796 on Build GRCh38
rs999261351 Chr.2: 239051712-239051712 on Build GRCh38
rs1023756739 Chr.2: 239051541-239051541 on Build GRCh38
rs867491749 Chr.2: 239051694-239051694 on Build GRCh38
rs923794602 Chr.2: 239051939-239051939 on Build GRCh38
rs1012477853 Chr.2: 239051529-239051529 on Build GRCh38
rs984133665 Chr.2: 239051615-239051615 on Build GRCh38
rs149593864 Chr.2: 239051826-239051826 on Build GRCh38
rs181201320 Chr.2: 239051847-239051847 on Build GRCh38
rs537215473 Chr.2: 239051984-239051984 on Build GRCh38
rs1046119169 Chr.2: 239051988-239051988 on Build GRCh38
rs566582705 Chr.2: 239051861-239051861 on Build GRCh38
rs79925675 Chr.2: 239051758-239051758 on Build GRCh38
rs867084484 Chr.2: 239051564-239051564 on Build GRCh38
rs921752893 Chr.2: 239051690-239051690 on Build GRCh38
rs974623990 Chr.2: 239051857-239051857 on Build GRCh38
rs933125184 Chr.2: 239051913-239051913 on Build GRCh38
rs1028785048 Chr.2: 239051912-239051912 on Build GRCh38
rs570335354 Chr.2: 239051959-239051959 on Build GRCh38
rs906799669 Chr.2: 239051971-239051971 on Build GRCh38
rs748631876 Chr.2: 239051592-239051592 on Build GRCh38
rs566640470 Chr.2: 239051914-239051917 on Build GRCh38
rs746524670 Chr.2: 239051910-239051913 on Build GRCh38
rs954107849 Chr.2: 239051526-239051526 on Build GRCh38
rs773389856 Chr.2: 239051732-239051732 on Build GRCh38
rs184968755 Chr.2: 239051907-239051907 on Build GRCh38
rs774707683 Chr.2: 239051569-239051569 on Build GRCh38
rs958280260 Chr.2: 239051963-239051963 on Build GRCh38
rs529709825 Chr.2: 239051735-239051736 on Build GRCh38
rs796844541 Chr.2: 239051848-239051849 on Build GRCh38
rs918683288 Chr.2: 239051584-239051584 on Build GRCh38
rs924155543 Chr.2: 239051701-239051701 on Build GRCh38
rs987333923 Chr.2: 239051920-239051920 on Build GRCh38
rs1045703811 Chr.2: 239051968-239051968 on Build GRCh38
rs951481965 Chr.2: 239051830-239051830 on Build GRCh38
rs1027484077 Chr.2: 239051508-239051508 on Build GRCh38
rs985458832 Chr.2: 239051540-239051540 on Build GRCh38
rs893439431 Chr.2: 239051951-239051951 on Build GRCh38
rs909911067 Chr.2: 239051618-239051618 on Build GRCh38
rs1053574168 Chr.2: 239051724-239051724 on Build GRCh38
rs549327637 Chr.2: 239051703-239051703 on Build GRCh38
rs1039007113 Chr.2: 239051663-239051663 on Build GRCh38
rs933176413 Chr.2: 239051700-239051700 on Build GRCh38
rs539416030 Chr.2: 239051808-239051808 on Build GRCh38
rs1051910331 Chr.2: 239051702-239051702 on Build GRCh38
rs921874347 Chr.2: 239051905-239051905 on Build GRCh38
rs376319727 Chr.2: 239051940-239051940 on Build GRCh38

View Full Product Details