Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240717124-240717631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
UniGene
Hs.516802
Gene Chromosome Location
Chr.2: 240713764-240821025 on Build GRCh38
Transcripts
NM_001244008.1 NM_001320705.1 NM_004321.6 XM_005247022.1 XM_005247024.1 XM_005247027.1 XM_005247028.2 XM_006712605.1 XM_011511364.1 XM_011511365.1 XM_011511366.2 XM_011511367.1 XM_017004383.1 XM_017004384.1 XM_017004385.1 XM_017004386.1 XM_017004387.1 XM_017004388.1 XM_017004389.1 XM_017004390.1 XM_017004391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4859414 c.5369G>A p.R1790Q Chr.2: 240717371-240717371 on Build GRCh38
5465494 c.5368C>T p.R1790W Chr.2: 240717372-240717372 on Build GRCh38
5465495 c.5065C>T p.R1689W Chr.2: 240717372-240717372 on Build GRCh38
720897 c.5066G>A p.R1689Q Chr.2: 240717371-240717371 on Build GRCh38
6091211 c.5354G>T p.R1785M Chr.2: 240717386-240717386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562109203 Chr.2: 240717255-240717255 on Build GRCh38
rs1003986227 Chr.2: 240717522-240717522 on Build GRCh38
rs762837283 Chr.2: 240717349-240717349 on Build GRCh38
rs531276835 Chr.2: 240717341-240717341 on Build GRCh38
rs776542312 Chr.2: 240717336-240717336 on Build GRCh38
rs1027993154 Chr.2: 240717557-240717557 on Build GRCh38
rs56051420 Chr.2: 240717280-240717280 on Build GRCh38
rs748374545 Chr.2: 240717324-240717324 on Build GRCh38
rs1061113 Chr.2: 240717256-240717256 on Build GRCh38
rs1015454204 Chr.2: 240717295-240717295 on Build GRCh38
rs886182139 Chr.2: 240717213-240717213 on Build GRCh38
rs769679424 Chr.2: 240717439-240717439 on Build GRCh38
rs987292027 Chr.2: 240717261-240717261 on Build GRCh38
rs557161979 Chr.2: 240717173-240717173 on Build GRCh38
rs986918139 Chr.2: 240717365-240717365 on Build GRCh38
rs907521890 Chr.2: 240717603-240717603 on Build GRCh38
rs372244073 Chr.2: 240717340-240717340 on Build GRCh38
rs772148613 Chr.2: 240717326-240717326 on Build GRCh38
rs954396239 Chr.2: 240717180-240717180 on Build GRCh38
rs898410979 Chr.2: 240717536-240717536 on Build GRCh38
rs949782720 Chr.2: 240717350-240717350 on Build GRCh38
rs558374091 Chr.2: 240717606-240717606 on Build GRCh38
rs905642704 Chr.2: 240717399-240717399 on Build GRCh38
rs1019361196 Chr.2: 240717581-240717581 on Build GRCh38
rs964200437 Chr.2: 240717491-240717491 on Build GRCh38
rs546283159 Chr.2: 240717492-240717492 on Build GRCh38
rs770633741 Chr.2: 240717367-240717367 on Build GRCh38
rs369622428 Chr.2: 240717429-240717429 on Build GRCh38
rs748416328 Chr.2: 240717401-240717401 on Build GRCh38
rs149656076 Chr.2: 240717604-240717604 on Build GRCh38
rs889424629 Chr.2: 240717566-240717566 on Build GRCh38
rs575986280 Chr.2: 240717586-240717586 on Build GRCh38
rs776337224 Chr.2: 240717467-240717467 on Build GRCh38
rs764018540 Chr.2: 240717456-240717456 on Build GRCh38
rs548422454 Chr.2: 240717259-240717259 on Build GRCh38
rs535790499 Chr.2: 240717521-240717521 on Build GRCh38
rs777787746 Chr.2: 240717422-240717422 on Build GRCh38
rs564598846 Chr.2: 240717203-240717203 on Build GRCh38
rs1033330948 Chr.2: 240717251-240717251 on Build GRCh38
rs780821029 Chr.2: 240717479-240717479 on Build GRCh38
rs540308680 Chr.2: 240717239-240717239 on Build GRCh38
rs563350862 Chr.2: 240717338-240717338 on Build GRCh38
rs779232085 Chr.2: 240717235-240717235 on Build GRCh38
rs1024104800 Chr.2: 240717236-240717236 on Build GRCh38
rs760590728 Chr.2: 240717385-240717387 on Build GRCh38
rs775303081 Chr.2: 240717351-240717371 on Build GRCh38
rs771850738 Chr.2: 240717341-240717341 on Build GRCh38
rs758806789 Chr.2: 240717410-240717410 on Build GRCh38
rs751236889 Chr.2: 240717355-240717355 on Build GRCh38
rs895227286 Chr.2: 240717248-240717248 on Build GRCh38
rs747320289 Chr.2: 240717428-240717428 on Build GRCh38
rs189282597 Chr.2: 240717469-240717469 on Build GRCh38
rs970315998 Chr.2: 240717243-240717243 on Build GRCh38
rs377466581 Chr.2: 240717164-240717166 on Build GRCh38
rs375509312 Chr.2: 240717372-240717372 on Build GRCh38
rs1001759107 Chr.2: 240717413-240717413 on Build GRCh38
rs573357958 Chr.2: 240717238-240717238 on Build GRCh38
rs756824696 Chr.2: 240717370-240717370 on Build GRCh38
rs986874899 Chr.2: 240717477-240717477 on Build GRCh38
rs534411093 Chr.2: 240717486-240717486 on Build GRCh38
rs1001432481 Chr.2: 240717249-240717249 on Build GRCh38
rs974297206 Chr.2: 240717308-240717308 on Build GRCh38
rs942876187 Chr.2: 240717302-240717302 on Build GRCh38
rs1007989919 Chr.2: 240717578-240717578 on Build GRCh38
rs921006450 Chr.2: 240717174-240717174 on Build GRCh38
rs777631361 Chr.2: 240717559-240717559 on Build GRCh38
rs920170527 Chr.2: 240717309-240717309 on Build GRCh38
rs779468930 Chr.2: 240717314-240717314 on Build GRCh38
rs920102677 Chr.2: 240717513-240717513 on Build GRCh38
rs747005832 Chr.2: 240717332-240717332 on Build GRCh38
rs1055639642 Chr.2: 240717244-240717244 on Build GRCh38
rs186456890 Chr.2: 240717221-240717221 on Build GRCh38
rs181750425 Chr.2: 240717496-240717496 on Build GRCh38
rs898802782 Chr.2: 240717187-240717187 on Build GRCh38
rs911369171 Chr.2: 240717290-240717290 on Build GRCh38
rs776738212 Chr.2: 240717431-240717431 on Build GRCh38
rs368053227 Chr.2: 240717335-240717335 on Build GRCh38
rs995796841 Chr.2: 240717540-240717540 on Build GRCh38
rs957295332 Chr.2: 240717253-240717253 on Build GRCh38
rs775518990 Chr.2: 240717441-240717441 on Build GRCh38
rs1046890706 Chr.2: 240717497-240717497 on Build GRCh38
rs779509876 Chr.2: 240717385-240717385 on Build GRCh38
rs570255531 Chr.2: 240717481-240717481 on Build GRCh38
rs755583402 Chr.2: 240717381-240717381 on Build GRCh38
rs763602305 Chr.2: 240717354-240717354 on Build GRCh38
rs1047424971 Chr.2: 240717204-240717204 on Build GRCh38
rs557605169 Chr.2: 240717552-240717552 on Build GRCh38
rs375874661 Chr.2: 240717329-240717329 on Build GRCh38
rs936642381 Chr.2: 240717240-240717240 on Build GRCh38
rs930262089 Chr.2: 240717323-240717323 on Build GRCh38
rs983015531 Chr.2: 240717524-240717524 on Build GRCh38
rs371994677 Chr.2: 240717432-240717432 on Build GRCh38

View Full Product Details