Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240719639-240720134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF1A
Entrez Gene ID
547
Gene Name
kinesin family member 1A
Gene Aliases
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
UniGene
Hs.516802
Gene Chromosome Location
Chr.2: 240713764-240821025 on Build GRCh38
Transcripts
NM_001244008.1 NM_001320705.1 NM_004321.6 XM_005247022.1 XM_005247024.1 XM_005247027.1 XM_005247028.2 XM_006712605.1 XM_011511364.1 XM_011511365.1 XM_011511366.2 XM_011511367.1 XM_017004383.1 XM_017004384.1 XM_017004385.1 XM_017004386.1 XM_017004387.1 XM_017004388.1 XM_017004389.1 XM_017004390.1 XM_017004391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6835889 c.4951G>A p.A1651T Chr.2: 240719844-240719844 on Build GRCh38
3910004 c.4570C>T p.P1524S Chr.2: 240719922-240719922 on Build GRCh38
6322544 c.4978C>A p.P1660T Chr.2: 240719817-240719817 on Build GRCh38
6091209 c.4909G>A p.E1637K Chr.2: 240719886-240719886 on Build GRCh38
6544136 c.4953C>A p.A1651A Chr.2: 240719842-240719842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138847355 Chr.2: 240719758-240719758 on Build GRCh38
rs756724937 Chr.2: 240719663-240719663 on Build GRCh38
rs769695239 Chr.2: 240719965-240719965 on Build GRCh38
rs191428830 Chr.2: 240719906-240719906 on Build GRCh38
rs776558618 Chr.2: 240719952-240719952 on Build GRCh38
rs750942838 Chr.2: 240719918-240719918 on Build GRCh38
rs1050346679 Chr.2: 240720056-240720056 on Build GRCh38
rs184939069 Chr.2: 240719824-240719824 on Build GRCh38
rs956276810 Chr.2: 240719837-240719837 on Build GRCh38
rs377034962 Chr.2: 240720011-240720011 on Build GRCh38
rs545087575 Chr.2: 240719726-240719726 on Build GRCh38
rs751768839 Chr.2: 240719767-240719768 on Build GRCh38
rs200317988 Chr.2: 240719789-240719789 on Build GRCh38
rs775849159 Chr.2: 240719754-240719754 on Build GRCh38
rs1048093628 Chr.2: 240719911-240719911 on Build GRCh38
rs979195520 Chr.2: 240719985-240719985 on Build GRCh38
rs797045653 Chr.2: 240719773-240719773 on Build GRCh38
rs373762639 Chr.2: 240719755-240719755 on Build GRCh38
rs377435468 Chr.2: 240719919-240719919 on Build GRCh38
rs764970503 Chr.2: 240719820-240719820 on Build GRCh38
rs201119646 Chr.2: 240719797-240719797 on Build GRCh38
rs372521322 Chr.2: 240719926-240719926 on Build GRCh38
rs556405858 Chr.2: 240719958-240719958 on Build GRCh38
rs200623445 Chr.2: 240719724-240719724 on Build GRCh38
rs754474100 Chr.2: 240719744-240719744 on Build GRCh38
rs34495497 Chr.2: 240719871-240719871 on Build GRCh38
rs992259915 Chr.2: 240719818-240719818 on Build GRCh38
rs201418175 Chr.2: 240719920-240719920 on Build GRCh38
rs554141220 Chr.2: 240720078-240720078 on Build GRCh38
rs1037734916 Chr.2: 240720074-240720074 on Build GRCh38
rs923205412 Chr.2: 240720038-240720038 on Build GRCh38
rs969057533 Chr.2: 240720112-240720112 on Build GRCh38
rs754893576 Chr.2: 240719796-240719796 on Build GRCh38
rs947853779 Chr.2: 240719682-240719682 on Build GRCh38
rs896037900 Chr.2: 240720084-240720084 on Build GRCh38
rs377032453 Chr.2: 240719886-240719886 on Build GRCh38
rs774256831 Chr.2: 240719778-240719778 on Build GRCh38
rs369475699 Chr.2: 240719843-240719843 on Build GRCh38
rs61561981 Chr.2: 240720111-240720111 on Build GRCh38
rs767301813 Chr.2: 240719782-240719782 on Build GRCh38
rs202164471 Chr.2: 240719785-240719785 on Build GRCh38
rs760657742 Chr.2: 240719880-240719880 on Build GRCh38
rs183768567 Chr.2: 240720089-240720089 on Build GRCh38
rs573887234 Chr.2: 240720005-240720005 on Build GRCh38
rs755324484 Chr.2: 240719862-240719864 on Build GRCh38
rs567604039 Chr.2: 240719921-240719921 on Build GRCh38
rs772261807 Chr.2: 240719872-240719872 on Build GRCh38
rs750065067 Chr.2: 240719689-240719689 on Build GRCh38
rs766136089 Chr.2: 240719881-240719881 on Build GRCh38
rs376581099 Chr.2: 240719869-240719869 on Build GRCh38
rs766631673 Chr.2: 240719725-240719726 on Build GRCh38
rs556381441 Chr.2: 240719699-240719699 on Build GRCh38
rs764884973 Chr.2: 240719664-240719664 on Build GRCh38
rs199557318 Chr.2: 240719810-240719810 on Build GRCh38
rs553614862 Chr.2: 240720034-240720034 on Build GRCh38
rs772025810 Chr.2: 240719939-240719939 on Build GRCh38
rs576331264 Chr.2: 240720083-240720083 on Build GRCh38
rs377748137 Chr.2: 240720076-240720076 on Build GRCh38
rs200141437 Chr.2: 240719868-240719868 on Build GRCh38
rs112808596 Chr.2: 240719844-240719844 on Build GRCh38
rs377641118 Chr.2: 240719980-240719980 on Build GRCh38
rs781567176 Chr.2: 240719977-240719990 on Build GRCh38
rs1039110469 Chr.2: 240720102-240720102 on Build GRCh38
rs925078759 Chr.2: 240720000-240720000 on Build GRCh38
rs370290675 Chr.2: 240719749-240719749 on Build GRCh38
rs772628882 Chr.2: 240720072-240720072 on Build GRCh38
rs748196598 Chr.2: 240719857-240719857 on Build GRCh38
rs960609527 Chr.2: 240719954-240719954 on Build GRCh38
rs992844539 Chr.2: 240720058-240720058 on Build GRCh38
rs368699290 Chr.2: 240720070-240720070 on Build GRCh38
rs896301370 Chr.2: 240719898-240719898 on Build GRCh38
rs538750744 Chr.2: 240719979-240719979 on Build GRCh38
rs762714105 Chr.2: 240719974-240719974 on Build GRCh38
rs1013589761 Chr.2: 240720088-240720088 on Build GRCh38
rs201747520 Chr.2: 240719747-240719747 on Build GRCh38
rs993680692 Chr.2: 240720077-240720077 on Build GRCh38
rs372226807 Chr.2: 240719811-240719811 on Build GRCh38
rs187810230 Chr.2: 240719878-240719878 on Build GRCh38
rs988418242 Chr.2: 240720024-240720024 on Build GRCh38
rs763752868 Chr.2: 240719915-240719915 on Build GRCh38
rs760037264 Chr.2: 240720039-240720039 on Build GRCh38
rs755376572 Chr.2: 240719924-240719924 on Build GRCh38
rs746442427 Chr.2: 240719841-240719841 on Build GRCh38
rs748464097 Chr.2: 240719929-240719929 on Build GRCh38
rs770668229 Chr.2: 240719686-240719686 on Build GRCh38
rs199930943 Chr.2: 240719746-240719746 on Build GRCh38
rs759393763 Chr.2: 240719887-240719887 on Build GRCh38
rs202072706 Chr.2: 240719907-240719907 on Build GRCh38
rs779791566 Chr.2: 240719748-240719748 on Build GRCh38
rs6437368 Chr.2: 240719951-240719951 on Build GRCh38
rs376658420 Chr.2: 240719840-240719840 on Build GRCh38
rs189618632 Chr.2: 240720110-240720110 on Build GRCh38
rs774989963 Chr.2: 240719970-240719970 on Build GRCh38
rs761974755 Chr.2: 240719780-240719780 on Build GRCh38
rs903766813 Chr.2: 240719702-240719702 on Build GRCh38
rs1030522378 Chr.2: 240719943-240719943 on Build GRCh38
rs774835128 Chr.2: 240720013-240720013 on Build GRCh38
rs777874917 Chr.2: 240719949-240719949 on Build GRCh38
rs571434431 Chr.2: 240719978-240719978 on Build GRCh38

View Full Product Details