Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67329503-67330013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB2
Entrez Gene ID
3595
Gene Name
interleukin 12 receptor subunit beta 2
Gene Aliases
-
UniGene
Hs.479347
Gene Chromosome Location
Chr.1: 67307351-67397730 on Build GRCh38
Transcripts
NM_001258214.1 NM_001258215.1 NM_001258216.1 NM_001319233.1 NM_001559.2 NR_047583.1 NR_047584.1 XM_005270825.2 XM_005270827.2 XM_005270828.3 XM_006710617.2 XM_011541383.2 XM_011541384.2 XM_017001202.1 XM_017001203.1 XM_017001204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1560516 c.779G>A p.R260Q Chr.1: 67329701-67329701 on Build GRCh38
74997 c.757C>A p.L253M Chr.1: 67329679-67329679 on Build GRCh38
4009655 c.752T>C p.L251P Chr.1: 67329674-67329674 on Build GRCh38
287192 c.680C>T p.P227L Chr.1: 67329602-67329602 on Build GRCh38
161699 c.800G>A p.W267* Chr.1: 67329722-67329722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779933372 Chr.1: 67329651-67329651 on Build GRCh38
rs759979028 Chr.1: 67329608-67329608 on Build GRCh38
rs1030769240 Chr.1: 67329807-67329807 on Build GRCh38
rs752675781 Chr.1: 67329777-67329777 on Build GRCh38
rs749220719 Chr.1: 67329573-67329573 on Build GRCh38
rs368134265 Chr.1: 67329632-67329632 on Build GRCh38
rs141167354 Chr.1: 67329713-67329713 on Build GRCh38
rs1495963 Chr.1: 67329636-67329636 on Build GRCh38
rs2307149 Chr.1: 67329767-67329767 on Build GRCh38
rs931765780 Chr.1: 67329860-67329860 on Build GRCh38
rs765685936 Chr.1: 67329717-67329717 on Build GRCh38
rs771574201 Chr.1: 67329586-67329586 on Build GRCh38
rs1036632013 Chr.1: 67329933-67329933 on Build GRCh38
rs546129313 Chr.1: 67329783-67329783 on Build GRCh38
rs890712622 Chr.1: 67329575-67329575 on Build GRCh38
rs751790078 Chr.1: 67329737-67329737 on Build GRCh38
rs140525594 Chr.1: 67329896-67329896 on Build GRCh38
rs1024699416 Chr.1: 67329649-67329649 on Build GRCh38
rs373809198 Chr.1: 67329751-67329751 on Build GRCh38
rs765307336 Chr.1: 67329610-67329610 on Build GRCh38
rs746659410 Chr.1: 67329700-67329700 on Build GRCh38
rs564937204 Chr.1: 67329673-67329673 on Build GRCh38
rs754796629 Chr.1: 67329765-67329765 on Build GRCh38
rs773929428 Chr.1: 67329576-67329576 on Build GRCh38
rs746407536 Chr.1: 67329805-67329805 on Build GRCh38
rs779507508 Chr.1: 67329563-67329563 on Build GRCh38
rs201771864 Chr.1: 67329629-67329629 on Build GRCh38
rs147313559 Chr.1: 67329604-67329604 on Build GRCh38
rs777500826 Chr.1: 67329631-67329631 on Build GRCh38
rs749238823 Chr.1: 67329657-67329657 on Build GRCh38
rs745604233 Chr.1: 67329560-67329560 on Build GRCh38
rs370130487 Chr.1: 67329728-67329728 on Build GRCh38
rs779603836 Chr.1: 67329803-67329803 on Build GRCh38
rs1039526799 Chr.1: 67329818-67329818 on Build GRCh38
rs747863379 Chr.1: 67329584-67329584 on Build GRCh38
rs764234305 Chr.1: 67329729-67329729 on Build GRCh38
rs911553075 Chr.1: 67329922-67329922 on Build GRCh38
rs906187252 Chr.1: 67329925-67329925 on Build GRCh38
rs762985506 Chr.1: 67329719-67329719 on Build GRCh38
rs776046469 Chr.1: 67329718-67329718 on Build GRCh38
rs374006496 Chr.1: 67329602-67329602 on Build GRCh38
rs994897196 Chr.1: 67329687-67329687 on Build GRCh38
rs530095686 Chr.1: 67329786-67329786 on Build GRCh38
rs751779031 Chr.1: 67329638-67329638 on Build GRCh38
rs545150086 Chr.1: 67329664-67329664 on Build GRCh38
rs778586536 Chr.1: 67329671-67329671 on Build GRCh38
rs200772326 Chr.1: 67329637-67329637 on Build GRCh38
rs539750407 Chr.1: 67329743-67329743 on Build GRCh38
rs897966480 Chr.1: 67329938-67329938 on Build GRCh38
rs970414661 Chr.1: 67329666-67329666 on Build GRCh38
rs140336744 Chr.1: 67329603-67329603 on Build GRCh38
rs771578695 Chr.1: 67329688-67329688 on Build GRCh38
rs930784998 Chr.1: 67329960-67329960 on Build GRCh38
rs199980983 Chr.1: 67329564-67329564 on Build GRCh38
rs944900711 Chr.1: 67329927-67329927 on Build GRCh38
rs1000829345 Chr.1: 67329592-67329592 on Build GRCh38
rs181692327 Chr.1: 67329863-67329863 on Build GRCh38
rs914981375 Chr.1: 67329850-67329850 on Build GRCh38
rs185002143 Chr.1: 67329689-67329689 on Build GRCh38
rs936490110 Chr.1: 67329842-67329842 on Build GRCh38
rs956553404 Chr.1: 67329820-67329820 on Build GRCh38
rs746542628 Chr.1: 67329760-67329760 on Build GRCh38
rs890520408 Chr.1: 67329550-67329550 on Build GRCh38
rs144347248 Chr.1: 67329645-67329645 on Build GRCh38
rs76664046 Chr.1: 67329809-67329809 on Build GRCh38
rs760069194 Chr.1: 67329716-67329716 on Build GRCh38
rs771299847 Chr.1: 67329701-67329701 on Build GRCh38
rs899689458 Chr.1: 67329827-67329827 on Build GRCh38
rs1009192561 Chr.1: 67329891-67329891 on Build GRCh38
rs190197815 Chr.1: 67329849-67329849 on Build GRCh38

View Full Product Details