Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3749822-3750359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA12B
Entrez Gene ID
116835
Gene Name
heat shock protein family A (Hsp70) member 12B
Gene Aliases
C20orf60
UniGene
Hs.516854
Gene Chromosome Location
Chr.20: 3732670-3753111 on Build GRCh38
Transcripts
NM_001197327.1 NM_001318322.1 NM_052970.4 XM_011529151.2 XM_017027632.1 XM_017027633.1 XM_017027634.1 XM_017027635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645396 c.1215G>A p.A405A Chr.20: 3750141-3750141 on Build GRCh38
723737 c.1063G>C p.G355R Chr.20: 3749989-3749989 on Build GRCh38
4637017 c.1097G>A p.R366H Chr.20: 3750023-3750023 on Build GRCh38
6682553 c.1181G>A p.R394H Chr.20: 3750107-3750107 on Build GRCh38
5517235 c.1227G>A p.S409S Chr.20: 3750153-3750153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34259366 Chr.20: 3749985-3749985 on Build GRCh38
rs774485093 Chr.20: 3749956-3749956 on Build GRCh38
rs368015429 Chr.20: 3749901-3749901 on Build GRCh38
rs763595452 Chr.20: 3750279-3750279 on Build GRCh38
rs770254202 Chr.20: 3750269-3750269 on Build GRCh38
rs753231490 Chr.20: 3749866-3749867 on Build GRCh38
rs756031970 Chr.20: 3750134-3750134 on Build GRCh38
rs750810788 Chr.20: 3749966-3749966 on Build GRCh38
rs374110537 Chr.20: 3749896-3749896 on Build GRCh38
rs535158079 Chr.20: 3750287-3750287 on Build GRCh38
rs565893726 Chr.20: 3750314-3750314 on Build GRCh38
rs778322208 Chr.20: 3749983-3749983 on Build GRCh38
rs755874590 Chr.20: 3750227-3750227 on Build GRCh38
rs558130978 Chr.20: 3749860-3749860 on Build GRCh38
rs776646116 Chr.20: 3749934-3749934 on Build GRCh38
rs778361086 Chr.20: 3749844-3749844 on Build GRCh38
rs73895608 Chr.20: 3750257-3750257 on Build GRCh38
rs927096811 Chr.20: 3749953-3749953 on Build GRCh38
rs139310351 Chr.20: 3750137-3750137 on Build GRCh38
rs767758993 Chr.20: 3749960-3749960 on Build GRCh38
rs372363417 Chr.20: 3750136-3750136 on Build GRCh38
rs1010308813 Chr.20: 3749903-3749903 on Build GRCh38
rs944947114 Chr.20: 3750066-3750066 on Build GRCh38
rs769196158 Chr.20: 3750026-3750026 on Build GRCh38
rs781177119 Chr.20: 3750132-3750132 on Build GRCh38
rs767837672 Chr.20: 3749886-3749886 on Build GRCh38
rs538311690 Chr.20: 3750022-3750022 on Build GRCh38
rs781125257 Chr.20: 3750014-3750014 on Build GRCh38
rs373399946 Chr.20: 3750273-3750273 on Build GRCh38
rs754387190 Chr.20: 3749904-3749904 on Build GRCh38
rs922155622 Chr.20: 3750301-3750301 on Build GRCh38
rs771021373 Chr.20: 3749929-3749929 on Build GRCh38
rs774344821 Chr.20: 3750031-3750031 on Build GRCh38
rs773133594 Chr.20: 3749870-3749870 on Build GRCh38
rs765725768 Chr.20: 3750093-3750093 on Build GRCh38
rs777756080 Chr.20: 3749926-3749926 on Build GRCh38
rs368513665 Chr.20: 3750120-3750120 on Build GRCh38
rs762086377 Chr.20: 3749882-3749882 on Build GRCh38
rs492010 Chr.20: 3749955-3749955 on Build GRCh38
rs763089289 Chr.20: 3749876-3749876 on Build GRCh38
rs745657847 Chr.20: 3750133-3750133 on Build GRCh38
rs760872422 Chr.20: 3750073-3750073 on Build GRCh38
rs111540710 Chr.20: 3750069-3750069 on Build GRCh38
rs754875285 Chr.20: 3750248-3750248 on Build GRCh38
rs746748857 Chr.20: 3749927-3749927 on Build GRCh38
rs1005081550 Chr.20: 3750177-3750177 on Build GRCh38
rs570002442 Chr.20: 3750222-3750222 on Build GRCh38
rs773362896 Chr.20: 3750068-3750068 on Build GRCh38
rs754523135 Chr.20: 3749888-3749915 on Build GRCh38
rs1054810962 Chr.20: 3750156-3750156 on Build GRCh38
rs372853784 Chr.20: 3750063-3750063 on Build GRCh38
rs776009719 Chr.20: 3750201-3750201 on Build GRCh38
rs200973186 Chr.20: 3750231-3750231 on Build GRCh38
rs751677682 Chr.20: 3749880-3749880 on Build GRCh38
rs892477609 Chr.20: 3749890-3749890 on Build GRCh38
rs775730976 Chr.20: 3749874-3749874 on Build GRCh38
rs773272336 Chr.20: 3750153-3750153 on Build GRCh38
rs752178720 Chr.20: 3750111-3750111 on Build GRCh38
rs966120130 Chr.20: 3749931-3749931 on Build GRCh38
rs758006104 Chr.20: 3749987-3749987 on Build GRCh38
rs972777671 Chr.20: 3750321-3750321 on Build GRCh38
rs763110677 Chr.20: 3749938-3749938 on Build GRCh38
rs530230071 Chr.20: 3750179-3750179 on Build GRCh38
rs769739525 Chr.20: 3750030-3750030 on Build GRCh38
rs777524890 Chr.20: 3749992-3749992 on Build GRCh38
rs952270997 Chr.20: 3749958-3749958 on Build GRCh38
rs199719933 Chr.20: 3750054-3750054 on Build GRCh38
rs532250171 Chr.20: 3749971-3749971 on Build GRCh38
rs140891573 Chr.20: 3749923-3749923 on Build GRCh38
rs763542746 Chr.20: 3750210-3750210 on Build GRCh38
rs370598013 Chr.20: 3750221-3750221 on Build GRCh38
rs370227269 Chr.20: 3749970-3749970 on Build GRCh38
rs971299591 Chr.20: 3749917-3749917 on Build GRCh38
rs771173094 Chr.20: 3750267-3750267 on Build GRCh38
rs745863915 Chr.20: 3750028-3750028 on Build GRCh38
rs763765071 Chr.20: 3750211-3750211 on Build GRCh38
rs909534809 Chr.20: 3749993-3749993 on Build GRCh38
rs751082743 Chr.20: 3750215-3750215 on Build GRCh38
rs141564153 Chr.20: 3750185-3750185 on Build GRCh38
rs995427677 Chr.20: 3749859-3749859 on Build GRCh38
rs962319981 Chr.20: 3750283-3750283 on Build GRCh38
rs748187474 Chr.20: 3750140-3750140 on Build GRCh38
rs771119929 Chr.20: 3750169-3750169 on Build GRCh38
rs747821198 Chr.20: 3749922-3749922 on Build GRCh38
rs144840007 Chr.20: 3750122-3750122 on Build GRCh38
rs747041765 Chr.20: 3750264-3750264 on Build GRCh38
rs375725643 Chr.20: 3750087-3750087 on Build GRCh38
rs780491469 Chr.20: 3749897-3749897 on Build GRCh38
rs367586620 Chr.20: 3750058-3750058 on Build GRCh38
rs369126307 Chr.20: 3750268-3750268 on Build GRCh38
rs754400404 Chr.20: 3749972-3749972 on Build GRCh38
rs979776341 Chr.20: 3750202-3750202 on Build GRCh38
rs750755788 Chr.20: 3749893-3749893 on Build GRCh38
rs747268390 Chr.20: 3750162-3750162 on Build GRCh38
rs757023154 Chr.20: 3750220-3750220 on Build GRCh38
rs766795451 Chr.20: 3750084-3750084 on Build GRCh38
rs778471854 Chr.20: 3749921-3749921 on Build GRCh38
rs1019418023 Chr.20: 3749913-3749914 on Build GRCh38
rs1008200723 Chr.20: 3749902-3749902 on Build GRCh38
rs1024427562 Chr.20: 3749915-3749915 on Build GRCh38
rs971077838 Chr.20: 3750193-3750193 on Build GRCh38
rs768680111 Chr.20: 3749941-3749941 on Build GRCh38
rs939651041 Chr.20: 3750037-3750037 on Build GRCh38
rs757855596 Chr.20: 3750113-3750113 on Build GRCh38
rs957276054 Chr.20: 3750223-3750223 on Build GRCh38
rs11696714 Chr.20: 3749925-3749925 on Build GRCh38
rs755504407 Chr.20: 3749974-3749974 on Build GRCh38
rs746807954 Chr.20: 3750003-3750003 on Build GRCh38
rs760009470 Chr.20: 3750090-3750090 on Build GRCh38
rs62206555 Chr.20: 3749877-3749877 on Build GRCh38
rs867453305 Chr.20: 3750023-3750023 on Build GRCh38
rs151101921 Chr.20: 3749845-3749845 on Build GRCh38
rs974874329 Chr.20: 3749950-3749950 on Build GRCh38
rs200034409 Chr.20: 3749943-3749943 on Build GRCh38
rs761981511 Chr.20: 3749959-3749959 on Build GRCh38
rs1039014694 Chr.20: 3750299-3750299 on Build GRCh38
rs760730514 Chr.20: 3749871-3749871 on Build GRCh38
rs553187145 Chr.20: 3750297-3750297 on Build GRCh38
rs759904018 Chr.20: 3750184-3750184 on Build GRCh38
rs770966334 Chr.20: 3749872-3749872 on Build GRCh38
rs771819791 Chr.20: 3749863-3749863 on Build GRCh38
rs763853731 Chr.20: 3750114-3750114 on Build GRCh38
rs6139196 Chr.20: 3750007-3750007 on Build GRCh38
rs944687067 Chr.20: 3750276-3750276 on Build GRCh38
rs753876891 Chr.20: 3750245-3750245 on Build GRCh38

View Full Product Details