Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45093429-45093939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNS1
Entrez Gene ID
3787
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Gene Aliases
Kv9.1
UniGene
Hs.117780
Gene Chromosome Location
Chr.20: 45091214-45101112 on Build GRCh38
Transcripts
NM_001322799.1 NM_002251.4 XM_017027846.1

SNP Detail

SNP ID Chromosome Location
rs540046984 Chr.20: 45093660-45093660 on Build GRCh38
rs1018180659 Chr.20: 45093593-45093593 on Build GRCh38
rs972885994 Chr.20: 45093530-45093530 on Build GRCh38
rs772238266 Chr.20: 45093538-45093538 on Build GRCh38
rs749815677 Chr.20: 45093712-45093712 on Build GRCh38
rs6031988 Chr.20: 45093488-45093488 on Build GRCh38
rs749486544 Chr.20: 45093455-45093455 on Build GRCh38
rs774332893 Chr.20: 45093496-45093496 on Build GRCh38
rs910243772 Chr.20: 45093565-45093565 on Build GRCh38
rs955558604 Chr.20: 45093869-45093869 on Build GRCh38
rs1038358609 Chr.20: 45093504-45093504 on Build GRCh38
rs79067117 Chr.20: 45093466-45093466 on Build GRCh38
rs898601746 Chr.20: 45093521-45093521 on Build GRCh38
rs6065785 Chr.20: 45093634-45093634 on Build GRCh38
rs972803337 Chr.20: 45093827-45093827 on Build GRCh38
rs532144035 Chr.20: 45093843-45093843 on Build GRCh38
rs919913431 Chr.20: 45093855-45093855 on Build GRCh38
rs943004099 Chr.20: 45093584-45093584 on Build GRCh38
rs934239214 Chr.20: 45093548-45093548 on Build GRCh38
rs554174105 Chr.20: 45093880-45093880 on Build GRCh38
rs1016859260 Chr.20: 45093783-45093783 on Build GRCh38
rs1031940209 Chr.20: 45093643-45093643 on Build GRCh38
rs1053558323 Chr.20: 45093614-45093614 on Build GRCh38
rs184678404 Chr.20: 45093618-45093618 on Build GRCh38
rs961447004 Chr.20: 45093784-45093784 on Build GRCh38
rs981607018 Chr.20: 45093694-45093694 on Build GRCh38
rs573394914 Chr.20: 45093854-45093854 on Build GRCh38
rs922760946 Chr.20: 45093536-45093536 on Build GRCh38
rs145773809 Chr.20: 45093654-45093654 on Build GRCh38
rs1033595354 Chr.20: 45093670-45093670 on Build GRCh38
rs971626097 Chr.20: 45093691-45093691 on Build GRCh38
rs1008569716 Chr.20: 45093577-45093577 on Build GRCh38
rs771172253 Chr.20: 45093477-45093477 on Build GRCh38
rs936214312 Chr.20: 45093908-45093908 on Build GRCh38
rs1030282400 Chr.20: 45093864-45093864 on Build GRCh38
rs927551875 Chr.20: 45093715-45093715 on Build GRCh38
rs958928792 Chr.20: 45093793-45093793 on Build GRCh38
rs981657838 Chr.20: 45093459-45093459 on Build GRCh38
rs188302375 Chr.20: 45093664-45093664 on Build GRCh38
rs945078988 Chr.20: 45093622-45093622 on Build GRCh38
rs759588629 Chr.20: 45093529-45093529 on Build GRCh38
rs1005525352 Chr.20: 45093681-45093681 on Build GRCh38
rs974081623 Chr.20: 45093794-45093794 on Build GRCh38
rs985727488 Chr.20: 45093564-45093564 on Build GRCh38
rs1020881345 Chr.20: 45093680-45093680 on Build GRCh38
rs761099480 Chr.20: 45093754-45093754 on Build GRCh38
rs895172787 Chr.20: 45093677-45093677 on Build GRCh38
rs911459405 Chr.20: 45093895-45093895 on Build GRCh38

View Full Product Details