Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45097938-45098441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNS1
Entrez Gene ID
3787
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Gene Aliases
Kv9.1
UniGene
Hs.117780
Gene Chromosome Location
Chr.20: 45091214-45101112 on Build GRCh38
Transcripts
NM_001322799.1 NM_002251.4 XM_017027846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211645 c.755C>T p.A252V Chr.20: 45098017-45098017 on Build GRCh38
3693524 c.586G>A p.A196T Chr.20: 45098186-45098186 on Build GRCh38
4632830 c.471G>A p.L157L Chr.20: 45098301-45098301 on Build GRCh38
4624258 c.384G>A p.E128E Chr.20: 45098388-45098388 on Build GRCh38
5811251 c.623T>A p.L208Q Chr.20: 45098149-45098149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764547218 Chr.20: 45098338-45098338 on Build GRCh38
rs754484302 Chr.20: 45098216-45098216 on Build GRCh38
rs781080055 Chr.20: 45098146-45098146 on Build GRCh38
rs968994291 Chr.20: 45098408-45098408 on Build GRCh38
rs755101813 Chr.20: 45098136-45098136 on Build GRCh38
rs776913037 Chr.20: 45098342-45098342 on Build GRCh38
rs971091515 Chr.20: 45098239-45098239 on Build GRCh38
rs774683642 Chr.20: 45098286-45098286 on Build GRCh38
rs752113669 Chr.20: 45097983-45097983 on Build GRCh38
rs985945503 Chr.20: 45098051-45098051 on Build GRCh38
rs761577755 Chr.20: 45097962-45097962 on Build GRCh38
rs768631916 Chr.20: 45098145-45098145 on Build GRCh38
rs562295055 Chr.20: 45098251-45098251 on Build GRCh38
rs1002984834 Chr.20: 45098288-45098288 on Build GRCh38
rs763020291 Chr.20: 45098163-45098163 on Build GRCh38
rs748006971 Chr.20: 45098148-45098148 on Build GRCh38
rs374752149 Chr.20: 45098206-45098206 on Build GRCh38
rs746974187 Chr.20: 45098144-45098144 on Build GRCh38
rs993471960 Chr.20: 45098315-45098315 on Build GRCh38
rs764306069 Chr.20: 45098194-45098194 on Build GRCh38
rs1045239162 Chr.20: 45098257-45098257 on Build GRCh38
rs776877543 Chr.20: 45098362-45098362 on Build GRCh38
rs746010157 Chr.20: 45098287-45098287 on Build GRCh38
rs541155099 Chr.20: 45098004-45098004 on Build GRCh38
rs147406486 Chr.20: 45098195-45098195 on Build GRCh38
rs781164466 Chr.20: 45098219-45098219 on Build GRCh38
rs985105120 Chr.20: 45098333-45098333 on Build GRCh38
rs751203756 Chr.20: 45098376-45098376 on Build GRCh38
rs761018535 Chr.20: 45098324-45098324 on Build GRCh38
rs773497188 Chr.20: 45098158-45098158 on Build GRCh38
rs766314841 Chr.20: 45097978-45097978 on Build GRCh38
rs772262134 Chr.20: 45098304-45098304 on Build GRCh38
rs902036716 Chr.20: 45098107-45098107 on Build GRCh38
rs761174654 Chr.20: 45098191-45098191 on Build GRCh38
rs1006383236 Chr.20: 45098261-45098261 on Build GRCh38
rs1040376123 Chr.20: 45098358-45098358 on Build GRCh38
rs751113930 Chr.20: 45098207-45098207 on Build GRCh38
rs777743004 Chr.20: 45098410-45098410 on Build GRCh38
rs752664327 Chr.20: 45098402-45098402 on Build GRCh38
rs1040845868 Chr.20: 45098095-45098095 on Build GRCh38
rs763151470 Chr.20: 45097976-45097976 on Build GRCh38
rs550109434 Chr.20: 45098179-45098179 on Build GRCh38
rs201608614 Chr.20: 45098118-45098118 on Build GRCh38
rs1017677972 Chr.20: 45098404-45098404 on Build GRCh38
rs778690478 Chr.20: 45098092-45098092 on Build GRCh38
rs954763965 Chr.20: 45098016-45098017 on Build GRCh38
rs948429725 Chr.20: 45098230-45098230 on Build GRCh38
rs979119191 Chr.20: 45098074-45098074 on Build GRCh38
rs776149926 Chr.20: 45098313-45098313 on Build GRCh38
rs756515952 Chr.20: 45098081-45098081 on Build GRCh38
rs766016752 Chr.20: 45098365-45098365 on Build GRCh38
rs748021274 Chr.20: 45098231-45098231 on Build GRCh38
rs924547755 Chr.20: 45098088-45098088 on Build GRCh38
rs552417867 Chr.20: 45098345-45098345 on Build GRCh38
rs370402014 Chr.20: 45098169-45098169 on Build GRCh38
rs532420183 Chr.20: 45098337-45098337 on Build GRCh38
rs905324525 Chr.20: 45098258-45098258 on Build GRCh38
rs755943859 Chr.20: 45098235-45098235 on Build GRCh38
rs754498637 Chr.20: 45098379-45098379 on Build GRCh38
rs368048059 Chr.20: 45098267-45098267 on Build GRCh38
rs968669932 Chr.20: 45098059-45098059 on Build GRCh38
rs371977105 Chr.20: 45098281-45098281 on Build GRCh38
rs766125887 Chr.20: 45098080-45098080 on Build GRCh38
rs749076353 Chr.20: 45098415-45098415 on Build GRCh38
rs762209311 Chr.20: 45098361-45098361 on Build GRCh38
rs940819587 Chr.20: 45098065-45098065 on Build GRCh38
rs910336517 Chr.20: 45098056-45098056 on Build GRCh38
rs369840753 Chr.20: 45098180-45098180 on Build GRCh38
rs750026831 Chr.20: 45098106-45098106 on Build GRCh38
rs760078538 Chr.20: 45097984-45097984 on Build GRCh38
rs779360976 Chr.20: 45098135-45098135 on Build GRCh38
rs770046040 Chr.20: 45097965-45097965 on Build GRCh38
rs755958406 Chr.20: 45098406-45098406 on Build GRCh38
rs768178246 Chr.20: 45098174-45098174 on Build GRCh38
rs536654737 Chr.20: 45098100-45098100 on Build GRCh38
rs774522244 Chr.20: 45098173-45098173 on Build GRCh38
rs747273011 Chr.20: 45098048-45098048 on Build GRCh38
rs201080873 Chr.20: 45098394-45098394 on Build GRCh38
rs749545248 Chr.20: 45098276-45098276 on Build GRCh38
rs370783761 Chr.20: 45097968-45097968 on Build GRCh38
rs757070162 Chr.20: 45098416-45098416 on Build GRCh38
rs1030401147 Chr.20: 45097977-45097977 on Build GRCh38
rs368484022 Chr.20: 45098150-45098150 on Build GRCh38
rs886435348 Chr.20: 45098057-45098057 on Build GRCh38
rs753161361 Chr.20: 45098078-45098078 on Build GRCh38

View Full Product Details