Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62202264-62202806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTG2
Entrez Gene ID
26164
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5, ObgH1, dJ1005F21.2
UniGene
Hs.340636
Gene Chromosome Location
Chr.20: 62183025-62203568 on Build GRCh38
Transcripts
NM_015666.3 XM_005260393.1 XM_006723774.3 XM_011528776.1 XM_017027805.1 XM_017027806.1 XM_017027807.1 XM_017027808.1

SNP Detail

SNP ID Chromosome Location
rs950694653 Chr.20: 62202692-62202692 on Build GRCh38
rs887598194 Chr.20: 62202769-62202769 on Build GRCh38
rs535305085 Chr.20: 62202291-62202291 on Build GRCh38
rs1054538337 Chr.20: 62202620-62202620 on Build GRCh38
rs950634879 Chr.20: 62202324-62202324 on Build GRCh38
rs200619495 Chr.20: 62202446-62202446 on Build GRCh38
rs911601382 Chr.20: 62202498-62202498 on Build GRCh38
rs991428795 Chr.20: 62202776-62202776 on Build GRCh38
rs535584485 Chr.20: 62202351-62202351 on Build GRCh38
rs199996996 Chr.20: 62202751-62202751 on Build GRCh38
rs1029341999 Chr.20: 62202473-62202473 on Build GRCh38
rs774747684 Chr.20: 62202753-62202765 on Build GRCh38
rs1012681457 Chr.20: 62202362-62202362 on Build GRCh38
rs903453639 Chr.20: 62202565-62202566 on Build GRCh38
rs150565894 Chr.20: 62202439-62202446 on Build GRCh38
rs367730632 Chr.20: 62202610-62202610 on Build GRCh38
rs971822008 Chr.20: 62202519-62202519 on Build GRCh38
rs993246049 Chr.20: 62202744-62202744 on Build GRCh38
rs1048618619 Chr.20: 62202587-62202587 on Build GRCh38
rs761898717 Chr.20: 62202443-62202446 on Build GRCh38
rs553607931 Chr.20: 62202340-62202340 on Build GRCh38
rs776707348 Chr.20: 62202301-62202301 on Build GRCh38
rs1025030639 Chr.20: 62202384-62202384 on Build GRCh38
rs11539718 Chr.20: 62202293-62202293 on Build GRCh38
rs773565375 Chr.20: 62202501-62202501 on Build GRCh38
rs561591304 Chr.20: 62202625-62202625 on Build GRCh38
rs563785817 Chr.20: 62202669-62202669 on Build GRCh38
rs575046398 Chr.20: 62202346-62202346 on Build GRCh38
rs1030839379 Chr.20: 62202408-62202408 on Build GRCh38
rs1048950914 Chr.20: 62202752-62202752 on Build GRCh38
rs571523663 Chr.20: 62202656-62202656 on Build GRCh38
rs1049151939 Chr.20: 62202337-62202337 on Build GRCh38
rs367544583 Chr.20: 62202458-62202461 on Build GRCh38
rs545800228 Chr.20: 62202529-62202529 on Build GRCh38
rs576655964 Chr.20: 62202735-62202736 on Build GRCh38
rs776940830 Chr.20: 62202364-62202364 on Build GRCh38
rs529052751 Chr.20: 62202626-62202626 on Build GRCh38
rs897368063 Chr.20: 62202300-62202300 on Build GRCh38
rs540099370 Chr.20: 62202624-62202624 on Build GRCh38
rs987549902 Chr.20: 62202495-62202495 on Build GRCh38
rs763495228 Chr.20: 62202566-62202566 on Build GRCh38
rs918742893 Chr.20: 62202528-62202528 on Build GRCh38
rs909846604 Chr.20: 62202405-62202405 on Build GRCh38
rs1046437529 Chr.20: 62202775-62202775 on Build GRCh38
rs372627222 Chr.20: 62202615-62202615 on Build GRCh38
rs1026679437 Chr.20: 62202287-62202287 on Build GRCh38
rs895130383 Chr.20: 62202339-62202339 on Build GRCh38
rs575166850 Chr.20: 62202441-62202441 on Build GRCh38
rs934891467 Chr.20: 62202569-62202569 on Build GRCh38
rs563899607 Chr.20: 62202548-62202548 on Build GRCh38
rs759974698 Chr.20: 62202480-62202480 on Build GRCh38
rs906626094 Chr.20: 62202394-62202394 on Build GRCh38
rs529898106 Chr.20: 62202471-62202471 on Build GRCh38
rs370925285 Chr.20: 62202454-62202461 on Build GRCh38
rs999321681 Chr.20: 62202407-62202407 on Build GRCh38
rs557014555 Chr.20: 62202422-62202422 on Build GRCh38
rs747329862 Chr.20: 62202439-62202442 on Build GRCh38
rs561092592 Chr.20: 62202465-62202466 on Build GRCh38
rs1026123069 Chr.20: 62202677-62202677 on Build GRCh38
rs543584895 Chr.20: 62202710-62202710 on Build GRCh38
rs111741946 Chr.20: 62202765-62202765 on Build GRCh38
rs1040156269 Chr.20: 62202401-62202401 on Build GRCh38
rs931497459 Chr.20: 62202576-62202576 on Build GRCh38
rs765825719 Chr.20: 62202613-62202613 on Build GRCh38
rs201844863 Chr.20: 62202693-62202693 on Build GRCh38
rs377274862 Chr.20: 62202685-62202685 on Build GRCh38
rs962722495 Chr.20: 62202748-62202748 on Build GRCh38

View Full Product Details