Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31873119-31873623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1

SNP Detail

SNP ID Chromosome Location
rs553262657 Chr.21: 31873172-31873172 on Build GRCh38
rs1038392663 Chr.21: 31873181-31873181 on Build GRCh38
rs980526714 Chr.21: 31873286-31873286 on Build GRCh38
rs542222932 Chr.21: 31873148-31873148 on Build GRCh38
rs966197985 Chr.21: 31873176-31873176 on Build GRCh38
rs926495044 Chr.21: 31873292-31873292 on Build GRCh38
rs1056315825 Chr.21: 31873299-31873299 on Build GRCh38
rs549844695 Chr.21: 31873381-31873381 on Build GRCh38
rs565178689 Chr.21: 31873387-31873387 on Build GRCh38
rs879628415 Chr.21: 31873431-31873431 on Build GRCh38
rs532527322 Chr.21: 31873393-31873393 on Build GRCh38
rs111796090 Chr.21: 31873380-31873380 on Build GRCh38
rs141312880 Chr.21: 31873477-31873478 on Build GRCh38
rs899328491 Chr.21: 31873197-31873197 on Build GRCh38
rs892343668 Chr.21: 31873328-31873328 on Build GRCh38
rs903839682 Chr.21: 31873385-31873385 on Build GRCh38
rs1017033085 Chr.21: 31873538-31873538 on Build GRCh38
rs764978599 Chr.21: 31873263-31873263 on Build GRCh38
rs1003272346 Chr.21: 31873309-31873309 on Build GRCh38
rs951032440 Chr.21: 31873258-31873258 on Build GRCh38
rs878892794 Chr.21: 31873511-31873511 on Build GRCh38
rs994037892 Chr.21: 31873206-31873206 on Build GRCh38
rs970427970 Chr.21: 31873270-31873270 on Build GRCh38
rs902021966 Chr.21: 31873440-31873440 on Build GRCh38
rs547687531 Chr.21: 31873401-31873401 on Build GRCh38
rs941138609 Chr.21: 31873278-31873278 on Build GRCh38
rs962400145 Chr.21: 31873560-31873564 on Build GRCh38
rs976367147 Chr.21: 31873187-31873187 on Build GRCh38
rs889142859 Chr.21: 31873156-31873156 on Build GRCh38
rs950917434 Chr.21: 31873497-31873497 on Build GRCh38
rs565999964 Chr.21: 31873463-31873463 on Build GRCh38
rs997646343 Chr.21: 31873466-31873466 on Build GRCh38
rs367676404 Chr.21: 31873547-31873547 on Build GRCh38
rs8126866 Chr.21: 31873493-31873493 on Build GRCh38
rs939178251 Chr.21: 31873297-31873298 on Build GRCh38
rs1008097068 Chr.21: 31873389-31873389 on Build GRCh38
rs1017694558 Chr.21: 31873414-31873414 on Build GRCh38
rs570029732 Chr.21: 31873557-31873557 on Build GRCh38
rs573204320 Chr.21: 31873213-31873213 on Build GRCh38
rs944112728 Chr.21: 31873274-31873287 on Build GRCh38
rs548321340 Chr.21: 31873519-31873519 on Build GRCh38

View Full Product Details