Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31873838-31874331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1 XM_011529537.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792647 c.261+2T>C p.? Chr.21: 31873937-31873937 on Build GRCh38
4101227 c.205G>T p.G69C Chr.21: 31873879-31873879 on Build GRCh38
6370070 c.165C>A p.H55Q Chr.21: 31873839-31873839 on Build GRCh38
6586611 c.251C>T p.T84I Chr.21: 31873925-31873925 on Build GRCh38
6035173 c.207C>A p.G69G Chr.21: 31873881-31873881 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906741570 Chr.21: 31874202-31874202 on Build GRCh38
rs867403784 Chr.21: 31874290-31874290 on Build GRCh38
rs541263368 Chr.21: 31874267-31874267 on Build GRCh38
rs1047842785 Chr.21: 31874157-31874157 on Build GRCh38
rs1006353966 Chr.21: 31874169-31874169 on Build GRCh38
rs756958824 Chr.21: 31873946-31873946 on Build GRCh38
rs554121934 Chr.21: 31873890-31873890 on Build GRCh38
rs763179584 Chr.21: 31874159-31874159 on Build GRCh38
rs763338038 Chr.21: 31873870-31873870 on Build GRCh38
rs752627020 Chr.21: 31873912-31873912 on Build GRCh38
rs764676766 Chr.21: 31874165-31874165 on Build GRCh38
rs1020668855 Chr.21: 31874295-31874295 on Build GRCh38
rs1047868285 Chr.21: 31874183-31874183 on Build GRCh38
rs943404249 Chr.21: 31874148-31874148 on Build GRCh38
rs773136184 Chr.21: 31873869-31873869 on Build GRCh38
rs559500337 Chr.21: 31874279-31874279 on Build GRCh38
rs922446303 Chr.21: 31874098-31874098 on Build GRCh38
rs753282638 Chr.21: 31873927-31873927 on Build GRCh38
rs759807242 Chr.21: 31874139-31874139 on Build GRCh38
rs532587813 Chr.21: 31874238-31874238 on Build GRCh38
rs755289079 Chr.21: 31873961-31873961 on Build GRCh38
rs1010952770 Chr.21: 31874235-31874235 on Build GRCh38
rs979765830 Chr.21: 31874120-31874120 on Build GRCh38
rs1051876863 Chr.21: 31874114-31874114 on Build GRCh38
rs898768964 Chr.21: 31874011-31874011 on Build GRCh38
rs763727972 Chr.21: 31874057-31874057 on Build GRCh38
rs991856632 Chr.21: 31873993-31873993 on Build GRCh38
rs766698607 Chr.21: 31873888-31873888 on Build GRCh38
rs925185623 Chr.21: 31874129-31874129 on Build GRCh38
rs564589746 Chr.21: 31874232-31874232 on Build GRCh38
rs542883295 Chr.21: 31873988-31873988 on Build GRCh38
rs977888561 Chr.21: 31874068-31874068 on Build GRCh38
rs781388640 Chr.21: 31873981-31873981 on Build GRCh38
rs967004079 Chr.21: 31874258-31874258 on Build GRCh38
rs752047611 Chr.21: 31874172-31874172 on Build GRCh38
rs759335599 Chr.21: 31873906-31873906 on Build GRCh38
rs542075947 Chr.21: 31873974-31873974 on Build GRCh38
rs71838131 Chr.21: 31874114-31874115 on Build GRCh38
rs147058521 Chr.21: 31873911-31873911 on Build GRCh38
rs966949999 Chr.21: 31874305-31874305 on Build GRCh38
rs778478686 Chr.21: 31873950-31873950 on Build GRCh38
rs906090189 Chr.21: 31874184-31874184 on Build GRCh38
rs989412409 Chr.21: 31874007-31874007 on Build GRCh38
rs1036962889 Chr.21: 31873881-31873881 on Build GRCh38
rs763698903 Chr.21: 31873921-31873921 on Build GRCh38
rs755979165 Chr.21: 31873914-31873914 on Build GRCh38
rs933372013 Chr.21: 31874104-31874104 on Build GRCh38
rs71790770 Chr.21: 31874221-31874222 on Build GRCh38
rs943480634 Chr.21: 31874294-31874294 on Build GRCh38
rs955224832 Chr.21: 31874289-31874289 on Build GRCh38
rs1021971794 Chr.21: 31874246-31874246 on Build GRCh38
rs894899911 Chr.21: 31874234-31874234 on Build GRCh38
rs933417838 Chr.21: 31874153-31874153 on Build GRCh38
rs1033334438 Chr.21: 31874250-31874250 on Build GRCh38
rs867754768 Chr.21: 31874291-31874291 on Build GRCh38
rs7277651 Chr.21: 31874142-31874142 on Build GRCh38
rs966658336 Chr.21: 31874052-31874052 on Build GRCh38
rs769966135 Chr.21: 31873868-31873868 on Build GRCh38
rs748201528 Chr.21: 31873867-31873867 on Build GRCh38
rs187318167 Chr.21: 31874009-31874009 on Build GRCh38
rs943384550 Chr.21: 31874136-31874136 on Build GRCh38
rs748730122 Chr.21: 31873982-31873982 on Build GRCh38
rs868716928 Chr.21: 31874192-31874192 on Build GRCh38
rs959334385 Chr.21: 31873985-31873985 on Build GRCh38
rs398101602 Chr.21: 31874116-31874117 on Build GRCh38
rs1033407886 Chr.21: 31873960-31873960 on Build GRCh38
rs1003241349 Chr.21: 31874215-31874215 on Build GRCh38
rs539433470 Chr.21: 31873887-31873887 on Build GRCh38
rs1037716359 Chr.21: 31874162-31874162 on Build GRCh38
rs376686896 Chr.21: 31874182-31874182 on Build GRCh38
rs747407336 Chr.21: 31873956-31873956 on Build GRCh38
rs1038359950 Chr.21: 31874180-31874180 on Build GRCh38
rs775954677 Chr.21: 31874147-31874147 on Build GRCh38
rs966562293 Chr.21: 31874037-31874037 on Build GRCh38
rs113788583 Chr.21: 31874116-31874116 on Build GRCh38

View Full Product Details