Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31974471-31974968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1 XM_011529537.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3758844 c.1080C>T p.S360S Chr.21: 31974624-31974624 on Build GRCh38
1254532 c.1091A>C p.N364T Chr.21: 31974635-31974635 on Build GRCh38
1666268 c.1111G>C p.A371P Chr.21: 31974655-31974655 on Build GRCh38
4001998 c.1106A>C p.N369T Chr.21: 31974650-31974650 on Build GRCh38
6683153 c.1090A>G p.N364D Chr.21: 31974634-31974634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750503400 Chr.21: 31974695-31974695 on Build GRCh38
rs764964333 Chr.21: 31974522-31974522 on Build GRCh38
rs376886034 Chr.21: 31974737-31974737 on Build GRCh38
rs998440758 Chr.21: 31974531-31974531 on Build GRCh38
rs530955254 Chr.21: 31974901-31974901 on Build GRCh38
rs55976679 Chr.21: 31974624-31974624 on Build GRCh38
rs201678062 Chr.21: 31974511-31974511 on Build GRCh38
rs886932795 Chr.21: 31974889-31974889 on Build GRCh38
rs769185515 Chr.21: 31974761-31974761 on Build GRCh38
rs934786572 Chr.21: 31974516-31974516 on Build GRCh38
rs747820113 Chr.21: 31974798-31974798 on Build GRCh38
rs780565997 Chr.21: 31974704-31974704 on Build GRCh38
rs759245800 Chr.21: 31974742-31974742 on Build GRCh38
rs372591679 Chr.21: 31974550-31974550 on Build GRCh38
rs201353197 Chr.21: 31974495-31974495 on Build GRCh38
rs776501076 Chr.21: 31974586-31974586 on Build GRCh38
rs765301879 Chr.21: 31974673-31974673 on Build GRCh38
rs751377501 Chr.21: 31974725-31974725 on Build GRCh38
rs775998273 Chr.21: 31974639-31974639 on Build GRCh38
rs779772512 Chr.21: 31974579-31974579 on Build GRCh38
rs770525630 Chr.21: 31974741-31974741 on Build GRCh38
rs1001173055 Chr.21: 31974841-31974841 on Build GRCh38
rs1039863127 Chr.21: 31974518-31974518 on Build GRCh38
rs377240004 Chr.21: 31974510-31974510 on Build GRCh38
rs746748107 Chr.21: 31974582-31974582 on Build GRCh38
rs761084683 Chr.21: 31974650-31974650 on Build GRCh38
rs559644700 Chr.21: 31974797-31974797 on Build GRCh38
rs758562782 Chr.21: 31974723-31974723 on Build GRCh38
rs76055360 Chr.21: 31974888-31974888 on Build GRCh38
rs911941716 Chr.21: 31974931-31974931 on Build GRCh38
rs369480730 Chr.21: 31974515-31974515 on Build GRCh38
rs956743727 Chr.21: 31974865-31974865 on Build GRCh38
rs748054042 Chr.21: 31974620-31974620 on Build GRCh38
rs199996026 Chr.21: 31974600-31974600 on Build GRCh38
rs780932641 Chr.21: 31974733-31974733 on Build GRCh38
rs142782450 Chr.21: 31974864-31974864 on Build GRCh38
rs1041426300 Chr.21: 31974500-31974500 on Build GRCh38
rs768611459 Chr.21: 31974585-31974585 on Build GRCh38
rs745694854 Chr.21: 31974509-31974509 on Build GRCh38
rs115793516 Chr.21: 31974809-31974809 on Build GRCh38
rs767833801 Chr.21: 31974539-31974539 on Build GRCh38
rs939820820 Chr.21: 31974914-31974914 on Build GRCh38
rs201828596 Chr.21: 31974578-31974578 on Build GRCh38
rs764724200 Chr.21: 31974654-31974654 on Build GRCh38
rs753094206 Chr.21: 31974540-31974540 on Build GRCh38
rs896158217 Chr.21: 31974751-31974751 on Build GRCh38
rs62221640 Chr.21: 31974653-31974653 on Build GRCh38
rs778491908 Chr.21: 31974739-31974739 on Build GRCh38
rs765995957 Chr.21: 31974628-31974628 on Build GRCh38
rs777158399 Chr.21: 31974738-31974738 on Build GRCh38
rs754448972 Chr.21: 31974655-31974655 on Build GRCh38
rs772787408 Chr.21: 31974527-31974527 on Build GRCh38
rs550043648 Chr.21: 31974775-31974775 on Build GRCh38
rs748099220 Chr.21: 31974734-31974734 on Build GRCh38
rs960792228 Chr.21: 31974714-31974714 on Build GRCh38
rs183149457 Chr.21: 31974521-31974521 on Build GRCh38
rs754005752 Chr.21: 31974551-31974551 on Build GRCh38
rs756154326 Chr.21: 31974492-31974492 on Build GRCh38
rs762567981 Chr.21: 31974627-31974627 on Build GRCh38
rs780209655 Chr.21: 31974724-31974724 on Build GRCh38
rs779031599 Chr.21: 31974563-31974563 on Build GRCh38
rs754815879 Chr.21: 31974726-31974726 on Build GRCh38
rs762632455 Chr.21: 31974533-31974533 on Build GRCh38
rs756698126 Chr.21: 31974544-31974544 on Build GRCh38
rs1011887139 Chr.21: 31974791-31974791 on Build GRCh38

View Full Product Details