Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33296165-33296670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL10RB
Entrez Gene ID
3588
Gene Name
interleukin 10 receptor subunit beta
Gene Aliases
CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2
UniGene
Hs.654593
Gene Chromosome Location
Chr.21: 33266360-33297234 on Build GRCh38
Transcripts
NM_000628.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4627881 c.922A>G p.N308D Chr.21: 33296301-33296301 on Build GRCh38
6867418 c.864G>A p.E288E Chr.21: 33296243-33296243 on Build GRCh38
6881142 c.871G>A p.V291I Chr.21: 33296250-33296250 on Build GRCh38
2843571 c.954G>A p.P318P Chr.21: 33296333-33296333 on Build GRCh38
1030274 c.857C>T p.S286L Chr.21: 33296236-33296236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374161818 Chr.21: 33296333-33296333 on Build GRCh38
rs886057004 Chr.21: 33296377-33296377 on Build GRCh38
rs776039316 Chr.21: 33296339-33296340 on Build GRCh38
rs540281964 Chr.21: 33296627-33296627 on Build GRCh38
rs370841672 Chr.21: 33296325-33296325 on Build GRCh38
rs764945131 Chr.21: 33296284-33296285 on Build GRCh38
rs746167504 Chr.21: 33296327-33296327 on Build GRCh38
rs376104508 Chr.21: 33296395-33296395 on Build GRCh38
rs146668107 Chr.21: 33296201-33296201 on Build GRCh38
rs781038420 Chr.21: 33296448-33296448 on Build GRCh38
rs780187887 Chr.21: 33296345-33296345 on Build GRCh38
rs757379186 Chr.21: 33296324-33296324 on Build GRCh38
rs778285637 Chr.21: 33296213-33296213 on Build GRCh38
rs376143910 Chr.21: 33296292-33296292 on Build GRCh38
rs776455147 Chr.21: 33296368-33296368 on Build GRCh38
rs755414697 Chr.21: 33296198-33296198 on Build GRCh38
rs753324475 Chr.21: 33296203-33296203 on Build GRCh38
rs147772699 Chr.21: 33296265-33296265 on Build GRCh38
rs141414815 Chr.21: 33296332-33296332 on Build GRCh38
rs762495022 Chr.21: 33296384-33296384 on Build GRCh38
rs772836914 Chr.21: 33296382-33296382 on Build GRCh38
rs776563865 Chr.21: 33296263-33296263 on Build GRCh38
rs763395229 Chr.21: 33296342-33296345 on Build GRCh38
rs8178561 Chr.21: 33296492-33296492 on Build GRCh38
rs544407415 Chr.21: 33296310-33296310 on Build GRCh38
rs752012391 Chr.21: 33296445-33296446 on Build GRCh38
rs770320812 Chr.21: 33296387-33296387 on Build GRCh38
rs561856540 Chr.21: 33296638-33296638 on Build GRCh38
rs1037746312 Chr.21: 33296422-33296422 on Build GRCh38
rs771890796 Chr.21: 33296236-33296236 on Build GRCh38
rs773041746 Chr.21: 33296286-33296286 on Build GRCh38
rs758406164 Chr.21: 33296423-33296423 on Build GRCh38
rs373486817 Chr.21: 33296270-33296270 on Build GRCh38
rs886057003 Chr.21: 33296374-33296374 on Build GRCh38
rs200681978 Chr.21: 33296217-33296217 on Build GRCh38
rs922350498 Chr.21: 33296339-33296339 on Build GRCh38
rs771006570 Chr.21: 33296224-33296224 on Build GRCh38
rs531666376 Chr.21: 33296225-33296225 on Build GRCh38
rs1019924295 Chr.21: 33296188-33296188 on Build GRCh38
rs998948453 Chr.21: 33296595-33296595 on Build GRCh38
rs750510923 Chr.21: 33296421-33296421 on Build GRCh38
rs764500130 Chr.21: 33296399-33296399 on Build GRCh38
rs11701888 Chr.21: 33296457-33296457 on Build GRCh38
rs3171425 Chr.21: 33296442-33296442 on Build GRCh38
rs528482502 Chr.21: 33296610-33296610 on Build GRCh38
rs555064260 Chr.21: 33296480-33296480 on Build GRCh38
rs748094856 Chr.21: 33296452-33296452 on Build GRCh38
rs753016855 Chr.21: 33296307-33296307 on Build GRCh38
rs780170183 Chr.21: 33296431-33296431 on Build GRCh38
rs764543444 Chr.21: 33296308-33296308 on Build GRCh38
rs762692366 Chr.21: 33296290-33296290 on Build GRCh38
rs778886949 Chr.21: 33296285-33296286 on Build GRCh38
rs768491723 Chr.21: 33296243-33296243 on Build GRCh38
rs756566292 Chr.21: 33296209-33296209 on Build GRCh38
rs895990590 Chr.21: 33296628-33296628 on Build GRCh38
rs760719286 Chr.21: 33296237-33296237 on Build GRCh38
rs139413536 Chr.21: 33296411-33296411 on Build GRCh38
rs770520463 Chr.21: 33296501-33296501 on Build GRCh38
rs769497995 Chr.21: 33296380-33296380 on Build GRCh38
rs757755568 Chr.21: 33296418-33296418 on Build GRCh38
rs879575028 Chr.21: 33296626-33296626 on Build GRCh38
rs764763018 Chr.21: 33296360-33296360 on Build GRCh38
rs911014085 Chr.21: 33296268-33296268 on Build GRCh38
rs768456830 Chr.21: 33296367-33296367 on Build GRCh38
rs760973106 Chr.21: 33296396-33296396 on Build GRCh38
rs781553501 Chr.21: 33296212-33296212 on Build GRCh38
rs1033176483 Chr.21: 33296641-33296641 on Build GRCh38
rs367791594 Chr.21: 33296636-33296636 on Build GRCh38
rs773217539 Chr.21: 33296481-33296481 on Build GRCh38
rs1058862 Chr.21: 33296360-33296360 on Build GRCh38
rs765354572 Chr.21: 33296420-33296420 on Build GRCh38
rs942530927 Chr.21: 33296291-33296291 on Build GRCh38
rs1014684753 Chr.21: 33296486-33296486 on Build GRCh38
rs747203732 Chr.21: 33296434-33296434 on Build GRCh38
rs746785015 Chr.21: 33296347-33296347 on Build GRCh38
rs773927781 Chr.21: 33296526-33296526 on Build GRCh38
rs746070714 Chr.21: 33296231-33296231 on Build GRCh38
rs748094483 Chr.21: 33296376-33296376 on Build GRCh38
rs754344615 Chr.21: 33296416-33296416 on Build GRCh38

View Full Product Details