Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33403204-33403699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNGR2
Entrez Gene ID
3460
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Gene Aliases
AF-1, IFGR2, IFNGT1, IMD28
UniGene
Hs.634632
Gene Chromosome Location
Chr.21: 33402896-33437521 on Build GRCh38
Transcripts
NM_005534.3 XM_005260969.2

SNP Detail

SNP ID Chromosome Location
rs759238264 Chr.21: 33403611-33403611 on Build GRCh38
rs759786879 Chr.21: 33403567-33403569 on Build GRCh38
rs776426451 Chr.21: 33403526-33403526 on Build GRCh38
rs995335196 Chr.21: 33403488-33403488 on Build GRCh38
rs964009116 Chr.21: 33403364-33403364 on Build GRCh38
rs912251110 Chr.21: 33403677-33403677 on Build GRCh38
rs1049363076 Chr.21: 33403442-33403442 on Build GRCh38
rs943716579 Chr.21: 33403678-33403678 on Build GRCh38
rs982552344 Chr.21: 33403436-33403436 on Build GRCh38
rs769756592 Chr.21: 33403538-33403538 on Build GRCh38
rs990250212 Chr.21: 33403626-33403626 on Build GRCh38
rs539352863 Chr.21: 33403422-33403422 on Build GRCh38
rs890879682 Chr.21: 33403445-33403445 on Build GRCh38
rs945735993 Chr.21: 33403224-33403224 on Build GRCh38
rs904460473 Chr.21: 33403252-33403252 on Build GRCh38
rs975085315 Chr.21: 33403370-33403370 on Build GRCh38
rs1026903027 Chr.21: 33403347-33403347 on Build GRCh38
rs922318778 Chr.21: 33403380-33403380 on Build GRCh38
rs1022250905 Chr.21: 33403358-33403358 on Build GRCh38
rs952051116 Chr.21: 33403365-33403365 on Build GRCh38
rs1026930622 Chr.21: 33403513-33403513 on Build GRCh38
rs747873434 Chr.21: 33403532-33403532 on Build GRCh38
rs897021799 Chr.21: 33403454-33403454 on Build GRCh38
rs751362234 Chr.21: 33403630-33403630 on Build GRCh38
rs914813287 Chr.21: 33403433-33403433 on Build GRCh38
rs1036776220 Chr.21: 33403453-33403453 on Build GRCh38
rs1008360191 Chr.21: 33403449-33403449 on Build GRCh38
rs767210895 Chr.21: 33403624-33403624 on Build GRCh38
rs553694549 Chr.21: 33403523-33403523 on Build GRCh38
rs1054673896 Chr.21: 33403429-33403429 on Build GRCh38
rs995865383 Chr.21: 33403332-33403332 on Build GRCh38
rs908692954 Chr.21: 33403469-33403469 on Build GRCh38
rs985101045 Chr.21: 33403648-33403648 on Build GRCh38
rs934837539 Chr.21: 33403423-33403423 on Build GRCh38
rs981966284 Chr.21: 33403591-33403602 on Build GRCh38
rs961328642 Chr.21: 33403307-33403307 on Build GRCh38
rs186418679 Chr.21: 33403643-33403643 on Build GRCh38
rs773932279 Chr.21: 33403596-33403596 on Build GRCh38
rs932332379 Chr.21: 33403440-33403440 on Build GRCh38
rs762420616 Chr.21: 33403552-33403552 on Build GRCh38
rs879598186 Chr.21: 33403386-33403386 on Build GRCh38
rs17882748 Chr.21: 33403415-33403415 on Build GRCh38
rs956427420 Chr.21: 33403599-33403599 on Build GRCh38
rs981635023 Chr.21: 33403400-33403400 on Build GRCh38
rs1012938610 Chr.21: 33403580-33403580 on Build GRCh38
rs1015805059 Chr.21: 33403292-33403292 on Build GRCh38
rs765468464 Chr.21: 33403591-33403593 on Build GRCh38
rs971513246 Chr.21: 33403590-33403591 on Build GRCh38
rs747254797 Chr.21: 33403512-33403512 on Build GRCh38
rs753866192 Chr.21: 33403566-33403567 on Build GRCh38
rs1041631611 Chr.21: 33403225-33403225 on Build GRCh38
rs971862261 Chr.21: 33403390-33403390 on Build GRCh38
rs556894037 Chr.21: 33403273-33403273 on Build GRCh38
rs571968917 Chr.21: 33403632-33403632 on Build GRCh38
rs773084508 Chr.21: 33403551-33403551 on Build GRCh38
rs765907908 Chr.21: 33403582-33403582 on Build GRCh38

View Full Product Details