Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39788068-39788583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF5
Entrez Gene ID
150084
Gene Name
immunoglobulin superfamily member 5
Gene Aliases
GSF5, JAM4
UniGene
Hs.422120
Gene Chromosome Location
Chr.21: 39725838-39802096 on Build GRCh38
Transcripts
NM_001080444.1 XM_011529472.1 XM_011529473.1 XM_017028279.1 XM_017028280.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3728106 c.935-2A>T p.? Chr.21: 39788165-39788165 on Build GRCh38
1030781 c.948A>C p.Q316H Chr.21: 39788180-39788180 on Build GRCh38
6874042 c.935-1G>A p.? Chr.21: 39788166-39788166 on Build GRCh38
4513506 c.940C>T p.R314C Chr.21: 39788172-39788172 on Build GRCh38
4922458 c.956+1G>A p.? Chr.21: 39788189-39788189 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765489479 Chr.21: 39788181-39788181 on Build GRCh38
rs533383487 Chr.21: 39788415-39788415 on Build GRCh38
rs916252997 Chr.21: 39788380-39788380 on Build GRCh38
rs979213034 Chr.21: 39788388-39788388 on Build GRCh38
rs376931297 Chr.21: 39788163-39788163 on Build GRCh38
rs1014238941 Chr.21: 39788113-39788113 on Build GRCh38
rs1052940967 Chr.21: 39788454-39788454 on Build GRCh38
rs145234708 Chr.21: 39788173-39788173 on Build GRCh38
rs766791278 Chr.21: 39788156-39788157 on Build GRCh38
rs754024637 Chr.21: 39788169-39788173 on Build GRCh38
rs889867582 Chr.21: 39788490-39788491 on Build GRCh38
rs931914950 Chr.21: 39788377-39788377 on Build GRCh38
rs554711350 Chr.21: 39788275-39788275 on Build GRCh38
rs1014702548 Chr.21: 39788341-39788341 on Build GRCh38
rs141389311 Chr.21: 39788314-39788314 on Build GRCh38
rs896608360 Chr.21: 39788470-39788470 on Build GRCh38
rs200738861 Chr.21: 39788132-39788132 on Build GRCh38
rs456818 Chr.21: 39788315-39788315 on Build GRCh38
rs9983347 Chr.21: 39788207-39788207 on Build GRCh38
rs139012406 Chr.21: 39788172-39788172 on Build GRCh38
rs759831123 Chr.21: 39788196-39788197 on Build GRCh38
rs371740172 Chr.21: 39788390-39788390 on Build GRCh38
rs752833487 Chr.21: 39788217-39788221 on Build GRCh38
rs149173446 Chr.21: 39788190-39788190 on Build GRCh38
rs775844513 Chr.21: 39788164-39788164 on Build GRCh38
rs904115654 Chr.21: 39788504-39788504 on Build GRCh38
rs370522613 Chr.21: 39788209-39788209 on Build GRCh38
rs752638386 Chr.21: 39788264-39788264 on Build GRCh38
rs1023892164 Chr.21: 39788279-39788279 on Build GRCh38
rs757074435 Chr.21: 39788227-39788227 on Build GRCh38
rs920303401 Chr.21: 39788145-39788145 on Build GRCh38
rs371421293 Chr.21: 39788259-39788259 on Build GRCh38
rs117474157 Chr.21: 39788360-39788360 on Build GRCh38
rs940606818 Chr.21: 39788452-39788452 on Build GRCh38
rs568239337 Chr.21: 39788525-39788525 on Build GRCh38
rs960967465 Chr.21: 39788111-39788111 on Build GRCh38
rs192464932 Chr.21: 39788192-39788192 on Build GRCh38
rs747779504 Chr.21: 39788133-39788133 on Build GRCh38
rs150001048 Chr.21: 39788156-39788156 on Build GRCh38
rs1004416662 Chr.21: 39788527-39788527 on Build GRCh38
rs769239364 Chr.21: 39788543-39788543 on Build GRCh38
rs1027579135 Chr.21: 39788336-39788336 on Build GRCh38
rs541520482 Chr.21: 39788301-39788301 on Build GRCh38
rs905414766 Chr.21: 39788501-39788501 on Build GRCh38
rs934066442 Chr.21: 39788468-39788468 on Build GRCh38
rs115802366 Chr.21: 39788263-39788263 on Build GRCh38
rs960869767 Chr.21: 39788547-39788547 on Build GRCh38
rs190442016 Chr.21: 39788404-39788404 on Build GRCh38
rs1023921859 Chr.21: 39788128-39788128 on Build GRCh38
rs765178987 Chr.21: 39788206-39788212 on Build GRCh38
rs887685495 Chr.21: 39788188-39788188 on Build GRCh38
rs769390462 Chr.21: 39788134-39788134 on Build GRCh38
rs530773428 Chr.21: 39788552-39788552 on Build GRCh38
rs147012493 Chr.21: 39788339-39788339 on Build GRCh38
rs1007157034 Chr.21: 39788248-39788248 on Build GRCh38
rs372177091 Chr.21: 39788235-39788235 on Build GRCh38
rs867876064 Chr.21: 39788167-39788167 on Build GRCh38
rs1045099234 Chr.21: 39788480-39788480 on Build GRCh38
rs547994524 Chr.21: 39788524-39788524 on Build GRCh38
rs200920678 Chr.21: 39788100-39788100 on Build GRCh38
rs951805568 Chr.21: 39788338-39788338 on Build GRCh38
rs781270955 Chr.21: 39788226-39788227 on Build GRCh38
rs373076256 Chr.21: 39788203-39788203 on Build GRCh38
rs112976253 Chr.21: 39788511-39788511 on Build GRCh38
rs984417450 Chr.21: 39788411-39788411 on Build GRCh38
rs772806933 Chr.21: 39788143-39788143 on Build GRCh38
rs921885362 Chr.21: 39788193-39788193 on Build GRCh38
rs184331966 Chr.21: 39788346-39788346 on Build GRCh38
rs901655967 Chr.21: 39788456-39788456 on Build GRCh38
rs1006936366 Chr.21: 39788497-39788497 on Build GRCh38
rs949326696 Chr.21: 39788476-39788476 on Build GRCh38
rs777230172 Chr.21: 39788240-39788240 on Build GRCh38
rs780106547 Chr.21: 39788202-39788202 on Build GRCh38
rs879845516 Chr.21: 39788509-39788509 on Build GRCh38
rs147888602 Chr.21: 39788269-39788269 on Build GRCh38
rs762300300 Chr.21: 39788155-39788155 on Build GRCh38

View Full Product Details