Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92168759-92169259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1107
Entrez Gene ID
23285
Gene Name
KIAA1107
Gene Aliases
-
UniGene
Hs.21554
Gene Chromosome Location
Chr.1: 92167052-92188509 on Build GRCh38
Transcripts
XM_017000800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7241846 c.225G>A p.L75L Chr.1: 92168961-92168961 on Build GRCh38
1344893 c.202C>T p.R68C Chr.1: 92168938-92168938 on Build GRCh38
1344892 c.157C>T p.R53C Chr.1: 92168893-92168893 on Build GRCh38
6835204 c.259+1G>C p.? Chr.1: 92168996-92168996 on Build GRCh38
6503058 c.246G>A p.Q82Q Chr.1: 92168982-92168982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988707676 Chr.1: 92169190-92169190 on Build GRCh38
rs1052209965 Chr.1: 92168992-92168992 on Build GRCh38
rs891406398 Chr.1: 92169027-92169027 on Build GRCh38
rs767790954 Chr.1: 92169030-92169030 on Build GRCh38
rs1044777569 Chr.1: 92169219-92169219 on Build GRCh38
rs774884224 Chr.1: 92168893-92168893 on Build GRCh38
rs201795862 Chr.1: 92168939-92168939 on Build GRCh38
rs926515516 Chr.1: 92169232-92169232 on Build GRCh38
rs912912882 Chr.1: 92168911-92168911 on Build GRCh38
rs944283122 Chr.1: 92169054-92169054 on Build GRCh38
rs980420873 Chr.1: 92168820-92168820 on Build GRCh38
rs142362153 Chr.1: 92169200-92169201 on Build GRCh38
rs949040550 Chr.1: 92169000-92169000 on Build GRCh38
rs533527827 Chr.1: 92169063-92169063 on Build GRCh38
rs995555521 Chr.1: 92169076-92169076 on Build GRCh38
rs147026255 Chr.1: 92169159-92169159 on Build GRCh38
rs959958597 Chr.1: 92168870-92168870 on Build GRCh38
rs770603621 Chr.1: 92168904-92168904 on Build GRCh38
rs769437604 Chr.1: 92168825-92168825 on Build GRCh38
rs947115269 Chr.1: 92168914-92168914 on Build GRCh38
rs1050412912 Chr.1: 92169026-92169026 on Build GRCh38
rs772988513 Chr.1: 92168878-92168878 on Build GRCh38
rs760879960 Chr.1: 92169114-92169114 on Build GRCh38
rs969861796 Chr.1: 92169200-92169200 on Build GRCh38
rs754141530 Chr.1: 92169201-92169201 on Build GRCh38
rs905152510 Chr.1: 92169034-92169034 on Build GRCh38
rs917710763 Chr.1: 92169209-92169209 on Build GRCh38
rs1036005355 Chr.1: 92169156-92169156 on Build GRCh38
rs925037105 Chr.1: 92168953-92168953 on Build GRCh38
rs184869066 Chr.1: 92169069-92169069 on Build GRCh38
rs560163981 Chr.1: 92169132-92169132 on Build GRCh38
rs536431300 Chr.1: 92169199-92169200 on Build GRCh38
rs74568907 Chr.1: 92169222-92169222 on Build GRCh38
rs573985324 Chr.1: 92168867-92168867 on Build GRCh38
rs760615713 Chr.1: 92168894-92168894 on Build GRCh38
rs548980717 Chr.1: 92169171-92169171 on Build GRCh38
rs1019696707 Chr.1: 92169104-92169104 on Build GRCh38
rs995323213 Chr.1: 92169041-92169041 on Build GRCh38
rs561205473 Chr.1: 92169228-92169228 on Build GRCh38
rs886958313 Chr.1: 92169071-92169071 on Build GRCh38
rs925434554 Chr.1: 92168972-92168972 on Build GRCh38
rs796575290 Chr.1: 92169144-92169144 on Build GRCh38
rs544838062 Chr.1: 92168947-92168947 on Build GRCh38
rs750609133 Chr.1: 92169044-92169044 on Build GRCh38
rs978485404 Chr.1: 92168933-92168933 on Build GRCh38
rs949032268 Chr.1: 92169210-92169210 on Build GRCh38
rs771792192 Chr.1: 92168837-92168837 on Build GRCh38
rs761863809 Chr.1: 92169007-92169007 on Build GRCh38
rs1019869416 Chr.1: 92169208-92169208 on Build GRCh38
rs555674305 Chr.1: 92168803-92168803 on Build GRCh38
rs935047389 Chr.1: 92168968-92168968 on Build GRCh38
rs980016054 Chr.1: 92169225-92169225 on Build GRCh38
rs764269086 Chr.1: 92168998-92168998 on Build GRCh38
rs141564225 Chr.1: 92168999-92168999 on Build GRCh38
rs566377338 Chr.1: 92168988-92168988 on Build GRCh38

View Full Product Details