Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92180139-92180660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1107
Entrez Gene ID
23285
Gene Name
KIAA1107
Gene Aliases
-
UniGene
Hs.21554
Gene Chromosome Location
Chr.1: 92167052-92188509 on Build GRCh38
Transcripts
XM_017000800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6540624 c.1607G>T p.R536L Chr.1: 92180604-92180604 on Build GRCh38
6883543 c.1442G>A p.G481E Chr.1: 92180274-92180274 on Build GRCh38
1602788 c.1606C>T p.R536C Chr.1: 92180603-92180603 on Build GRCh38
161928 c.1279G>A p.E427K Chr.1: 92180276-92180276 on Build GRCh38
5760732 c.1521G>T p.Q507H Chr.1: 92180518-92180518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559780200 Chr.1: 92180479-92180479 on Build GRCh38
rs375022441 Chr.1: 92180494-92180494 on Build GRCh38
rs762339923 Chr.1: 92180336-92180336 on Build GRCh38
rs973485604 Chr.1: 92180196-92180196 on Build GRCh38
rs200439049 Chr.1: 92180519-92180519 on Build GRCh38
rs753458005 Chr.1: 92180435-92180435 on Build GRCh38
rs969286143 Chr.1: 92180255-92180255 on Build GRCh38
rs371842534 Chr.1: 92180607-92180607 on Build GRCh38
rs752915828 Chr.1: 92180602-92180602 on Build GRCh38
rs113321393 Chr.1: 92180291-92180291 on Build GRCh38
rs760963436 Chr.1: 92180247-92180247 on Build GRCh38
rs61741973 Chr.1: 92180595-92180595 on Build GRCh38
rs764350466 Chr.1: 92180612-92180614 on Build GRCh38
rs950839005 Chr.1: 92180293-92180293 on Build GRCh38
rs766924871 Chr.1: 92180319-92180319 on Build GRCh38
rs766087232 Chr.1: 92180421-92180421 on Build GRCh38
rs17131606 Chr.1: 92180377-92180377 on Build GRCh38
rs778723037 Chr.1: 92180469-92180469 on Build GRCh38
rs570186992 Chr.1: 92180609-92180609 on Build GRCh38
rs1023405140 Chr.1: 92180233-92180233 on Build GRCh38
rs1483022 Chr.1: 92180464-92180464 on Build GRCh38
rs895471108 Chr.1: 92180528-92180528 on Build GRCh38
rs769117720 Chr.1: 92180234-92180234 on Build GRCh38
rs866597241 Chr.1: 92180568-92180568 on Build GRCh38
rs1012561949 Chr.1: 92180537-92180537 on Build GRCh38
rs7552286 Chr.1: 92180396-92180396 on Build GRCh38
rs780469162 Chr.1: 92180527-92180527 on Build GRCh38
rs199735511 Chr.1: 92180511-92180511 on Build GRCh38
rs982427400 Chr.1: 92180273-92180273 on Build GRCh38
rs184101262 Chr.1: 92180368-92180368 on Build GRCh38
rs1052365994 Chr.1: 92180552-92180552 on Build GRCh38
rs760165097 Chr.1: 92180399-92180399 on Build GRCh38
rs994878468 Chr.1: 92180623-92180623 on Build GRCh38
rs767845526 Chr.1: 92180471-92180471 on Build GRCh38
rs751702442 Chr.1: 92180395-92180395 on Build GRCh38
rs769171218 Chr.1: 92180604-92180604 on Build GRCh38
rs935265995 Chr.1: 92180375-92180375 on Build GRCh38
rs890689640 Chr.1: 92180563-92180563 on Build GRCh38
rs987577052 Chr.1: 92180349-92180349 on Build GRCh38
rs576456064 Chr.1: 92180242-92180242 on Build GRCh38
rs915524654 Chr.1: 92180442-92180442 on Build GRCh38

View Full Product Details