Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14488856-14489383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
UniGene
Hs.517819
Gene Chromosome Location
Chr.3: 14489111-14602996 on Build GRCh38
Transcripts
NM_001080423.3 NM_001134367.3 NM_003043.5 NR_103507.2 XM_006713307.2 XM_011534030.2 XM_011534031.2

Gene Symbol
SLC6A6
Entrez Gene ID
6533
Gene Name
solute carrier family 6 member 6
Gene Aliases
TAUT
UniGene
Hs.529488
Gene Chromosome Location
Chr.3: 14402576-14489349 on Build GRCh38
Transcripts
NM_001080423.3 NM_001134367.3 NM_003043.5 NR_103507.2 XM_006713307.2 XM_011534030.2 XM_011534031.2

SNP Detail

SNP ID Chromosome Location
rs958132263 Chr.3: 14489245-14489245 on Build GRCh38
rs940432325 Chr.3: 14489224-14489225 on Build GRCh38
rs557371192 Chr.3: 14489080-14489081 on Build GRCh38
rs560894501 Chr.3: 14489121-14489121 on Build GRCh38
rs1032940484 Chr.3: 14489046-14489046 on Build GRCh38
rs755512611 Chr.3: 14488931-14488931 on Build GRCh38
rs941537625 Chr.3: 14489353-14489353 on Build GRCh38
rs9036 Chr.3: 14489213-14489213 on Build GRCh38
rs559672512 Chr.3: 14489279-14489279 on Build GRCh38
rs936746842 Chr.3: 14488885-14488885 on Build GRCh38
rs539851892 Chr.3: 14489224-14489224 on Build GRCh38
rs1030164064 Chr.3: 14489205-14489205 on Build GRCh38
rs996250522 Chr.3: 14489191-14489191 on Build GRCh38
rs1045292698 Chr.3: 14489231-14489231 on Build GRCh38
rs1047368310 Chr.3: 14489343-14489343 on Build GRCh38
rs931691847 Chr.3: 14488880-14488880 on Build GRCh38
rs758581924 Chr.3: 14489297-14489297 on Build GRCh38
rs771334430 Chr.3: 14489344-14489344 on Build GRCh38
rs989552722 Chr.3: 14489247-14489247 on Build GRCh38
rs1046610241 Chr.3: 14489003-14489003 on Build GRCh38
rs1036212941 Chr.3: 14488972-14488972 on Build GRCh38
rs917637964 Chr.3: 14489225-14489225 on Build GRCh38
rs757145298 Chr.3: 14488952-14488952 on Build GRCh38
rs553373527 Chr.3: 14489318-14489318 on Build GRCh38
rs916563686 Chr.3: 14488996-14488996 on Build GRCh38
rs779109671 Chr.3: 14489160-14489160 on Build GRCh38
rs893224888 Chr.3: 14489049-14489050 on Build GRCh38
rs969377622 Chr.3: 14489271-14489271 on Build GRCh38
rs372896141 Chr.3: 14489166-14489166 on Build GRCh38
rs1005951510 Chr.3: 14489081-14489081 on Build GRCh38
rs984465962 Chr.3: 14489123-14489123 on Build GRCh38
rs1013822234 Chr.3: 14489007-14489007 on Build GRCh38
rs77607340 Chr.3: 14489035-14489035 on Build GRCh38
rs1020423834 Chr.3: 14489081-14489081 on Build GRCh38
rs770022263 Chr.3: 14488911-14488912 on Build GRCh38
rs910028135 Chr.3: 14489352-14489352 on Build GRCh38
rs916662510 Chr.3: 14489258-14489258 on Build GRCh38
rs1045011737 Chr.3: 14489013-14489013 on Build GRCh38
rs147207210 Chr.3: 14489042-14489042 on Build GRCh38
rs140308593 Chr.3: 14489050-14489050 on Build GRCh38
rs989997277 Chr.3: 14488987-14488987 on Build GRCh38
rs539105060 Chr.3: 14489000-14489000 on Build GRCh38
rs1047253980 Chr.3: 14489073-14489073 on Build GRCh38
rs1037873488 Chr.3: 14489090-14489090 on Build GRCh38
rs1048757574 Chr.3: 14488912-14488912 on Build GRCh38
rs982519166 Chr.3: 14489294-14489294 on Build GRCh38
rs1030775856 Chr.3: 14489226-14489226 on Build GRCh38
rs1029287545 Chr.3: 14489092-14489095 on Build GRCh38
rs908998778 Chr.3: 14488937-14488937 on Build GRCh38
rs535010390 Chr.3: 14489311-14489311 on Build GRCh38
rs866505142 Chr.3: 14489101-14489101 on Build GRCh38
rs888645145 Chr.3: 14489074-14489074 on Build GRCh38

View Full Product Details