Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19253515-19254013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH8
Entrez Gene ID
131096
Gene Name
potassium voltage-gated channel subfamily H member 8
Gene Aliases
ELK, ELK1, Kv12.1, elk3
UniGene
Hs.475656
Gene Chromosome Location
Chr.3: 19148454-19535643 on Build GRCh38
Transcripts
NM_144633.2 XM_017005698.1 XM_017005699.1 XM_017005700.1 XM_017005701.1 XM_017005702.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799486 c.272C>A p.T91K Chr.3: 19253849-19253849 on Build GRCh38
4457318 c.103C>T p.Q35* Chr.3: 19253680-19253680 on Build GRCh38
729948 c.267G>A p.E89E Chr.3: 19253844-19253844 on Build GRCh38
1484964 c.172A>G p.T58A Chr.3: 19253749-19253749 on Build GRCh38
6097329 c.110C>T p.A37V Chr.3: 19253687-19253687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556934057 Chr.3: 19253651-19253651 on Build GRCh38
rs776045327 Chr.3: 19253636-19253638 on Build GRCh38
rs375011458 Chr.3: 19253933-19253933 on Build GRCh38
rs370167832 Chr.3: 19253859-19253859 on Build GRCh38
rs375524089 Chr.3: 19253649-19253649 on Build GRCh38
rs959425070 Chr.3: 19253598-19253598 on Build GRCh38
rs761421443 Chr.3: 19253743-19253743 on Build GRCh38
rs773011184 Chr.3: 19253858-19253858 on Build GRCh38
rs74640554 Chr.3: 19253905-19253905 on Build GRCh38
rs1000834982 Chr.3: 19253570-19253570 on Build GRCh38
rs760167301 Chr.3: 19253719-19253719 on Build GRCh38
rs775955215 Chr.3: 19253833-19253833 on Build GRCh38
rs758839261 Chr.3: 19253759-19253759 on Build GRCh38
rs764731093 Chr.3: 19253857-19253857 on Build GRCh38
rs564126399 Chr.3: 19253746-19253746 on Build GRCh38
rs768191040 Chr.3: 19253831-19253831 on Build GRCh38
rs762735278 Chr.3: 19253922-19253922 on Build GRCh38
rs148905975 Chr.3: 19253665-19253665 on Build GRCh38
rs748376306 Chr.3: 19253794-19253794 on Build GRCh38
rs373622554 Chr.3: 19253804-19253804 on Build GRCh38
rs768438674 Chr.3: 19253901-19253901 on Build GRCh38
rs770724667 Chr.3: 19253678-19253678 on Build GRCh38
rs368456035 Chr.3: 19253921-19253921 on Build GRCh38
rs766933038 Chr.3: 19253764-19253764 on Build GRCh38
rs747018462 Chr.3: 19253644-19253644 on Build GRCh38
rs887578385 Chr.3: 19253765-19253765 on Build GRCh38
rs967190300 Chr.3: 19253562-19253562 on Build GRCh38
rs541398243 Chr.3: 19253578-19253578 on Build GRCh38
rs373918603 Chr.3: 19253890-19253890 on Build GRCh38
rs766916681 Chr.3: 19253868-19253868 on Build GRCh38
rs763395090 Chr.3: 19253864-19253864 on Build GRCh38
rs979666911 Chr.3: 19253573-19253573 on Build GRCh38
rs919051762 Chr.3: 19253667-19253667 on Build GRCh38
rs192627517 Chr.3: 19253906-19253906 on Build GRCh38
rs774193937 Chr.3: 19253795-19253795 on Build GRCh38
rs202115249 Chr.3: 19253693-19253693 on Build GRCh38
rs368719351 Chr.3: 19253823-19253823 on Build GRCh38
rs758162637 Chr.3: 19253887-19253887 on Build GRCh38
rs199638242 Chr.3: 19253869-19253869 on Build GRCh38
rs925406601 Chr.3: 19253596-19253596 on Build GRCh38
rs533263551 Chr.3: 19253980-19253980 on Build GRCh38
rs912850946 Chr.3: 19253935-19253935 on Build GRCh38
rs565744148 Chr.3: 19253875-19253875 on Build GRCh38
rs750921698 Chr.3: 19253631-19253631 on Build GRCh38
rs780738930 Chr.3: 19253632-19253632 on Build GRCh38
rs111725285 Chr.3: 19253650-19253650 on Build GRCh38
rs969043260 Chr.3: 19253547-19253547 on Build GRCh38
rs753496962 Chr.3: 19253618-19253618 on Build GRCh38
rs865920585 Chr.3: 19253841-19253841 on Build GRCh38
rs373010498 Chr.3: 19253914-19253914 on Build GRCh38
rs961604431 Chr.3: 19253738-19253738 on Build GRCh38
rs529173044 Chr.3: 19253701-19253701 on Build GRCh38
rs772150706 Chr.3: 19253614-19253614 on Build GRCh38
rs768086928 Chr.3: 19253873-19253873 on Build GRCh38
rs369906313 Chr.3: 19253616-19253616 on Build GRCh38
rs781461081 Chr.3: 19253655-19253655 on Build GRCh38
rs1021373193 Chr.3: 19253865-19253865 on Build GRCh38
rs771954038 Chr.3: 19253711-19253711 on Build GRCh38
rs1001325367 Chr.3: 19253896-19253896 on Build GRCh38
rs746992706 Chr.3: 19253824-19253824 on Build GRCh38
rs11923699 Chr.3: 19253775-19253775 on Build GRCh38
rs1040435877 Chr.3: 19253663-19253663 on Build GRCh38
rs745683327 Chr.3: 19253708-19253708 on Build GRCh38
rs377586865 Chr.3: 19253818-19253818 on Build GRCh38
rs965615790 Chr.3: 19253966-19253966 on Build GRCh38
rs776320230 Chr.3: 19253734-19253734 on Build GRCh38
rs373290866 Chr.3: 19253886-19253886 on Build GRCh38
rs778085313 Chr.3: 19253807-19253807 on Build GRCh38
rs750779500 Chr.3: 19253756-19253756 on Build GRCh38
rs779187063 Chr.3: 19253624-19253624 on Build GRCh38
rs545887787 Chr.3: 19253728-19253728 on Build GRCh38
rs761420458 Chr.3: 19253847-19253847 on Build GRCh38
rs756220179 Chr.3: 19253878-19253878 on Build GRCh38
rs757639009 Chr.3: 19253619-19253619 on Build GRCh38
rs752168041 Chr.3: 19253773-19253773 on Build GRCh38
rs1013575993 Chr.3: 19253826-19253826 on Build GRCh38
rs781169194 Chr.3: 19253789-19253789 on Build GRCh38
rs746829538 Chr.3: 19253900-19253900 on Build GRCh38
rs755483864 Chr.3: 19253871-19253871 on Build GRCh38
rs117407062 Chr.3: 19253932-19253932 on Build GRCh38
rs369890839 Chr.3: 19253715-19253715 on Build GRCh38

View Full Product Details