Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19394816-19395318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH8
Entrez Gene ID
131096
Gene Name
potassium voltage-gated channel subfamily H member 8
Gene Aliases
ELK, ELK1, Kv12.1, elk3
UniGene
Hs.475656
Gene Chromosome Location
Chr.3: 19148454-19535643 on Build GRCh38
Transcripts
NM_144633.2 XM_011533347.2 XM_017005698.1 XM_017005699.1 XM_017005700.1 XM_017005701.1 XM_017005702.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6482019 c.1051C>A p.H351N Chr.3: 19395185-19395185 on Build GRCh38
3334229 c.1010G>A p.R337H Chr.3: 19395144-19395144 on Build GRCh38
245150 c.1053C>T p.H351H Chr.3: 19395187-19395187 on Build GRCh38
1566503 c.1113G>A p.A371A Chr.3: 19395247-19395247 on Build GRCh38
729876 c.1177+1G>A p.? Chr.3: 19395312-19395312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754907543 Chr.3: 19395247-19395247 on Build GRCh38
rs755740067 Chr.3: 19395275-19395275 on Build GRCh38
rs768032265 Chr.3: 19395113-19395113 on Build GRCh38
rs537658979 Chr.3: 19395174-19395174 on Build GRCh38
rs1050736470 Chr.3: 19394861-19394861 on Build GRCh38
rs771177562 Chr.3: 19395286-19395286 on Build GRCh38
rs774727569 Chr.3: 19395291-19395291 on Build GRCh38
rs745912732 Chr.3: 19395272-19395273 on Build GRCh38
rs10470642 Chr.3: 19395095-19395095 on Build GRCh38
rs973485055 Chr.3: 19395118-19395118 on Build GRCh38
rs756278505 Chr.3: 19395056-19395056 on Build GRCh38
rs1024137285 Chr.3: 19394876-19394876 on Build GRCh38
rs1040641388 Chr.3: 19395047-19395047 on Build GRCh38
rs773009170 Chr.3: 19395144-19395144 on Build GRCh38
rs747601313 Chr.3: 19395271-19395271 on Build GRCh38
rs890701904 Chr.3: 19394884-19394884 on Build GRCh38
rs575576619 Chr.3: 19395193-19395193 on Build GRCh38
rs749009930 Chr.3: 19395280-19395280 on Build GRCh38
rs777977711 Chr.3: 19395070-19395070 on Build GRCh38
rs529197468 Chr.3: 19394920-19394920 on Build GRCh38
rs892103258 Chr.3: 19394910-19394910 on Build GRCh38
rs138407554 Chr.3: 19395086-19395087 on Build GRCh38
rs746247633 Chr.3: 19395292-19395292 on Build GRCh38
rs373564560 Chr.3: 19395149-19395149 on Build GRCh38
rs1052039879 Chr.3: 19394908-19394908 on Build GRCh38
rs17005915 Chr.3: 19394942-19394942 on Build GRCh38
rs200575141 Chr.3: 19395196-19395196 on Build GRCh38
rs993035863 Chr.3: 19394856-19394856 on Build GRCh38
rs536078878 Chr.3: 19395042-19395042 on Build GRCh38
rs977486225 Chr.3: 19394890-19394890 on Build GRCh38
rs944987357 Chr.3: 19394993-19394993 on Build GRCh38
rs755123795 Chr.3: 19394946-19394946 on Build GRCh38
rs11721265 Chr.3: 19395206-19395206 on Build GRCh38
rs1034982313 Chr.3: 19395040-19395040 on Build GRCh38
rs775851110 Chr.3: 19395284-19395285 on Build GRCh38
rs910758312 Chr.3: 19394991-19394991 on Build GRCh38
rs11721268 Chr.3: 19395229-19395229 on Build GRCh38
rs1002438259 Chr.3: 19394986-19394986 on Build GRCh38
rs376188228 Chr.3: 19395153-19395153 on Build GRCh38
rs760137238 Chr.3: 19395165-19395165 on Build GRCh38
rs747694669 Chr.3: 19395126-19395126 on Build GRCh38
rs769502038 Chr.3: 19395134-19395134 on Build GRCh38
rs767931626 Chr.3: 19395173-19395173 on Build GRCh38
rs960888343 Chr.3: 19395046-19395046 on Build GRCh38
rs781283040 Chr.3: 19395259-19395259 on Build GRCh38
rs958651526 Chr.3: 19394849-19394849 on Build GRCh38
rs758272449 Chr.3: 19395087-19395087 on Build GRCh38
rs779087039 Chr.3: 19395143-19395143 on Build GRCh38
rs145114975 Chr.3: 19395181-19395181 on Build GRCh38
rs772479127 Chr.3: 19395277-19395277 on Build GRCh38
rs574912623 Chr.3: 19395009-19395009 on Build GRCh38
rs751337521 Chr.3: 19395246-19395246 on Build GRCh38
rs369829387 Chr.3: 19395121-19395121 on Build GRCh38
rs766250526 Chr.3: 19395241-19395241 on Build GRCh38
rs986168629 Chr.3: 19394966-19394966 on Build GRCh38
rs777559557 Chr.3: 19395279-19395279 on Build GRCh38
rs566420938 Chr.3: 19394894-19394894 on Build GRCh38
rs370083901 Chr.3: 19395159-19395159 on Build GRCh38
rs750161969 Chr.3: 19395084-19395084 on Build GRCh38
rs747011626 Chr.3: 19395112-19395112 on Build GRCh38

View Full Product Details