Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20072251-20072762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT2B
Entrez Gene ID
8850
Gene Name
lysine acetyltransferase 2B
Gene Aliases
CAF, P/CAF, PCAF
UniGene
Hs.533055
Gene Chromosome Location
Chr.3: 20040032-20154404 on Build GRCh38
Transcripts
NM_003884.4 XM_005265528.4 XM_017007423.1 XM_017007424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4418905 c.309G>T p.E103D Chr.3: 20072338-20072338 on Build GRCh38
3767384 c.429A>C p.L143L Chr.3: 20072458-20072458 on Build GRCh38
5864238 c.358delC p.R122fs*7 Chr.3: 20072387-20072387 on Build GRCh38
3823573 c.410G>A p.R137Q Chr.3: 20072439-20072439 on Build GRCh38
6690152 c.389T>C p.V130A Chr.3: 20072418-20072418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565153650 Chr.3: 20072647-20072647 on Build GRCh38
rs780301580 Chr.3: 20072319-20072319 on Build GRCh38
rs201855469 Chr.3: 20072302-20072302 on Build GRCh38
rs971050133 Chr.3: 20072727-20072727 on Build GRCh38
rs962623632 Chr.3: 20072369-20072369 on Build GRCh38
rs769969403 Chr.3: 20072320-20072320 on Build GRCh38
rs1016738426 Chr.3: 20072702-20072702 on Build GRCh38
rs761510735 Chr.3: 20072373-20072373 on Build GRCh38
rs749538432 Chr.3: 20072399-20072399 on Build GRCh38
rs372460930 Chr.3: 20072489-20072489 on Build GRCh38
rs763665386 Chr.3: 20072492-20072493 on Build GRCh38
rs764920595 Chr.3: 20072374-20072374 on Build GRCh38
rs756102146 Chr.3: 20072491-20072491 on Build GRCh38
rs1053926412 Chr.3: 20072602-20072602 on Build GRCh38
rs201922668 Chr.3: 20072398-20072398 on Build GRCh38
rs1054058665 Chr.3: 20072290-20072290 on Build GRCh38
rs752088443 Chr.3: 20072378-20072378 on Build GRCh38
rs565923626 Chr.3: 20072723-20072723 on Build GRCh38
rs750101129 Chr.3: 20072375-20072375 on Build GRCh38
rs542928956 Chr.3: 20072650-20072650 on Build GRCh38
rs768530791 Chr.3: 20072410-20072410 on Build GRCh38
rs199589780 Chr.3: 20072363-20072363 on Build GRCh38
rs747550075 Chr.3: 20072286-20072286 on Build GRCh38
rs779867237 Chr.3: 20072402-20072402 on Build GRCh38
rs200560854 Chr.3: 20072391-20072391 on Build GRCh38
rs375101044 Chr.3: 20072721-20072721 on Build GRCh38
rs894033666 Chr.3: 20072335-20072335 on Build GRCh38
rs982700206 Chr.3: 20072308-20072308 on Build GRCh38
rs758794416 Chr.3: 20072376-20072376 on Build GRCh38
rs201056968 Chr.3: 20072309-20072309 on Build GRCh38
rs766871792 Chr.3: 20072377-20072377 on Build GRCh38
rs770651239 Chr.3: 20072511-20072511 on Build GRCh38
rs767377212 Chr.3: 20072390-20072390 on Build GRCh38
rs748438929 Chr.3: 20072518-20072518 on Build GRCh38
rs983887757 Chr.3: 20072397-20072397 on Build GRCh38
rs772950002 Chr.3: 20072446-20072446 on Build GRCh38
rs3021408 Chr.3: 20072338-20072338 on Build GRCh38
rs894067762 Chr.3: 20072622-20072622 on Build GRCh38
rs781543067 Chr.3: 20072506-20072506 on Build GRCh38
rs951306553 Chr.3: 20072395-20072395 on Build GRCh38
rs746964628 Chr.3: 20072405-20072405 on Build GRCh38
rs748137421 Chr.3: 20072504-20072504 on Build GRCh38
rs780640772 Chr.3: 20072284-20072284 on Build GRCh38
rs751927546 Chr.3: 20072477-20072477 on Build GRCh38
rs41285055 Chr.3: 20072287-20072287 on Build GRCh38
rs767845859 Chr.3: 20072482-20072482 on Build GRCh38
rs981064863 Chr.3: 20072728-20072728 on Build GRCh38
rs376264367 Chr.3: 20072575-20072577 on Build GRCh38
rs760581446 Chr.3: 20072341-20072341 on Build GRCh38
rs771514707 Chr.3: 20072652-20072652 on Build GRCh38
rs769459234 Chr.3: 20072426-20072426 on Build GRCh38
rs749819325 Chr.3: 20072592-20072592 on Build GRCh38
rs962966827 Chr.3: 20072712-20072712 on Build GRCh38
rs3828349 Chr.3: 20072552-20072552 on Build GRCh38
rs3828348 Chr.3: 20072556-20072556 on Build GRCh38
rs756292917 Chr.3: 20072498-20072498 on Build GRCh38
rs776067737 Chr.3: 20072368-20072368 on Build GRCh38
rs755477559 Chr.3: 20072386-20072386 on Build GRCh38
rs544983010 Chr.3: 20072495-20072495 on Build GRCh38
rs754030761 Chr.3: 20072492-20072492 on Build GRCh38
rs771888629 Chr.3: 20072336-20072336 on Build GRCh38
rs563142891 Chr.3: 20072719-20072719 on Build GRCh38
rs966118993 Chr.3: 20072277-20072277 on Build GRCh38
rs144078165 Chr.3: 20072624-20072624 on Build GRCh38
rs547780759 Chr.3: 20072699-20072699 on Build GRCh38
rs759961036 Chr.3: 20072479-20072479 on Build GRCh38
rs755614826 Chr.3: 20072360-20072361 on Build GRCh38
rs1003519777 Chr.3: 20072676-20072676 on Build GRCh38
rs148170754 Chr.3: 20072505-20072505 on Build GRCh38
rs997537370 Chr.3: 20072280-20072280 on Build GRCh38
rs776641465 Chr.3: 20072420-20072420 on Build GRCh38
rs893393894 Chr.3: 20072532-20072532 on Build GRCh38
rs184573877 Chr.3: 20072634-20072634 on Build GRCh38
rs555906565 Chr.3: 20072392-20072392 on Build GRCh38
rs970751902 Chr.3: 20072382-20072382 on Build GRCh38
rs773436380 Chr.3: 20072324-20072324 on Build GRCh38
rs906549059 Chr.3: 20072638-20072638 on Build GRCh38
rs745595746 Chr.3: 20072325-20072325 on Build GRCh38
rs3749180 Chr.3: 20072497-20072497 on Build GRCh38
rs143856943 Chr.3: 20072470-20072470 on Build GRCh38
rs113213072 Chr.3: 20072680-20072680 on Build GRCh38
rs1026159961 Chr.3: 20072677-20072677 on Build GRCh38

View Full Product Details