Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20114772-20115282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT2B
Entrez Gene ID
8850
Gene Name
lysine acetyltransferase 2B
Gene Aliases
CAF, P/CAF, PCAF
UniGene
Hs.533055
Gene Chromosome Location
Chr.3: 20040032-20154404 on Build GRCh38
Transcripts
NM_003884.4 XM_005265528.4 XM_017007423.1 XM_017007424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6577686 c.1044-4C>T p.? Chr.3: 20114878-20114878 on Build GRCh38
4939641 c.1150+2T>G p.? Chr.3: 20114990-20114990 on Build GRCh38
3823574 c.1083C>G p.N361K Chr.3: 20114921-20114921 on Build GRCh38
6423265 c.1049T>A p.L350Q Chr.3: 20114887-20114887 on Build GRCh38
6690153 c.1103A>G p.D368G Chr.3: 20114941-20114941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902049226 Chr.3: 20114854-20114858 on Build GRCh38
rs190875179 Chr.3: 20115137-20115137 on Build GRCh38
rs768015791 Chr.3: 20114964-20114964 on Build GRCh38
rs978041007 Chr.3: 20114936-20114936 on Build GRCh38
rs984989673 Chr.3: 20114797-20114797 on Build GRCh38
rs997604094 Chr.3: 20114860-20114860 on Build GRCh38
rs748292958 Chr.3: 20114843-20114844 on Build GRCh38
rs916437288 Chr.3: 20115044-20115044 on Build GRCh38
rs914840420 Chr.3: 20114821-20114821 on Build GRCh38
rs925406968 Chr.3: 20114967-20114967 on Build GRCh38
rs775645714 Chr.3: 20114801-20114801 on Build GRCh38
rs4515067 Chr.3: 20114856-20114856 on Build GRCh38
rs199916721 Chr.3: 20114934-20114934 on Build GRCh38
rs552844014 Chr.3: 20115252-20115252 on Build GRCh38
rs776344007 Chr.3: 20114991-20114991 on Build GRCh38
rs747786674 Chr.3: 20115006-20115006 on Build GRCh38
rs754962896 Chr.3: 20114862-20114862 on Build GRCh38
rs886424705 Chr.3: 20114920-20114920 on Build GRCh38
rs769848771 Chr.3: 20114844-20114844 on Build GRCh38
rs769510529 Chr.3: 20115034-20115034 on Build GRCh38
rs772277545 Chr.3: 20114803-20114803 on Build GRCh38
rs563995164 Chr.3: 20115208-20115208 on Build GRCh38
rs745597798 Chr.3: 20114914-20114914 on Build GRCh38
rs765688747 Chr.3: 20114843-20114843 on Build GRCh38
rs1015745349 Chr.3: 20114915-20114915 on Build GRCh38
rs1012823077 Chr.3: 20115056-20115056 on Build GRCh38
rs779811352 Chr.3: 20114930-20114930 on Build GRCh38
rs965254842 Chr.3: 20115253-20115253 on Build GRCh38
rs960181343 Chr.3: 20115254-20115254 on Build GRCh38
rs761593639 Chr.3: 20114832-20114832 on Build GRCh38
rs934343344 Chr.3: 20114807-20114807 on Build GRCh38
rs924286402 Chr.3: 20114804-20114804 on Build GRCh38
rs993850602 Chr.3: 20115144-20115144 on Build GRCh38
rs576967787 Chr.3: 20115049-20115049 on Build GRCh38
rs1055195451 Chr.3: 20115040-20115040 on Build GRCh38
rs777985994 Chr.3: 20114888-20114888 on Build GRCh38
rs752913448 Chr.3: 20114869-20114869 on Build GRCh38
rs548830211 Chr.3: 20115159-20115159 on Build GRCh38
rs936782253 Chr.3: 20114983-20114983 on Build GRCh38
rs976339882 Chr.3: 20114825-20114825 on Build GRCh38
rs147518139 Chr.3: 20115128-20115128 on Build GRCh38
rs1018016178 Chr.3: 20115243-20115243 on Build GRCh38
rs531081326 Chr.3: 20115239-20115239 on Build GRCh38
rs781373205 Chr.3: 20114863-20114863 on Build GRCh38
rs774564899 Chr.3: 20114908-20114911 on Build GRCh38
rs2365794 Chr.3: 20114810-20114810 on Build GRCh38
rs764403827 Chr.3: 20115231-20115231 on Build GRCh38
rs375169189 Chr.3: 20114925-20114925 on Build GRCh38
rs541066627 Chr.3: 20115051-20115051 on Build GRCh38
rs766728301 Chr.3: 20114854-20114854 on Build GRCh38
rs914788284 Chr.3: 20115177-20115177 on Build GRCh38
rs372053800 Chr.3: 20114998-20114998 on Build GRCh38
rs1022787595 Chr.3: 20115059-20115059 on Build GRCh38
rs200480938 Chr.3: 20114852-20114852 on Build GRCh38
rs752138277 Chr.3: 20114859-20114859 on Build GRCh38
rs767729527 Chr.3: 20115013-20115015 on Build GRCh38
rs888106267 Chr.3: 20115170-20115170 on Build GRCh38
rs559728786 Chr.3: 20115130-20115130 on Build GRCh38
rs750873507 Chr.3: 20114844-20114844 on Build GRCh38
rs1048307974 Chr.3: 20114865-20114865 on Build GRCh38
rs564999991 Chr.3: 20114876-20114876 on Build GRCh38

View Full Product Details