Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20125778-20126281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT2B
Entrez Gene ID
8850
Gene Name
lysine acetyltransferase 2B
Gene Aliases
CAF, P/CAF, PCAF
UniGene
Hs.533055
Gene Chromosome Location
Chr.3: 20040032-20154404 on Build GRCh38
Transcripts
NM_003884.4 XM_005265528.4 XM_017007423.1 XM_017007424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377431 c.1432C>A p.H478N Chr.3: 20125923-20125923 on Build GRCh38
7190490 c.1502A>G p.N501S Chr.3: 20125993-20125993 on Build GRCh38
1043293 c.1622+2T>C p.? Chr.3: 20126115-20126115 on Build GRCh38
7171223 c.1527G>C p.K509N Chr.3: 20126018-20126018 on Build GRCh38
1239183 c.1492G>A p.V498M Chr.3: 20125983-20125983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772124073 Chr.3: 20126038-20126038 on Build GRCh38
rs746113235 Chr.3: 20125898-20125898 on Build GRCh38
rs764591571 Chr.3: 20126134-20126134 on Build GRCh38
rs762920270 Chr.3: 20126095-20126095 on Build GRCh38
rs751236716 Chr.3: 20125866-20125866 on Build GRCh38
rs746086370 Chr.3: 20126036-20126036 on Build GRCh38
rs769554331 Chr.3: 20125820-20125820 on Build GRCh38
rs143548388 Chr.3: 20125928-20125928 on Build GRCh38
rs75200486 Chr.3: 20125861-20125861 on Build GRCh38
rs777398044 Chr.3: 20126079-20126079 on Build GRCh38
rs773908616 Chr.3: 20126098-20126098 on Build GRCh38
rs937321141 Chr.3: 20125964-20125964 on Build GRCh38
rs376017300 Chr.3: 20126133-20126133 on Build GRCh38
rs542043280 Chr.3: 20126216-20126216 on Build GRCh38
rs1007198862 Chr.3: 20126203-20126203 on Build GRCh38
rs367786973 Chr.3: 20126010-20126010 on Build GRCh38
rs756532890 Chr.3: 20125997-20125997 on Build GRCh38
rs866273086 Chr.3: 20125980-20125980 on Build GRCh38
rs369657764 Chr.3: 20125819-20125824 on Build GRCh38
rs1056293777 Chr.3: 20125983-20125983 on Build GRCh38
rs141445570 Chr.3: 20125996-20125996 on Build GRCh38
rs767911128 Chr.3: 20125960-20125960 on Build GRCh38
rs747861358 Chr.3: 20126069-20126069 on Build GRCh38
rs888796871 Chr.3: 20126187-20126187 on Build GRCh38
rs760923919 Chr.3: 20126122-20126122 on Build GRCh38
rs755350046 Chr.3: 20125870-20125870 on Build GRCh38
rs572889418 Chr.3: 20125805-20125805 on Build GRCh38
rs756637853 Chr.3: 20125886-20125886 on Build GRCh38
rs909139231 Chr.3: 20125839-20125839 on Build GRCh38
rs950116860 Chr.3: 20126013-20126013 on Build GRCh38
rs754389731 Chr.3: 20126141-20126141 on Build GRCh38
rs762403927 Chr.3: 20125927-20125927 on Build GRCh38
rs759100700 Chr.3: 20126103-20126103 on Build GRCh38
rs529058312 Chr.3: 20126201-20126201 on Build GRCh38
rs768489478 Chr.3: 20125910-20125910 on Build GRCh38
rs765021821 Chr.3: 20125924-20125924 on Build GRCh38
rs1048930176 Chr.3: 20126177-20126177 on Build GRCh38
rs773114824 Chr.3: 20126081-20126081 on Build GRCh38
rs949149161 Chr.3: 20125906-20125906 on Build GRCh38
rs778429289 Chr.3: 20125888-20125888 on Build GRCh38
rs769601797 Chr.3: 20126078-20126078 on Build GRCh38
rs373721059 Chr.3: 20125922-20125922 on Build GRCh38
rs754654855 Chr.3: 20125959-20125959 on Build GRCh38
rs936342669 Chr.3: 20125931-20125931 on Build GRCh38
rs757377789 Chr.3: 20126023-20126023 on Build GRCh38
rs757911346 Chr.3: 20125855-20125855 on Build GRCh38
rs1019014894 Chr.3: 20126205-20126205 on Build GRCh38
rs1026153211 Chr.3: 20126193-20126193 on Build GRCh38
rs748685129 Chr.3: 20125878-20125878 on Build GRCh38
rs770425092 Chr.3: 20126097-20126097 on Build GRCh38
rs752416354 Chr.3: 20126119-20126119 on Build GRCh38
rs952711818 Chr.3: 20126213-20126213 on Build GRCh38
rs925956674 Chr.3: 20125943-20125943 on Build GRCh38
rs1005550103 Chr.3: 20126253-20126253 on Build GRCh38
rs774691663 Chr.3: 20125894-20125894 on Build GRCh38
rs201537605 Chr.3: 20126034-20126034 on Build GRCh38
rs749339385 Chr.3: 20125889-20125889 on Build GRCh38
rs200471371 Chr.3: 20126113-20126113 on Build GRCh38
rs771156251 Chr.3: 20125893-20125893 on Build GRCh38
rs917763924 Chr.3: 20125905-20125905 on Build GRCh38
rs779198091 Chr.3: 20126027-20126027 on Build GRCh38
rs780331317 Chr.3: 20126057-20126057 on Build GRCh38
rs778374040 Chr.3: 20126003-20126003 on Build GRCh38
rs773167944 Chr.3: 20125925-20125925 on Build GRCh38

View Full Product Details