Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48694502-48694995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K2
Entrez Gene ID
51447
Gene Name
inositol hexakisphosphate kinase 2
Gene Aliases
IHPK2, PIUS
UniGene
Hs.595983
Gene Chromosome Location
Chr.3: 48688003-48717278 on Build GRCh38
Transcripts
NM_001005909.2 NM_001005910.2 NM_001005911.2 NM_001146178.2 NM_001146179.2 NM_001190316.1 NM_001190317.1 NM_016291.3 NR_027437.2 NR_027438.2 XM_006713199.3 XM_006713200.1 XM_006713201.1 XM_006713202.1 XM_011533816.1 XM_011533817.1 XM_011533818.1 XM_011533822.1 XM_011533823.1 XM_017006583.1 XM_017006584.1 XM_017006585.1 XM_017006586.1 XM_017006587.1 XM_017006588.1 XM_017006589.1 XM_017006590.1 XM_017006591.1 XM_017006592.1 XM_017006593.1

SNP Detail

SNP ID Chromosome Location
rs555966751 Chr.3: 48694662-48694662 on Build GRCh38
rs72929172 Chr.3: 48694528-48694528 on Build GRCh38
rs186030998 Chr.3: 48694704-48694704 on Build GRCh38
rs887416560 Chr.3: 48694951-48694951 on Build GRCh38
rs755701022 Chr.3: 48694957-48694957 on Build GRCh38
rs796901701 Chr.3: 48694875-48694877 on Build GRCh38
rs1035604543 Chr.3: 48694710-48694710 on Build GRCh38
rs760054255 Chr.3: 48694547-48694547 on Build GRCh38
rs766904497 Chr.3: 48694788-48694788 on Build GRCh38
rs574460998 Chr.3: 48694548-48694551 on Build GRCh38
rs573154172 Chr.3: 48694740-48694740 on Build GRCh38
rs563115137 Chr.3: 48694892-48694892 on Build GRCh38
rs772019329 Chr.3: 48694944-48694944 on Build GRCh38
rs1005944578 Chr.3: 48694641-48694641 on Build GRCh38
rs779815577 Chr.3: 48694956-48694956 on Build GRCh38
rs1005521204 Chr.3: 48694591-48694591 on Build GRCh38
rs879086979 Chr.3: 48694916-48694916 on Build GRCh38
rs897116518 Chr.3: 48694873-48694873 on Build GRCh38
rs560021874 Chr.3: 48694738-48694738 on Build GRCh38
rs988477657 Chr.3: 48694735-48694735 on Build GRCh38
rs1041284461 Chr.3: 48694759-48694759 on Build GRCh38
rs945640827 Chr.3: 48694749-48694749 on Build GRCh38
rs868699417 Chr.3: 48694624-48694624 on Build GRCh38
rs760932933 Chr.3: 48694958-48694960 on Build GRCh38
rs779631475 Chr.3: 48694917-48694918 on Build GRCh38
rs778063050 Chr.3: 48694843-48694843 on Build GRCh38
rs746532621 Chr.3: 48694963-48694963 on Build GRCh38
rs562184464 Chr.3: 48694803-48694803 on Build GRCh38
rs1037030132 Chr.3: 48694839-48694839 on Build GRCh38
rs969879349 Chr.3: 48694942-48694942 on Build GRCh38
rs774322013 Chr.3: 48694955-48694957 on Build GRCh38
rs757232672 Chr.3: 48694777-48694777 on Build GRCh38
rs74931524 Chr.3: 48694748-48694748 on Build GRCh38
rs1005987951 Chr.3: 48694822-48694822 on Build GRCh38
rs551179391 Chr.3: 48694891-48694891 on Build GRCh38
rs538015401 Chr.3: 48694656-48694656 on Build GRCh38
rs182441058 Chr.3: 48694655-48694655 on Build GRCh38
rs749294706 Chr.3: 48694782-48694783 on Build GRCh38
rs1028371902 Chr.3: 48694536-48694536 on Build GRCh38
rs763386239 Chr.3: 48694781-48694781 on Build GRCh38
rs776079531 Chr.3: 48694780-48694780 on Build GRCh38
rs200668427 Chr.3: 48694930-48694930 on Build GRCh38
rs767991254 Chr.3: 48694971-48694971 on Build GRCh38
rs961297677 Chr.3: 48694618-48694618 on Build GRCh38
rs979841371 Chr.3: 48694694-48694694 on Build GRCh38
rs756110385 Chr.3: 48694840-48694840 on Build GRCh38
rs778746899 Chr.3: 48694926-48694926 on Build GRCh38
rs757233119 Chr.3: 48694550-48694550 on Build GRCh38
rs887541445 Chr.3: 48694608-48694608 on Build GRCh38
rs943529676 Chr.3: 48694866-48694866 on Build GRCh38
rs753759928 Chr.3: 48694703-48694703 on Build GRCh38
rs73830476 Chr.3: 48694599-48694599 on Build GRCh38
rs957292846 Chr.3: 48694711-48694711 on Build GRCh38
rs11557629 Chr.3: 48694790-48694790 on Build GRCh38
rs1024102586 Chr.3: 48694686-48694686 on Build GRCh38
rs928982533 Chr.3: 48694597-48694597 on Build GRCh38
rs768385005 Chr.3: 48694573-48694573 on Build GRCh38
rs888525094 Chr.3: 48694812-48694812 on Build GRCh38
rs533664278 Chr.3: 48694895-48694895 on Build GRCh38
rs548510443 Chr.3: 48694687-48694687 on Build GRCh38
rs529397870 Chr.3: 48694813-48694813 on Build GRCh38

View Full Product Details