Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48694885-48695384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K2
Entrez Gene ID
51447
Gene Name
inositol hexakisphosphate kinase 2
Gene Aliases
IHPK2, PIUS
UniGene
Hs.595983
Gene Chromosome Location
Chr.3: 48688003-48717278 on Build GRCh38
Transcripts
NM_001005909.2 NM_001005910.2 NM_001005911.2 NM_001146178.2 NM_001146179.2 NM_001190316.1 NM_001190317.1 NM_016291.3 NR_027437.2 NR_027438.2 XM_006713199.3 XM_006713200.1 XM_006713201.1 XM_006713202.1 XM_011533816.1 XM_011533817.1 XM_011533818.1 XM_011533822.1 XM_011533823.1 XM_017006583.1 XM_017006584.1 XM_017006585.1 XM_017006586.1 XM_017006587.1 XM_017006588.1 XM_017006589.1 XM_017006590.1 XM_017006591.1 XM_017006592.1 XM_017006593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6686610 c.94G>A p.V32M Chr.3: 48695198-48695198 on Build GRCh38
5726687 c.24G>A p.M8I Chr.3: 48695268-48695268 on Build GRCh38
7168959 c.186C>T p.F62F Chr.3: 48695106-48695106 on Build GRCh38
3595121 c.46G>A p.G16S Chr.3: 48695246-48695246 on Build GRCh38
7284584 c.78C>T p.V26V Chr.3: 48695214-48695214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887416560 Chr.3: 48694951-48694951 on Build GRCh38
rs750875000 Chr.3: 48695048-48695050 on Build GRCh38
rs368729339 Chr.3: 48695059-48695059 on Build GRCh38
rs759340203 Chr.3: 48695254-48695254 on Build GRCh38
rs772019329 Chr.3: 48694944-48694944 on Build GRCh38
rs57768168 Chr.3: 48695243-48695243 on Build GRCh38
rs757512024 Chr.3: 48695045-48695045 on Build GRCh38
rs779815577 Chr.3: 48694956-48694956 on Build GRCh38
rs567177917 Chr.3: 48695022-48695022 on Build GRCh38
rs879086979 Chr.3: 48694916-48694916 on Build GRCh38
rs146702068 Chr.3: 48695251-48695251 on Build GRCh38
rs766151601 Chr.3: 48695177-48695177 on Build GRCh38
rs148012691 Chr.3: 48695315-48695315 on Build GRCh38
rs372961390 Chr.3: 48695253-48695253 on Build GRCh38
rs753369478 Chr.3: 48695281-48695281 on Build GRCh38
rs776189556 Chr.3: 48694980-48694980 on Build GRCh38
rs761795320 Chr.3: 48695038-48695039 on Build GRCh38
rs770251392 Chr.3: 48695306-48695306 on Build GRCh38
rs769006659 Chr.3: 48695297-48695297 on Build GRCh38
rs752626041 Chr.3: 48695256-48695256 on Build GRCh38
rs965662769 Chr.3: 48695048-48695048 on Build GRCh38
rs753082181 Chr.3: 48694997-48694997 on Build GRCh38
rs777584179 Chr.3: 48695192-48695192 on Build GRCh38
rs766598024 Chr.3: 48695252-48695252 on Build GRCh38
rs951013834 Chr.3: 48695235-48695235 on Build GRCh38
rs753311097 Chr.3: 48695348-48695348 on Build GRCh38
rs756752514 Chr.3: 48695351-48695351 on Build GRCh38
rs200668427 Chr.3: 48694930-48694930 on Build GRCh38
rs750176207 Chr.3: 48694986-48694986 on Build GRCh38
rs769017739 Chr.3: 48694986-48694987 on Build GRCh38
rs201967339 Chr.3: 48695330-48695330 on Build GRCh38
rs778746899 Chr.3: 48694926-48694926 on Build GRCh38
rs758231437 Chr.3: 48695035-48695035 on Build GRCh38
rs372064416 Chr.3: 48695110-48695110 on Build GRCh38
rs772962383 Chr.3: 48695077-48695077 on Build GRCh38
rs774900608 Chr.3: 48694984-48694984 on Build GRCh38
rs748720088 Chr.3: 48695301-48695301 on Build GRCh38
rs760824854 Chr.3: 48695008-48695008 on Build GRCh38
rs755623255 Chr.3: 48695191-48695191 on Build GRCh38
rs866449293 Chr.3: 48695309-48695309 on Build GRCh38
rs540601199 Chr.3: 48695292-48695292 on Build GRCh38
rs941465286 Chr.3: 48695130-48695130 on Build GRCh38
rs754446508 Chr.3: 48695310-48695310 on Build GRCh38
rs11557631 Chr.3: 48695221-48695221 on Build GRCh38
rs761841215 Chr.3: 48695123-48695123 on Build GRCh38
rs756111244 Chr.3: 48695257-48695257 on Build GRCh38
rs754935195 Chr.3: 48695062-48695062 on Build GRCh38
rs139349177 Chr.3: 48695189-48695189 on Build GRCh38
rs905728889 Chr.3: 48694994-48694994 on Build GRCh38
rs780602870 Chr.3: 48695066-48695066 on Build GRCh38
rs759267272 Chr.3: 48695329-48695329 on Build GRCh38
rs776376522 Chr.3: 48694983-48694983 on Build GRCh38
rs755701022 Chr.3: 48694957-48694957 on Build GRCh38
rs764102951 Chr.3: 48695345-48695345 on Build GRCh38
rs773272512 Chr.3: 48695244-48695244 on Build GRCh38
rs774570078 Chr.3: 48695328-48695328 on Build GRCh38
rs192057551 Chr.3: 48695226-48695226 on Build GRCh38
rs1044156395 Chr.3: 48694975-48694975 on Build GRCh38
rs757981572 Chr.3: 48695355-48695355 on Build GRCh38
rs760736385 Chr.3: 48695340-48695340 on Build GRCh38
rs375230535 Chr.3: 48695195-48695195 on Build GRCh38
rs747377400 Chr.3: 48695202-48695202 on Build GRCh38
rs778442720 Chr.3: 48695352-48695352 on Build GRCh38
rs750584549 Chr.3: 48695148-48695148 on Build GRCh38
rs760932933 Chr.3: 48694958-48694960 on Build GRCh38
rs765423817 Chr.3: 48695049-48695049 on Build GRCh38
rs17080319 Chr.3: 48695047-48695047 on Build GRCh38
rs779631475 Chr.3: 48694917-48694918 on Build GRCh38
rs765448591 Chr.3: 48695136-48695136 on Build GRCh38
rs746532621 Chr.3: 48694963-48694963 on Build GRCh38
rs548722165 Chr.3: 48695067-48695067 on Build GRCh38
rs969879349 Chr.3: 48694942-48694942 on Build GRCh38
rs774322013 Chr.3: 48694955-48694957 on Build GRCh38
rs111397089 Chr.3: 48695185-48695185 on Build GRCh38
rs776709547 Chr.3: 48695037-48695045 on Build GRCh38
rs765416158 Chr.3: 48695040-48695041 on Build GRCh38
rs750247896 Chr.3: 48695053-48695053 on Build GRCh38
rs527477360 Chr.3: 48694981-48694981 on Build GRCh38
rs764632711 Chr.3: 48695113-48695113 on Build GRCh38
rs768173933 Chr.3: 48694989-48694989 on Build GRCh38
rs369490009 Chr.3: 48695199-48695199 on Build GRCh38
rs752539825 Chr.3: 48695339-48695339 on Build GRCh38
rs758121580 Chr.3: 48695054-48695054 on Build GRCh38
rs746242061 Chr.3: 48695078-48695078 on Build GRCh38
rs148477592 Chr.3: 48695214-48695214 on Build GRCh38
rs758603715 Chr.3: 48695159-48695159 on Build GRCh38
rs200143726 Chr.3: 48695300-48695300 on Build GRCh38
rs763491534 Chr.3: 48695263-48695264 on Build GRCh38
rs777494112 Chr.3: 48695268-48695268 on Build GRCh38
rs767991254 Chr.3: 48694971-48694971 on Build GRCh38
rs571065789 Chr.3: 48695196-48695196 on Build GRCh38
rs369011363 Chr.3: 48695296-48695296 on Build GRCh38
rs957141808 Chr.3: 48695029-48695029 on Build GRCh38
rs34911461 Chr.3: 48695131-48695132 on Build GRCh38
rs777055597 Chr.3: 48695223-48695223 on Build GRCh38
rs769596688 Chr.3: 48695076-48695076 on Build GRCh38
rs776740658 Chr.3: 48695114-48695114 on Build GRCh38
rs1049901488 Chr.3: 48695104-48695104 on Build GRCh38
rs968006325 Chr.3: 48695125-48695125 on Build GRCh38
rs756909204 Chr.3: 48695284-48695284 on Build GRCh38
rs780052236 Chr.3: 48695057-48695057 on Build GRCh38
rs772269322 Chr.3: 48695100-48695100 on Build GRCh38
rs201009012 Chr.3: 48695190-48695190 on Build GRCh38
rs748487372 Chr.3: 48695358-48695358 on Build GRCh38
rs750148345 Chr.3: 48695354-48695354 on Build GRCh38
rs141695869 Chr.3: 48695320-48695320 on Build GRCh38
rs374945808 Chr.3: 48695255-48695255 on Build GRCh38

View Full Product Details