Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50299922-50300433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL1
Entrez Gene ID
3373
Gene Name
hyaluronoglucosaminidase 1
Gene Aliases
HYAL-1, LUCA1, MPS9, NAT6
UniGene
Hs.75619
Gene Chromosome Location
Chr.3: 50299889-50312951 on Build GRCh38
Transcripts
NM_033159.3 NM_153281.1 NM_153282.2 NM_153283.2 NM_153285.2 NR_047690.1 XM_011533667.2 XM_011533668.2 XM_011533669.2

SNP Detail

SNP ID Chromosome Location
rs1011385276 Chr.3: 50299993-50299993 on Build GRCh38
rs938035040 Chr.3: 50300290-50300290 on Build GRCh38
rs1041537653 Chr.3: 50300375-50300375 on Build GRCh38
rs1055668304 Chr.3: 50299989-50299989 on Build GRCh38
rs903056291 Chr.3: 50300377-50300377 on Build GRCh38
rs1020047984 Chr.3: 50299995-50299995 on Build GRCh38
rs182987423 Chr.3: 50300049-50300049 on Build GRCh38
rs1283 Chr.3: 50299991-50299991 on Build GRCh38
rs1011415105 Chr.3: 50300347-50300347 on Build GRCh38
rs192692460 Chr.3: 50300262-50300262 on Build GRCh38
rs782409309 Chr.3: 50299945-50299945 on Build GRCh38
rs782042512 Chr.3: 50299988-50299988 on Build GRCh38
rs184872974 Chr.3: 50300310-50300310 on Build GRCh38
rs963599165 Chr.3: 50300090-50300090 on Build GRCh38
rs967164645 Chr.3: 50300003-50300003 on Build GRCh38
rs187533933 Chr.3: 50300250-50300250 on Build GRCh38
rs150976767 Chr.3: 50300024-50300024 on Build GRCh38
rs117698094 Chr.3: 50300301-50300301 on Build GRCh38
rs141677727 Chr.3: 50300098-50300098 on Build GRCh38
rs909486002 Chr.3: 50300088-50300088 on Build GRCh38
rs587729250 Chr.3: 50300047-50300047 on Build GRCh38
rs949498950 Chr.3: 50300159-50300159 on Build GRCh38
rs782198562 Chr.3: 50299944-50299944 on Build GRCh38
rs953195289 Chr.3: 50300041-50300041 on Build GRCh38
rs145434547 Chr.3: 50300332-50300332 on Build GRCh38
rs926715169 Chr.3: 50300263-50300263 on Build GRCh38
rs972237393 Chr.3: 50300121-50300121 on Build GRCh38
rs782422891 Chr.3: 50300265-50300265 on Build GRCh38
rs919391355 Chr.3: 50300123-50300123 on Build GRCh38
rs1030076924 Chr.3: 50300028-50300028 on Build GRCh38

View Full Product Details