Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50301974-50302467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL1
Entrez Gene ID
3373
Gene Name
hyaluronoglucosaminidase 1
Gene Aliases
HYAL-1, LUCA1, MPS9, NAT6
UniGene
Hs.75619
Gene Chromosome Location
Chr.3: 50299889-50312951 on Build GRCh38
Transcripts
NM_033159.3 NM_153281.1 NM_153282.2 NM_153283.2 NM_153285.2 NR_047690.1 XM_011533667.2 XM_011533668.2 XM_011533669.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836048 c.535G>T p.A179S Chr.3: 50302422-50302422 on Build GRCh38
5424304 c.532G>T p.A178S Chr.3: 50302425-50302425 on Build GRCh38
320898 c.528G>T p.Q176H Chr.3: 50302429-50302429 on Build GRCh38
5423303 c.896C>T p.P299L Chr.3: 50302061-50302061 on Build GRCh38
2851277 c.577C>T p.L193L Chr.3: 50302380-50302380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782513063 Chr.3: 50302246-50302246 on Build GRCh38
rs138951582 Chr.3: 50302212-50302212 on Build GRCh38
rs587679732 Chr.3: 50302347-50302347 on Build GRCh38
rs781783393 Chr.3: 50302345-50302345 on Build GRCh38
rs781928640 Chr.3: 50302390-50302390 on Build GRCh38
rs782798734 Chr.3: 50302410-50302410 on Build GRCh38
rs782343493 Chr.3: 50302387-50302387 on Build GRCh38
rs587652432 Chr.3: 50302171-50302171 on Build GRCh38
rs587748533 Chr.3: 50302148-50302148 on Build GRCh38
rs782458770 Chr.3: 50302420-50302420 on Build GRCh38
rs782812048 Chr.3: 50302124-50302124 on Build GRCh38
rs373351472 Chr.3: 50302039-50302039 on Build GRCh38
rs138263782 Chr.3: 50302180-50302180 on Build GRCh38
rs782708747 Chr.3: 50302425-50302425 on Build GRCh38
rs1047509699 Chr.3: 50302014-50302014 on Build GRCh38
rs781943397 Chr.3: 50302186-50302186 on Build GRCh38
rs782348476 Chr.3: 50302301-50302301 on Build GRCh38
rs782601965 Chr.3: 50302369-50302369 on Build GRCh38
rs374928005 Chr.3: 50302357-50302357 on Build GRCh38
rs753549050 Chr.3: 50302140-50302140 on Build GRCh38
rs782758646 Chr.3: 50302324-50302324 on Build GRCh38
rs782608355 Chr.3: 50302253-50302253 on Build GRCh38
rs147106267 Chr.3: 50302385-50302385 on Build GRCh38
rs142160277 Chr.3: 50302209-50302209 on Build GRCh38
rs782564609 Chr.3: 50302275-50302275 on Build GRCh38
rs781785872 Chr.3: 50302025-50302025 on Build GRCh38
rs782385337 Chr.3: 50302107-50302107 on Build GRCh38
rs782701738 Chr.3: 50302022-50302022 on Build GRCh38
rs1018254791 Chr.3: 50302164-50302164 on Build GRCh38
rs781976895 Chr.3: 50302213-50302213 on Build GRCh38
rs782012779 Chr.3: 50302414-50302414 on Build GRCh38
rs782094366 Chr.3: 50302111-50302111 on Build GRCh38
rs143791794 Chr.3: 50302386-50302386 on Build GRCh38
rs781796869 Chr.3: 50302231-50302231 on Build GRCh38
rs1011078230 Chr.3: 50302057-50302057 on Build GRCh38
rs587670275 Chr.3: 50302419-50302419 on Build GRCh38
rs782028491 Chr.3: 50302091-50302091 on Build GRCh38
rs781834437 Chr.3: 50302051-50302052 on Build GRCh38
rs143176318 Chr.3: 50302278-50302278 on Build GRCh38
rs782042919 Chr.3: 50302393-50302393 on Build GRCh38
rs141022886 Chr.3: 50302093-50302093 on Build GRCh38
rs781980498 Chr.3: 50302110-50302110 on Build GRCh38
rs781873681 Chr.3: 50302127-50302127 on Build GRCh38
rs782086155 Chr.3: 50302222-50302222 on Build GRCh38
rs202165326 Chr.3: 50302368-50302368 on Build GRCh38
rs782189959 Chr.3: 50302189-50302189 on Build GRCh38
rs150255984 Chr.3: 50302145-50302145 on Build GRCh38
rs782299729 Chr.3: 50302161-50302161 on Build GRCh38
rs916207090 Chr.3: 50302308-50302308 on Build GRCh38
rs781838448 Chr.3: 50302327-50302327 on Build GRCh38
rs782023654 Chr.3: 50302317-50302317 on Build GRCh38
rs782793045 Chr.3: 50302238-50302238 on Build GRCh38
rs782533590 Chr.3: 50302034-50302034 on Build GRCh38
rs782285336 Chr.3: 50302445-50302446 on Build GRCh38
rs782692817 Chr.3: 50302376-50302376 on Build GRCh38
rs782203268 Chr.3: 50302079-50302079 on Build GRCh38
rs782292968 Chr.3: 50302187-50302187 on Build GRCh38
rs374889084 Chr.3: 50302294-50302294 on Build GRCh38
rs782051318 Chr.3: 50302303-50302303 on Build GRCh38
rs782602522 Chr.3: 50302073-50302073 on Build GRCh38
rs782722149 Chr.3: 50302342-50302342 on Build GRCh38
rs149384463 Chr.3: 50302427-50302427 on Build GRCh38
rs782428159 Chr.3: 50302305-50302305 on Build GRCh38
rs782313024 Chr.3: 50302085-50302085 on Build GRCh38
rs902728091 Chr.3: 50302104-50302104 on Build GRCh38
rs782275827 Chr.3: 50302377-50302377 on Build GRCh38
rs781935297 Chr.3: 50302302-50302302 on Build GRCh38
rs929130961 Chr.3: 50302012-50302012 on Build GRCh38
rs782752459 Chr.3: 50302146-50302146 on Build GRCh38
rs200718831 Chr.3: 50302258-50302258 on Build GRCh38
rs782132370 Chr.3: 50302320-50302320 on Build GRCh38
rs782525991 Chr.3: 50302163-50302163 on Build GRCh38
rs141770421 Chr.3: 50302371-50302371 on Build GRCh38
rs782643536 Chr.3: 50302440-50302440 on Build GRCh38
rs782655370 Chr.3: 50302165-50302165 on Build GRCh38
rs149438911 Chr.3: 50302156-50302156 on Build GRCh38
rs782389049 Chr.3: 50302384-50302384 on Build GRCh38
rs370585179 Chr.3: 50302281-50302281 on Build GRCh38
rs782170292 Chr.3: 50302236-50302236 on Build GRCh38
rs782240278 Chr.3: 50302170-50302170 on Build GRCh38
rs376502179 Chr.3: 50302223-50302223 on Build GRCh38
rs104893743 Chr.3: 50302155-50302155 on Build GRCh38
rs148862952 Chr.3: 50302348-50302348 on Build GRCh38
rs782200654 Chr.3: 50302370-50302370 on Build GRCh38
rs139187462 Chr.3: 50302210-50302210 on Build GRCh38
rs782355058 Chr.3: 50302184-50302184 on Build GRCh38
rs782751421 Chr.3: 50302418-50302418 on Build GRCh38
rs878328 Chr.3: 50302109-50302109 on Build GRCh38
rs199678542 Chr.3: 50302352-50302352 on Build GRCh38
rs782256106 Chr.3: 50302046-50302046 on Build GRCh38
rs939067401 Chr.3: 50302288-50302288 on Build GRCh38
rs782723000 Chr.3: 50302112-50302112 on Build GRCh38
rs376026547 Chr.3: 50302415-50302415 on Build GRCh38
rs1029776116 Chr.3: 50302121-50302121 on Build GRCh38
rs782395202 Chr.3: 50302280-50302280 on Build GRCh38
rs587665493 Chr.3: 50302239-50302239 on Build GRCh38
rs116482870 Chr.3: 50302191-50302191 on Build GRCh38
rs587658260 Chr.3: 50302071-50302071 on Build GRCh38
rs781902076 Chr.3: 50302434-50302434 on Build GRCh38
rs782041421 Chr.3: 50302123-50302123 on Build GRCh38
rs781835664 Chr.3: 50302008-50302008 on Build GRCh38
rs782079716 Chr.3: 50302339-50302339 on Build GRCh38
rs781932281 Chr.3: 50302118-50302118 on Build GRCh38

View Full Product Details