Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52794667-52795207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH3
Entrez Gene ID
3699
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Gene Aliases
H3P, ITI-HC3, SHAP
UniGene
Hs.76716
Gene Chromosome Location
Chr.3: 52794768-52809009 on Build GRCh38
Transcripts
NM_002217.3 XM_005265105.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1255064 c.83G>A p.R28Q Chr.3: 52794886-52794886 on Build GRCh38
1255063 c.83G>A p.R28Q Chr.3: 52794886-52794886 on Build GRCh38
4572147 c.65T>G p.F22C Chr.3: 52794868-52794868 on Build GRCh38
6877289 c.45C>T p.S15S Chr.3: 52794848-52794848 on Build GRCh38
2780231 c.4G>A p.A2T Chr.3: 52794807-52794807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997877910 Chr.3: 52794986-52794986 on Build GRCh38
rs756489463 Chr.3: 52794925-52794925 on Build GRCh38
rs747815332 Chr.3: 52794804-52794804 on Build GRCh38
rs560024879 Chr.3: 52794850-52794850 on Build GRCh38
rs546733679 Chr.3: 52794816-52794816 on Build GRCh38
rs567756674 Chr.3: 52794906-52794906 on Build GRCh38
rs531503965 Chr.3: 52794950-52794950 on Build GRCh38
rs888746005 Chr.3: 52795005-52795005 on Build GRCh38
rs989888608 Chr.3: 52794736-52794736 on Build GRCh38
rs750914193 Chr.3: 52794907-52794907 on Build GRCh38
rs994191768 Chr.3: 52794748-52794748 on Build GRCh38
rs757911598 Chr.3: 52794815-52794815 on Build GRCh38
rs1021304022 Chr.3: 52795062-52795062 on Build GRCh38
rs201110320 Chr.3: 52794803-52794803 on Build GRCh38
rs960974084 Chr.3: 52795123-52795123 on Build GRCh38
rs373235618 Chr.3: 52794825-52794825 on Build GRCh38
rs1005654623 Chr.3: 52794800-52794800 on Build GRCh38
rs916597070 Chr.3: 52794811-52794811 on Build GRCh38
rs923836220 Chr.3: 52794948-52794948 on Build GRCh38
rs576702487 Chr.3: 52794725-52794725 on Build GRCh38
rs554112830 Chr.3: 52795047-52795047 on Build GRCh38
rs528961799 Chr.3: 52794871-52794871 on Build GRCh38
rs990952091 Chr.3: 52794874-52794874 on Build GRCh38
rs756577207 Chr.3: 52794802-52794802 on Build GRCh38
rs901762869 Chr.3: 52794692-52794692 on Build GRCh38
rs969346136 Chr.3: 52795157-52795157 on Build GRCh38
rs564076595 Chr.3: 52794779-52794779 on Build GRCh38
rs754420593 Chr.3: 52794884-52794884 on Build GRCh38
rs766854942 Chr.3: 52794928-52794928 on Build GRCh38
rs775723113 Chr.3: 52794764-52794764 on Build GRCh38
rs377359593 Chr.3: 52794984-52794984 on Build GRCh38
rs546861404 Chr.3: 52795085-52795085 on Build GRCh38
rs567699397 Chr.3: 52795056-52795056 on Build GRCh38
rs568992581 Chr.3: 52794933-52794933 on Build GRCh38
rs948144050 Chr.3: 52794832-52794832 on Build GRCh38
rs759295905 Chr.3: 52794760-52794760 on Build GRCh38
rs140701550 Chr.3: 52795165-52795165 on Build GRCh38
rs916854978 Chr.3: 52795147-52795147 on Build GRCh38
rs768027400 Chr.3: 52794886-52794886 on Build GRCh38
rs548955356 Chr.3: 52794872-52794872 on Build GRCh38
rs778956898 Chr.3: 52794919-52794919 on Build GRCh38
rs762975686 Chr.3: 52794765-52794765 on Build GRCh38
rs746556196 Chr.3: 52794831-52794831 on Build GRCh38
rs549449651 Chr.3: 52794769-52794769 on Build GRCh38
rs776470753 Chr.3: 52794755-52794755 on Build GRCh38
rs147337675 Chr.3: 52794724-52794724 on Build GRCh38
rs770875847 Chr.3: 52794833-52794833 on Build GRCh38
rs1014818961 Chr.3: 52794691-52794691 on Build GRCh38
rs774478674 Chr.3: 52794876-52794876 on Build GRCh38
rs1047072881 Chr.3: 52795099-52795099 on Build GRCh38
rs556492051 Chr.3: 52795128-52795128 on Build GRCh38
rs1031628071 Chr.3: 52795091-52795091 on Build GRCh38
rs992842520 Chr.3: 52795146-52795146 on Build GRCh38
rs936487382 Chr.3: 52794690-52794690 on Build GRCh38
rs533963695 Chr.3: 52795046-52795046 on Build GRCh38
rs202203995 Chr.3: 52794778-52794778 on Build GRCh38
rs901131322 Chr.3: 52794743-52794743 on Build GRCh38
rs545740325 Chr.3: 52795167-52795167 on Build GRCh38
rs78407765 Chr.3: 52794726-52794726 on Build GRCh38
rs376872714 Chr.3: 52794885-52794885 on Build GRCh38
rs765417629 Chr.3: 52794761-52794761 on Build GRCh38
rs997150527 Chr.3: 52794716-52794717 on Build GRCh38
rs911035383 Chr.3: 52795041-52795041 on Build GRCh38
rs775358463 Chr.3: 52794826-52794826 on Build GRCh38
rs978243433 Chr.3: 52794767-52794767 on Build GRCh38
rs756853240 Chr.3: 52794906-52794906 on Build GRCh38
rs368987550 Chr.3: 52794789-52794789 on Build GRCh38
rs1053642899 Chr.3: 52794721-52794721 on Build GRCh38
rs763927280 Chr.3: 52795042-52795042 on Build GRCh38
rs775635272 Chr.3: 52794868-52794868 on Build GRCh38
rs745624330 Chr.3: 52794853-52794853 on Build GRCh38
rs751633939 Chr.3: 52795172-52795172 on Build GRCh38
rs999611570 Chr.3: 52795090-52795090 on Build GRCh38
rs7629493 Chr.3: 52794952-52794952 on Build GRCh38
rs1041155974 Chr.3: 52794740-52794740 on Build GRCh38
rs769603753 Chr.3: 52794861-52794861 on Build GRCh38

View Full Product Details