Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52797724-52798272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH3
Entrez Gene ID
3699
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Gene Aliases
H3P, ITI-HC3, SHAP
UniGene
Hs.76716
Gene Chromosome Location
Chr.3: 52794768-52809009 on Build GRCh38
Transcripts
NM_002217.3 XM_005265105.4 XM_017006356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1291749 c.658delA p.K221fs*10 Chr.3: 52797925-52797925 on Build GRCh38
7331347 c.609C>A p.F203L Chr.3: 52797876-52797876 on Build GRCh38
1211070 c.651C>A p.F217L Chr.3: 52797918-52797918 on Build GRCh38
1211069 c.651C>A p.F217L Chr.3: 52797918-52797918 on Build GRCh38
2780251 c.655G>T p.G219W Chr.3: 52797922-52797922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368997998 Chr.3: 52798088-52798089 on Build GRCh38
rs749480189 Chr.3: 52797935-52797935 on Build GRCh38
rs780953381 Chr.3: 52797819-52797819 on Build GRCh38
rs756518754 Chr.3: 52797921-52797921 on Build GRCh38
rs58960890 Chr.3: 52797991-52797991 on Build GRCh38
rs892200692 Chr.3: 52798070-52798070 on Build GRCh38
rs745713888 Chr.3: 52797820-52797820 on Build GRCh38
rs745623976 Chr.3: 52797924-52797924 on Build GRCh38
rs757857469 Chr.3: 52797901-52797901 on Build GRCh38
rs894731891 Chr.3: 52798194-52798194 on Build GRCh38
rs750728076 Chr.3: 52798149-52798149 on Build GRCh38
rs769599746 Chr.3: 52797930-52797930 on Build GRCh38
rs772665013 Chr.3: 52797967-52797967 on Build GRCh38
rs749158060 Chr.3: 52797834-52797834 on Build GRCh38
rs921761354 Chr.3: 52797828-52797828 on Build GRCh38
rs920572535 Chr.3: 52798246-52798246 on Build GRCh38
rs559478524 Chr.3: 52797876-52797876 on Build GRCh38
rs750783492 Chr.3: 52797881-52797881 on Build GRCh38
rs62255414 Chr.3: 52798094-52798094 on Build GRCh38
rs751136891 Chr.3: 52797911-52797911 on Build GRCh38
rs748275884 Chr.3: 52797944-52797944 on Build GRCh38
rs768992367 Chr.3: 52797937-52797937 on Build GRCh38
rs772172934 Chr.3: 52797867-52797867 on Build GRCh38
rs533443123 Chr.3: 52798089-52798089 on Build GRCh38
rs1053461019 Chr.3: 52798059-52798059 on Build GRCh38
rs776717948 Chr.3: 52798004-52798004 on Build GRCh38
rs565501270 Chr.3: 52798244-52798244 on Build GRCh38
rs550747222 Chr.3: 52797975-52797975 on Build GRCh38
rs763793671 Chr.3: 52797925-52797925 on Build GRCh38
rs368383671 Chr.3: 52797885-52797885 on Build GRCh38
rs762189975 Chr.3: 52798031-52798031 on Build GRCh38
rs774283833 Chr.3: 52797942-52797942 on Build GRCh38
rs746924022 Chr.3: 52797803-52797803 on Build GRCh38
rs774858257 Chr.3: 52797855-52797855 on Build GRCh38
rs139300508 Chr.3: 52797845-52797845 on Build GRCh38
rs904203714 Chr.3: 52798152-52798152 on Build GRCh38
rs945172477 Chr.3: 52798072-52798072 on Build GRCh38
rs757865045 Chr.3: 52797782-52797782 on Build GRCh38
rs760894658 Chr.3: 52797912-52797912 on Build GRCh38
rs114325431 Chr.3: 52797914-52797914 on Build GRCh38
rs528595557 Chr.3: 52797884-52797884 on Build GRCh38
rs752207891 Chr.3: 52797768-52797768 on Build GRCh38
rs552194543 Chr.3: 52798165-52798165 on Build GRCh38
rs372440070 Chr.3: 52797886-52797886 on Build GRCh38
rs1045746634 Chr.3: 52797923-52797923 on Build GRCh38
rs763430800 Chr.3: 52798222-52798222 on Build GRCh38
rs780269806 Chr.3: 52797933-52797933 on Build GRCh38
rs1027021537 Chr.3: 52798223-52798223 on Build GRCh38
rs113350909 Chr.3: 52798090-52798090 on Build GRCh38
rs907432852 Chr.3: 52797754-52797754 on Build GRCh38
rs762224522 Chr.3: 52797866-52797866 on Build GRCh38
rs972137796 Chr.3: 52798245-52798245 on Build GRCh38
rs936308368 Chr.3: 52798048-52798048 on Build GRCh38
rs777708244 Chr.3: 52797793-52797793 on Build GRCh38
rs953228686 Chr.3: 52798210-52798210 on Build GRCh38
rs921194941 Chr.3: 52798007-52798007 on Build GRCh38
rs1029330542 Chr.3: 52798167-52798167 on Build GRCh38
rs968692654 Chr.3: 52797764-52797764 on Build GRCh38
rs895592158 Chr.3: 52798057-52798057 on Build GRCh38
rs371603059 Chr.3: 52797977-52797977 on Build GRCh38
rs768184448 Chr.3: 52797873-52797873 on Build GRCh38
rs34352685 Chr.3: 52797900-52797900 on Build GRCh38
rs1018968899 Chr.3: 52798231-52798231 on Build GRCh38
rs920833435 Chr.3: 52797913-52797913 on Build GRCh38
rs770919801 Chr.3: 52797810-52797810 on Build GRCh38
rs752941232 Chr.3: 52797858-52797858 on Build GRCh38
rs769972968 Chr.3: 52797824-52797824 on Build GRCh38
rs979097924 Chr.3: 52797773-52797773 on Build GRCh38
rs562327889 Chr.3: 52797910-52797910 on Build GRCh38
rs763043771 Chr.3: 52797835-52797835 on Build GRCh38
rs765809669 Chr.3: 52798086-52798086 on Build GRCh38
rs940694512 Chr.3: 52797909-52797909 on Build GRCh38
rs143340291 Chr.3: 52797941-52797941 on Build GRCh38

View Full Product Details