Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58491895-58492423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD6
Entrez Gene ID
200845
Gene Name
potassium channel tetramerization domain containing 6
Gene Aliases
KCASH3
UniGene
Hs.13982
Gene Chromosome Location
Chr.3: 58492096-58502360 on Build GRCh38
Transcripts
NM_001128214.1

SNP Detail

SNP ID Chromosome Location
rs929630760 Chr.3: 58492159-58492159 on Build GRCh38
rs958826121 Chr.3: 58492381-58492381 on Build GRCh38
rs1025358970 Chr.3: 58492022-58492022 on Build GRCh38
rs1005319061 Chr.3: 58492031-58492031 on Build GRCh38
rs111898477 Chr.3: 58492099-58492099 on Build GRCh38
rs891962740 Chr.3: 58491914-58491914 on Build GRCh38
rs994240299 Chr.3: 58492008-58492008 on Build GRCh38
rs1047188092 Chr.3: 58492186-58492187 on Build GRCh38
rs539941124 Chr.3: 58492333-58492333 on Build GRCh38
rs927479104 Chr.3: 58492140-58492140 on Build GRCh38
rs1029550372 Chr.3: 58492301-58492301 on Build GRCh38
rs577485003 Chr.3: 58492231-58492231 on Build GRCh38
rs989229668 Chr.3: 58492096-58492096 on Build GRCh38
rs1018058235 Chr.3: 58492225-58492225 on Build GRCh38
rs1043263586 Chr.3: 58491939-58491939 on Build GRCh38
rs553461206 Chr.3: 58492404-58492404 on Build GRCh38
rs937574375 Chr.3: 58492145-58492145 on Build GRCh38
rs866053485 Chr.3: 58492067-58492067 on Build GRCh38
rs184647973 Chr.3: 58492026-58492026 on Build GRCh38
rs1018087832 Chr.3: 58492058-58492058 on Build GRCh38
rs1020324496 Chr.3: 58492331-58492331 on Build GRCh38
rs947474209 Chr.3: 58492127-58492127 on Build GRCh38
rs764323627 Chr.3: 58492363-58492363 on Build GRCh38
rs1057344926 Chr.3: 58492152-58492153 on Build GRCh38
rs978910277 Chr.3: 58492128-58492128 on Build GRCh38
rs895521024 Chr.3: 58492006-58492006 on Build GRCh38
rs906093742 Chr.3: 58491985-58491985 on Build GRCh38
rs947505980 Chr.3: 58491926-58491926 on Build GRCh38
rs557603709 Chr.3: 58492229-58492229 on Build GRCh38
rs913313128 Chr.3: 58491925-58491925 on Build GRCh38
rs1030746991 Chr.3: 58492072-58492072 on Build GRCh38
rs1053248848 Chr.3: 58491921-58491921 on Build GRCh38
rs896063811 Chr.3: 58492158-58492158 on Build GRCh38
rs963917851 Chr.3: 58492064-58492064 on Build GRCh38
rs535701651 Chr.3: 58492366-58492376 on Build GRCh38
rs937605450 Chr.3: 58492001-58492001 on Build GRCh38
rs1057193486 Chr.3: 58492002-58492002 on Build GRCh38
rs1005289701 Chr.3: 58492211-58492211 on Build GRCh38
rs956851626 Chr.3: 58492302-58492302 on Build GRCh38
rs954696678 Chr.3: 58492073-58492074 on Build GRCh38
rs927364549 Chr.3: 58492379-58492379 on Build GRCh38
rs998430960 Chr.3: 58492268-58492268 on Build GRCh38
rs988808625 Chr.3: 58492305-58492305 on Build GRCh38
rs537325348 Chr.3: 58492065-58492065 on Build GRCh38
rs888178090 Chr.3: 58492190-58492190 on Build GRCh38

View Full Product Details